Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 36999123-36999618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG5
Entrez Gene ID
29880
Gene Name
ALG5, dolichyl-phosphate beta-glucosyltransferase
Gene Aliases
bA421P11.2
UniGene
Hs.507769
Gene Chromosome Location
Chr.13: 36949770-36999367 on Build GRCh38
Transcripts
NM_001142364.1 NM_013338.4 XM_017020552.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1238170 c.10C>T p.L4F Chr.13: 36999291-36999291 on Build GRCh38
6553912 c.49G>A p.A17T Chr.13: 36999252-36999252 on Build GRCh38
3468678 c.60C>T p.L20L Chr.13: 36999241-36999241 on Build GRCh38
696646 c.24G>A p.L8L Chr.13: 36999277-36999277 on Build GRCh38
7150502 c.41C>T p.A14V Chr.13: 36999260-36999260 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1015008785 Chr.13: 36999178-36999178 on Build GRCh38
rs565406391 Chr.13: 36999357-36999357 on Build GRCh38
rs373213937 Chr.13: 36999188-36999188 on Build GRCh38
rs767613936 Chr.13: 36999303-36999303 on Build GRCh38
rs1036630059 Chr.13: 36999367-36999367 on Build GRCh38
rs886282036 Chr.13: 36999494-36999494 on Build GRCh38
rs574584993 Chr.13: 36999271-36999271 on Build GRCh38
rs535486849 Chr.13: 36999294-36999294 on Build GRCh38
rs770491390 Chr.13: 36999330-36999330 on Build GRCh38
rs560115384 Chr.13: 36999548-36999548 on Build GRCh38
rs775475124 Chr.13: 36999338-36999338 on Build GRCh38
rs749775352 Chr.13: 36999227-36999227 on Build GRCh38
rs905312858 Chr.13: 36999400-36999400 on Build GRCh38
rs956371282 Chr.13: 36999355-36999355 on Build GRCh38
rs200894954 Chr.13: 36999226-36999226 on Build GRCh38
rs772633936 Chr.13: 36999289-36999289 on Build GRCh38
rs940840484 Chr.13: 36999544-36999545 on Build GRCh38
rs755285334 Chr.13: 36999314-36999314 on Build GRCh38
rs755340077 Chr.13: 36999275-36999275 on Build GRCh38
rs1037493134 Chr.13: 36999524-36999524 on Build GRCh38
rs748677219 Chr.13: 36999283-36999283 on Build GRCh38
rs759530016 Chr.13: 36999250-36999250 on Build GRCh38
rs368835581 Chr.13: 36999307-36999307 on Build GRCh38
rs975150375 Chr.13: 36999329-36999329 on Build GRCh38
rs1028683056 Chr.13: 36999565-36999565 on Build GRCh38
rs376280362 Chr.13: 36999315-36999315 on Build GRCh38
rs779005586 Chr.13: 36999193-36999193 on Build GRCh38
rs892320710 Chr.13: 36999413-36999413 on Build GRCh38
rs770743615 Chr.13: 36999216-36999216 on Build GRCh38
rs540943786 Chr.13: 36999322-36999322 on Build GRCh38
rs147574138 Chr.13: 36999270-36999270 on Build GRCh38
rs988946400 Chr.13: 36999374-36999374 on Build GRCh38
rs771496374 Chr.13: 36999238-36999238 on Build GRCh38
rs757220549 Chr.13: 36999328-36999328 on Build GRCh38
rs776395229 Chr.13: 36999366-36999366 on Build GRCh38
rs759139581 Chr.13: 36999409-36999409 on Build GRCh38
rs535981612 Chr.13: 36999176-36999176 on Build GRCh38
rs779333644 Chr.13: 36999282-36999282 on Build GRCh38
rs757367488 Chr.13: 36999261-36999261 on Build GRCh38
rs766467895 Chr.13: 36999352-36999352 on Build GRCh38
rs756364079 Chr.13: 36999249-36999250 on Build GRCh38
rs761443993 Chr.13: 36999184-36999184 on Build GRCh38
rs890088314 Chr.13: 36999583-36999583 on Build GRCh38
rs374546822 Chr.13: 36999234-36999234 on Build GRCh38
rs374355673 Chr.13: 36999306-36999306 on Build GRCh38
rs769684716 Chr.13: 36999333-36999333 on Build GRCh38
rs757806780 Chr.13: 36999314-36999323 on Build GRCh38
rs372353224 Chr.13: 36999345-36999345 on Build GRCh38
rs758874777 Chr.13: 36999321-36999321 on Build GRCh38
rs577401807 Chr.13: 36999395-36999395 on Build GRCh38
rs767528777 Chr.13: 36999263-36999263 on Build GRCh38
rs566465782 Chr.13: 36999232-36999232 on Build GRCh38
rs898890874 Chr.13: 36999536-36999536 on Build GRCh38
rs940509405 Chr.13: 36999498-36999498 on Build GRCh38
rs762935307 Chr.13: 36999342-36999342 on Build GRCh38
rs899124027 Chr.13: 36999369-36999369 on Build GRCh38
rs757420403 Chr.13: 36999214-36999214 on Build GRCh38
rs764244031 Chr.13: 36999194-36999194 on Build GRCh38
rs776337687 Chr.13: 36999248-36999248 on Build GRCh38
rs754071553 Chr.13: 36999312-36999312 on Build GRCh38
rs530123284 Chr.13: 36999450-36999450 on Build GRCh38
rs955328752 Chr.13: 36999491-36999491 on Build GRCh38
rs914639019 Chr.13: 36999376-36999376 on Build GRCh38
rs750528515 Chr.13: 36999305-36999305 on Build GRCh38
rs746017779 Chr.13: 36999324-36999325 on Build GRCh38
rs746649079 Chr.13: 36999291-36999291 on Build GRCh38
rs571901428 Chr.13: 36999311-36999311 on Build GRCh38
rs777747414 Chr.13: 36999560-36999560 on Build GRCh38
rs765149659 Chr.13: 36999296-36999296 on Build GRCh38
rs371119626 Chr.13: 36999302-36999302 on Build GRCh38
rs986579414 Chr.13: 36999496-36999496 on Build GRCh38
rs1028054051 Chr.13: 36999469-36999469 on Build GRCh38
rs747588147 Chr.13: 36999235-36999235 on Build GRCh38
rs770544173 Chr.13: 36999292-36999292 on Build GRCh38
rs886248536 Chr.13: 36999266-36999266 on Build GRCh38
rs921051692 Chr.13: 36999340-36999340 on Build GRCh38
rs4293244 Chr.13: 36999488-36999488 on Build GRCh38
rs779280718 Chr.13: 36999320-36999320 on Build GRCh38
rs767222637 Chr.13: 36999187-36999187 on Build GRCh38
rs772872038 Chr.13: 36999243-36999243 on Build GRCh38
rs754246428 Chr.13: 36999280-36999310 on Build GRCh38
rs377460720 Chr.13: 36999193-36999193 on Build GRCh38
rs146902400 Chr.13: 36999265-36999265 on Build GRCh38
rs1004637094 Chr.13: 36999268-36999268 on Build GRCh38
rs562688195 Chr.13: 36999436-36999436 on Build GRCh38
rs753765291 Chr.13: 36999191-36999191 on Build GRCh38
rs1003827976 Chr.13: 36999310-36999310 on Build GRCh38
rs534101761 Chr.13: 36999245-36999245 on Build GRCh38
rs760612138 Chr.13: 36999348-36999348 on Build GRCh38
rs1003020471 Chr.13: 36999465-36999484 on Build GRCh38
rs778228464 Chr.13: 36999252-36999272 on Build GRCh38
rs371089333 Chr.13: 36999485-36999485 on Build GRCh38
rs967680292 Chr.13: 36999453-36999453 on Build GRCh38
rs544377113 Chr.13: 36999412-36999412 on Build GRCh38
rs780945745 Chr.13: 36999331-36999331 on Build GRCh38
rs751759577 Chr.13: 36999203-36999203 on Build GRCh38
rs561093528 Chr.13: 36999256-36999256 on Build GRCh38
rs771566757 Chr.13: 36999286-36999286 on Build GRCh38
rs775532363 Chr.13: 36999297-36999297 on Build GRCh38
rs781422130 Chr.13: 36999215-36999215 on Build GRCh38
rs376568505 Chr.13: 36999247-36999247 on Build GRCh38
rs776431151 Chr.13: 36999293-36999293 on Build GRCh38

View Full Product Details