Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 41670857-41671355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VWA8
Entrez Gene ID
23078
Gene Name
von Willebrand factor A domain containing 8
Gene Aliases
KIAA0564
UniGene
Hs.368282
Gene Chromosome Location
Chr.13: 41566825-41961111 on Build GRCh38
Transcripts
NM_015058.1 XM_006719791.3 XM_017020469.1 XM_017020470.1 XM_017020472.1 XM_017020473.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
696559 c.4495G>A p.V1499I Chr.13: 41671062-41671062 on Build GRCh38
947474 c.4491C>T p.S1497S Chr.13: 41671066-41671066 on Build GRCh38
5970990 c.4546G>A p.E1516K Chr.13: 41671011-41671011 on Build GRCh38
7001481 c.4423_4424insG p.S1475fs*19 Chr.13: 41671133-41671134 on Build GRCh38
6821023 c.4489delA p.S1497fs*32 Chr.13: 41671068-41671068 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777308406 Chr.13: 41671074-41671074 on Build GRCh38
rs754253409 Chr.13: 41671316-41671316 on Build GRCh38
rs200757216 Chr.13: 41671032-41671032 on Build GRCh38
rs1023214123 Chr.13: 41671259-41671259 on Build GRCh38
rs752536500 Chr.13: 41671000-41671000 on Build GRCh38
rs566492078 Chr.13: 41671318-41671318 on Build GRCh38
rs761310308 Chr.13: 41670940-41670940 on Build GRCh38
rs768150088 Chr.13: 41671199-41671199 on Build GRCh38
rs934432280 Chr.13: 41670979-41670979 on Build GRCh38
rs12875049 Chr.13: 41671313-41671313 on Build GRCh38
rs758721806 Chr.13: 41671199-41671199 on Build GRCh38
rs771328858 Chr.13: 41670951-41670951 on Build GRCh38
rs769479504 Chr.13: 41671079-41671079 on Build GRCh38
rs779133353 Chr.13: 41670911-41670911 on Build GRCh38
rs781674647 Chr.13: 41671162-41671162 on Build GRCh38
rs764801777 Chr.13: 41670966-41670966 on Build GRCh38
rs748765756 Chr.13: 41671194-41671194 on Build GRCh38
rs375522079 Chr.13: 41670984-41670984 on Build GRCh38
rs751056495 Chr.13: 41671111-41671111 on Build GRCh38
rs371068026 Chr.13: 41671221-41671221 on Build GRCh38
rs771260024 Chr.13: 41671066-41671066 on Build GRCh38
rs747347146 Chr.13: 41671065-41671065 on Build GRCh38
rs761539962 Chr.13: 41671118-41671118 on Build GRCh38
rs374092988 Chr.13: 41671129-41671129 on Build GRCh38
rs934193941 Chr.13: 41671058-41671058 on Build GRCh38
rs751311007 Chr.13: 41670998-41670998 on Build GRCh38
rs746329626 Chr.13: 41671075-41671075 on Build GRCh38
rs750751946 Chr.13: 41671094-41671095 on Build GRCh38
rs747215747 Chr.13: 41671143-41671143 on Build GRCh38
rs774000962 Chr.13: 41671112-41671112 on Build GRCh38
rs775305708 Chr.13: 41670969-41670969 on Build GRCh38
rs370376270 Chr.13: 41671122-41671122 on Build GRCh38
rs762528080 Chr.13: 41671007-41671007 on Build GRCh38
rs763992144 Chr.13: 41670989-41670989 on Build GRCh38
rs756972845 Chr.13: 41671268-41671268 on Build GRCh38
rs1000500850 Chr.13: 41671312-41671312 on Build GRCh38
rs748317697 Chr.13: 41670912-41670912 on Build GRCh38
rs770422637 Chr.13: 41671166-41671166 on Build GRCh38
rs577232520 Chr.13: 41671059-41671059 on Build GRCh38
rs775200953 Chr.13: 41671171-41671171 on Build GRCh38
rs368335023 Chr.13: 41671213-41671213 on Build GRCh38
rs778938748 Chr.13: 41671042-41671042 on Build GRCh38
rs896535659 Chr.13: 41671235-41671235 on Build GRCh38
rs1038403732 Chr.13: 41671209-41671209 on Build GRCh38
rs752758480 Chr.13: 41671130-41671130 on Build GRCh38
rs149681227 Chr.13: 41670929-41670929 on Build GRCh38
rs370112959 Chr.13: 41670999-41670999 on Build GRCh38
rs376861790 Chr.13: 41670961-41670961 on Build GRCh38
rs768652380 Chr.13: 41670904-41670904 on Build GRCh38
rs757585303 Chr.13: 41671148-41671148 on Build GRCh38
rs185503998 Chr.13: 41671141-41671141 on Build GRCh38
rs1050311295 Chr.13: 41671088-41671088 on Build GRCh38
rs763455968 Chr.13: 41671054-41671054 on Build GRCh38
rs200645898 Chr.13: 41671163-41671163 on Build GRCh38
rs750407003 Chr.13: 41671121-41671121 on Build GRCh38
rs979952069 Chr.13: 41671178-41671178 on Build GRCh38
rs907886845 Chr.13: 41671216-41671217 on Build GRCh38
rs759122244 Chr.13: 41670964-41670964 on Build GRCh38
rs187938483 Chr.13: 41671280-41671280 on Build GRCh38
rs748407488 Chr.13: 41671053-41671053 on Build GRCh38
rs922358501 Chr.13: 41671329-41671329 on Build GRCh38
rs762763924 Chr.13: 41671107-41671107 on Build GRCh38
rs372498736 Chr.13: 41671008-41671008 on Build GRCh38
rs3818591 Chr.13: 41671243-41671243 on Build GRCh38
rs140258204 Chr.13: 41671215-41671215 on Build GRCh38
rs764810773 Chr.13: 41670950-41670951 on Build GRCh38
rs773416842 Chr.13: 41670937-41670937 on Build GRCh38
rs546080502 Chr.13: 41671120-41671120 on Build GRCh38
rs758444868 Chr.13: 41671132-41671132 on Build GRCh38
rs978482826 Chr.13: 41670962-41670962 on Build GRCh38
rs775110894 Chr.13: 41671099-41671099 on Build GRCh38
rs554693683 Chr.13: 41670884-41670884 on Build GRCh38

View Full Product Details