Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 43107063-43107600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC15
Entrez Gene ID
29103
Gene Name
DnaJ heat shock protein family (Hsp40) member C15
Gene Aliases
DNAJD1, HSD18, MCJ
UniGene
Hs.438830
Gene Chromosome Location
Chr.13: 43023226-43109170 on Build GRCh38
Transcripts
NM_013238.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3704642 c.391C>T p.P131S Chr.13: 43107186-43107186 on Build GRCh38
7341236 c.416A>T p.E139V Chr.13: 43107211-43107211 on Build GRCh38
5376768 c.392C>T p.P131L Chr.13: 43107187-43107187 on Build GRCh38
947571 c.450T>G p.H150Q Chr.13: 43107245-43107245 on Build GRCh38
4678722 c.446A>C p.K149T Chr.13: 43107241-43107241 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs554277829 Chr.13: 43107387-43107387 on Build GRCh38
rs770839736 Chr.13: 43107147-43107147 on Build GRCh38
rs376958924 Chr.13: 43107525-43107534 on Build GRCh38
rs368339708 Chr.13: 43107157-43107157 on Build GRCh38
rs772364285 Chr.13: 43107292-43107292 on Build GRCh38
rs987190318 Chr.13: 43107107-43107107 on Build GRCh38
rs370731770 Chr.13: 43107256-43107256 on Build GRCh38
rs535378779 Chr.13: 43107291-43107292 on Build GRCh38
rs769526825 Chr.13: 43107290-43107290 on Build GRCh38
rs370009265 Chr.13: 43107131-43107131 on Build GRCh38
rs72088427 Chr.13: 43107525-43107530 on Build GRCh38
rs779844332 Chr.13: 43107148-43107157 on Build GRCh38
rs201602896 Chr.13: 43107173-43107173 on Build GRCh38
rs756964252 Chr.13: 43107524-43107529 on Build GRCh38
rs142535946 Chr.13: 43107357-43107357 on Build GRCh38
rs545582541 Chr.13: 43107292-43107293 on Build GRCh38
rs765496709 Chr.13: 43107213-43107213 on Build GRCh38
rs138061775 Chr.13: 43107192-43107192 on Build GRCh38
rs181477714 Chr.13: 43107474-43107474 on Build GRCh38
rs776754011 Chr.13: 43107428-43107428 on Build GRCh38
rs889179583 Chr.13: 43107118-43107118 on Build GRCh38
rs57085120 Chr.13: 43107551-43107556 on Build GRCh38
rs188992233 Chr.13: 43107235-43107235 on Build GRCh38
rs779492473 Chr.13: 43107524-43107527 on Build GRCh38
rs551971889 Chr.13: 43107308-43107308 on Build GRCh38
rs756760442 Chr.13: 43107244-43107244 on Build GRCh38
rs530121565 Chr.13: 43107290-43107291 on Build GRCh38
rs61651931 Chr.13: 43107303-43107304 on Build GRCh38
rs1018704458 Chr.13: 43107455-43107455 on Build GRCh38
rs925400914 Chr.13: 43107380-43107380 on Build GRCh38
rs1047762 Chr.13: 43107261-43107261 on Build GRCh38
rs201270930 Chr.13: 43107553-43107553 on Build GRCh38
rs1026443557 Chr.13: 43107098-43107099 on Build GRCh38
rs760852761 Chr.13: 43107202-43107202 on Build GRCh38
rs568585702 Chr.13: 43107321-43107321 on Build GRCh38
rs747686723 Chr.13: 43107271-43107271 on Build GRCh38
rs761913590 Chr.13: 43107188-43107188 on Build GRCh38
rs761724309 Chr.13: 43107530-43107531 on Build GRCh38
rs879108526 Chr.13: 43107525-43107526 on Build GRCh38
rs768725441 Chr.13: 43107527-43107530 on Build GRCh38
rs200847296 Chr.13: 43107145-43107145 on Build GRCh38
rs988768517 Chr.13: 43107394-43107394 on Build GRCh38
rs1048050884 Chr.13: 43107548-43107548 on Build GRCh38
rs202074985 Chr.13: 43107556-43107557 on Build GRCh38
rs796302397 Chr.13: 43107291-43107292 on Build GRCh38
rs751584745 Chr.13: 43107164-43107175 on Build GRCh38
rs776894488 Chr.13: 43107300-43107301 on Build GRCh38
rs768505381 Chr.13: 43107479-43107479 on Build GRCh38
rs367738022 Chr.13: 43107243-43107243 on Build GRCh38
rs796628626 Chr.13: 43107292-43107293 on Build GRCh38
rs1026995972 Chr.13: 43107395-43107395 on Build GRCh38
rs777231665 Chr.13: 43107136-43107136 on Build GRCh38
rs778963550 Chr.13: 43107552-43107557 on Build GRCh38
rs558103629 Chr.13: 43107574-43107574 on Build GRCh38
rs1047775 Chr.13: 43107291-43107291 on Build GRCh38
rs67714115 Chr.13: 43107291-43107291 on Build GRCh38
rs200502838 Chr.13: 43107557-43107557 on Build GRCh38
rs950968253 Chr.13: 43107102-43107103 on Build GRCh38
rs933348170 Chr.13: 43107097-43107097 on Build GRCh38
rs969795143 Chr.13: 43107345-43107345 on Build GRCh38
rs9562463 Chr.13: 43107290-43107290 on Build GRCh38
rs766926766 Chr.13: 43107204-43107204 on Build GRCh38
rs9594889 Chr.13: 43107148-43107148 on Build GRCh38
rs746320381 Chr.13: 43107291-43107293 on Build GRCh38
rs745764821 Chr.13: 43107483-43107483 on Build GRCh38
rs150004773 Chr.13: 43107209-43107209 on Build GRCh38
rs1052226406 Chr.13: 43107104-43107104 on Build GRCh38
rs144620914 Chr.13: 43107176-43107176 on Build GRCh38
rs948714056 Chr.13: 43107314-43107314 on Build GRCh38
rs968129336 Chr.13: 43107526-43107526 on Build GRCh38
rs1051712388 Chr.13: 43107444-43107444 on Build GRCh38
rs533828110 Chr.13: 43107469-43107469 on Build GRCh38
rs778561659 Chr.13: 43107127-43107127 on Build GRCh38
rs72139916 Chr.13: 43107525-43107532 on Build GRCh38
rs543030130 Chr.13: 43107292-43107292 on Build GRCh38
rs145366836 Chr.13: 43107220-43107220 on Build GRCh38
rs895716901 Chr.13: 43107573-43107573 on Build GRCh38
rs760797822 Chr.13: 43107296-43107296 on Build GRCh38
rs199640403 Chr.13: 43107245-43107245 on Build GRCh38
rs17064181 Chr.13: 43107399-43107399 on Build GRCh38
rs1021925660 Chr.13: 43107516-43107516 on Build GRCh38
rs933591075 Chr.13: 43107442-43107442 on Build GRCh38
rs147456352 Chr.13: 43107191-43107191 on Build GRCh38
rs769812992 Chr.13: 43107282-43107282 on Build GRCh38
rs773573866 Chr.13: 43107293-43107293 on Build GRCh38
rs754879722 Chr.13: 43107175-43107176 on Build GRCh38
rs528425525 Chr.13: 43107270-43107271 on Build GRCh38
rs757979800 Chr.13: 43107234-43107234 on Build GRCh38
rs749126567 Chr.13: 43107447-43107447 on Build GRCh38
rs141469879 Chr.13: 43107525-43107528 on Build GRCh38
rs183874042 Chr.13: 43107128-43107128 on Build GRCh38
rs993359421 Chr.13: 43107518-43107518 on Build GRCh38
rs566761905 Chr.13: 43107286-43107286 on Build GRCh38
rs779711541 Chr.13: 43107284-43107284 on Build GRCh38

View Full Product Details