Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 45408834-45409384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A30
Entrez Gene ID
253512
Gene Name
solute carrier family 25 member 30
Gene Aliases
KMCP1
UniGene
Hs.591230
Gene Chromosome Location
Chr.13: 45393316-45434016 on Build GRCh38
Transcripts
NM_001010875.3 NM_001286806.1 NM_001286807.1 XM_005266321.2 XM_005266322.1 XM_006719793.3 XM_006719794.1 XM_011535026.1 XM_011535027.2 XM_017020523.1 XM_017020524.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4047780 c.135G>T p.K45N Chr.13: 45409004-45409004 on Build GRCh38
1747456 c.195G>A p.L65L Chr.13: 45408944-45408944 on Build GRCh38
4047779 c.163G>A p.A55T Chr.13: 45408976-45408976 on Build GRCh38
6780690 c.162C>T p.H54H Chr.13: 45408977-45408977 on Build GRCh38
1677644 c.199G>A p.A67T Chr.13: 45408940-45408940 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370511501 Chr.13: 45408963-45408963 on Build GRCh38
rs1011955680 Chr.13: 45409344-45409344 on Build GRCh38
rs371468082 Chr.13: 45408892-45408892 on Build GRCh38
rs776928023 Chr.13: 45408952-45408952 on Build GRCh38
rs781448606 Chr.13: 45409051-45409051 on Build GRCh38
rs764167986 Chr.13: 45409098-45409098 on Build GRCh38
rs756538416 Chr.13: 45409103-45409103 on Build GRCh38
rs750085119 Chr.13: 45409008-45409008 on Build GRCh38
rs902363415 Chr.13: 45409307-45409307 on Build GRCh38
rs758888803 Chr.13: 45408878-45408878 on Build GRCh38
rs762373234 Chr.13: 45409108-45409108 on Build GRCh38
rs1042616943 Chr.13: 45408954-45408954 on Build GRCh38
rs199725741 Chr.13: 45408964-45408964 on Build GRCh38
rs1029928425 Chr.13: 45408930-45408930 on Build GRCh38
rs541533010 Chr.13: 45409341-45409341 on Build GRCh38
rs966575586 Chr.13: 45409034-45409034 on Build GRCh38
rs144927807 Chr.13: 45409273-45409273 on Build GRCh38
rs778494590 Chr.13: 45409321-45409321 on Build GRCh38
rs761654563 Chr.13: 45408946-45408946 on Build GRCh38
rs775959430 Chr.13: 45409107-45409109 on Build GRCh38
rs771645923 Chr.13: 45408910-45408911 on Build GRCh38
rs201579618 Chr.13: 45408951-45408951 on Build GRCh38
rs368357937 Chr.13: 45409023-45409023 on Build GRCh38
rs778938365 Chr.13: 45409037-45409037 on Build GRCh38
rs927387626 Chr.13: 45409133-45409133 on Build GRCh38
rs148910106 Chr.13: 45409057-45409057 on Build GRCh38
rs752079823 Chr.13: 45408991-45408991 on Build GRCh38
rs370602370 Chr.13: 45409045-45409045 on Build GRCh38
rs752771414 Chr.13: 45408977-45408977 on Build GRCh38
rs61733507 Chr.13: 45409058-45409058 on Build GRCh38
rs1057191473 Chr.13: 45409167-45409167 on Build GRCh38
rs753053811 Chr.13: 45409093-45409093 on Build GRCh38
rs780771009 Chr.13: 45408904-45408904 on Build GRCh38
rs1020379381 Chr.13: 45409347-45409347 on Build GRCh38
rs763083921 Chr.13: 45408910-45408910 on Build GRCh38
rs1001035017 Chr.13: 45408979-45408979 on Build GRCh38
rs142058458 Chr.13: 45408990-45408990 on Build GRCh38
rs768305010 Chr.13: 45409026-45409026 on Build GRCh38
rs117699935 Chr.13: 45409022-45409022 on Build GRCh38
rs144455593 Chr.13: 45409014-45409014 on Build GRCh38
rs973547275 Chr.13: 45409132-45409132 on Build GRCh38
rs765351144 Chr.13: 45408968-45408968 on Build GRCh38
rs937399361 Chr.13: 45409151-45409151 on Build GRCh38
rs532319598 Chr.13: 45409336-45409336 on Build GRCh38
rs949335371 Chr.13: 45409272-45409272 on Build GRCh38
rs1038133108 Chr.13: 45409257-45409257 on Build GRCh38
rs763488313 Chr.13: 45408978-45408978 on Build GRCh38
rs917216676 Chr.13: 45409175-45409175 on Build GRCh38
rs753792185 Chr.13: 45409038-45409038 on Build GRCh38
rs528796034 Chr.13: 45408874-45408874 on Build GRCh38
rs548561020 Chr.13: 45409044-45409044 on Build GRCh38
rs774462993 Chr.13: 45408934-45408934 on Build GRCh38
rs778092492 Chr.13: 45408883-45408883 on Build GRCh38
rs373778087 Chr.13: 45408929-45408929 on Build GRCh38
rs766447336 Chr.13: 45408916-45408916 on Build GRCh38
rs1051681082 Chr.13: 45409275-45409275 on Build GRCh38
rs749233489 Chr.13: 45408926-45408926 on Build GRCh38
rs565053875 Chr.13: 45409326-45409326 on Build GRCh38
rs755780243 Chr.13: 45409016-45409016 on Build GRCh38
rs1018267256 Chr.13: 45409327-45409327 on Build GRCh38
rs751958411 Chr.13: 45408891-45408891 on Build GRCh38
rs373917288 Chr.13: 45409004-45409004 on Build GRCh38
rs891406173 Chr.13: 45409287-45409287 on Build GRCh38
rs769280434 Chr.13: 45408912-45408912 on Build GRCh38
rs1038983484 Chr.13: 45408887-45408887 on Build GRCh38
rs748101825 Chr.13: 45409040-45409040 on Build GRCh38
rs757555494 Chr.13: 45409000-45409000 on Build GRCh38

View Full Product Details