Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49259039-49259542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDADC1
Entrez Gene ID
81602
Gene Name
cytidine and dCMP deaminase domain containing 1
Gene Aliases
NYD-SP15, bA103J18.1
UniGene
Hs.388220
Gene Chromosome Location
Chr.13: 49247875-49293486 on Build GRCh38
Transcripts
NM_001193478.1 NM_030911.3 NR_036439.1 XM_011535250.2 XM_011535251.1 XM_011535253.1 XM_017020778.1 XM_017020779.1 XM_017020780.1 XM_017020781.1 XM_017020782.1 XM_017020783.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6442887 c.379C>G p.L127V Chr.13: 49259472-49259472 on Build GRCh38
4047957 c.388T>C p.S130P Chr.13: 49259481-49259481 on Build GRCh38
4047956 c.388T>C p.S130P Chr.13: 49259481-49259481 on Build GRCh38
3356402 c.331G>A p.G111R Chr.13: 49259424-49259424 on Build GRCh38
7323778 c.343C>G p.L115V Chr.13: 49259436-49259436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1031463863 Chr.13: 49259244-49259244 on Build GRCh38
rs893067025 Chr.13: 49259270-49259270 on Build GRCh38
rs948495454 Chr.13: 49259123-49259123 on Build GRCh38
rs778510767 Chr.13: 49259439-49259439 on Build GRCh38
rs557394020 Chr.13: 49259095-49259095 on Build GRCh38
rs780505887 Chr.13: 49259424-49259424 on Build GRCh38
rs752121960 Chr.13: 49259307-49259307 on Build GRCh38
rs377627444 Chr.13: 49259300-49259300 on Build GRCh38
rs374038513 Chr.13: 49259446-49259446 on Build GRCh38
rs768221697 Chr.13: 49259321-49259321 on Build GRCh38
rs768177985 Chr.13: 49259413-49259413 on Build GRCh38
rs376196398 Chr.13: 49259409-49259409 on Build GRCh38
rs981516065 Chr.13: 49259287-49259287 on Build GRCh38
rs1022345096 Chr.13: 49259335-49259335 on Build GRCh38
rs766160557 Chr.13: 49259371-49259371 on Build GRCh38
rs771951868 Chr.13: 49259329-49259329 on Build GRCh38
rs149539333 Chr.13: 49259326-49259326 on Build GRCh38
rs138009628 Chr.13: 49259305-49259305 on Build GRCh38
rs373879209 Chr.13: 49259387-49259387 on Build GRCh38
rs1032298821 Chr.13: 49259132-49259132 on Build GRCh38
rs546357990 Chr.13: 49259276-49259276 on Build GRCh38
rs773203015 Chr.13: 49259449-49259449 on Build GRCh38
rs770250667 Chr.13: 49259317-49259317 on Build GRCh38
rs376076436 Chr.13: 49259466-49259466 on Build GRCh38
rs770727651 Chr.13: 49259484-49259484 on Build GRCh38
rs773664140 Chr.13: 49259324-49259324 on Build GRCh38
rs780073811 Chr.13: 49259517-49259523 on Build GRCh38
rs966562111 Chr.13: 49259100-49259100 on Build GRCh38
rs999112977 Chr.13: 49259236-49259236 on Build GRCh38
rs763268368 Chr.13: 49259501-49259501 on Build GRCh38
rs758572045 Chr.13: 49259304-49259304 on Build GRCh38
rs781706959 Chr.13: 49259311-49259311 on Build GRCh38
rs764238517 Chr.13: 49259390-49259390 on Build GRCh38
rs950197427 Chr.13: 49259252-49259252 on Build GRCh38
rs943019534 Chr.13: 49259388-49259388 on Build GRCh38
rs374943987 Chr.13: 49259295-49259295 on Build GRCh38
rs372330771 Chr.13: 49259491-49259491 on Build GRCh38
rs760506329 Chr.13: 49259346-49259346 on Build GRCh38
rs772905973 Chr.13: 49259492-49259492 on Build GRCh38
rs370394327 Chr.13: 49259483-49259483 on Build GRCh38
rs748971034 Chr.13: 49259425-49259425 on Build GRCh38
rs748876140 Chr.13: 49259320-49259320 on Build GRCh38
rs915970163 Chr.13: 49259248-49259251 on Build GRCh38
rs1039126827 Chr.13: 49259400-49259400 on Build GRCh38
rs747664302 Chr.13: 49259441-49259441 on Build GRCh38
rs199866241 Chr.13: 49259421-49259421 on Build GRCh38
rs746282959 Chr.13: 49259314-49259314 on Build GRCh38
rs754542836 Chr.13: 49259427-49259427 on Build GRCh38
rs775747848 Chr.13: 49259379-49259379 on Build GRCh38
rs758550740 Chr.13: 49259460-49259461 on Build GRCh38
rs1020779381 Chr.13: 49259068-49259068 on Build GRCh38
rs746937744 Chr.13: 49259480-49259480 on Build GRCh38
rs561981629 Chr.13: 49259419-49259419 on Build GRCh38
rs757638434 Chr.13: 49259310-49259310 on Build GRCh38
rs370311963 Chr.13: 49259423-49259423 on Build GRCh38
rs972646699 Chr.13: 49259515-49259515 on Build GRCh38
rs779916883 Chr.13: 49259318-49259318 on Build GRCh38
rs575803424 Chr.13: 49259170-49259170 on Build GRCh38
rs764864602 Chr.13: 49259436-49259436 on Build GRCh38
rs1045875815 Chr.13: 49259204-49259204 on Build GRCh38
rs979336091 Chr.13: 49259119-49259119 on Build GRCh38
rs773114764 Chr.13: 49259334-49259334 on Build GRCh38
rs906922206 Chr.13: 49259211-49259211 on Build GRCh38

View Full Product Details