Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 52092580-52093129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEK5
Entrez Gene ID
341676
Gene Name
NIMA related kinase 5
Gene Aliases
-
UniGene
Hs.672144
Gene Chromosome Location
Chr.13: 52064762-52129092 on Build GRCh38
Transcripts
NM_199289.2 XM_006719808.3 XM_011535066.2 XM_011535068.2 XM_017020555.1 XM_017020556.1 XM_017020557.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4448726 c.1208+1G>A p.? Chr.13: 52093053-52093053 on Build GRCh38
3384873 c.1147G>A p.E383K Chr.13: 52093115-52093115 on Build GRCh38
948032 c.1181C>T p.T394M Chr.13: 52093081-52093081 on Build GRCh38
459266 c.1144C>T p.Q382* Chr.13: 52093118-52093118 on Build GRCh38
469510 c.1208G>A p.W403* Chr.13: 52093054-52093054 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145214408 Chr.13: 52093081-52093081 on Build GRCh38
rs1023054197 Chr.13: 52092842-52092842 on Build GRCh38
rs762236810 Chr.13: 52092937-52092937 on Build GRCh38
rs765679945 Chr.13: 52093025-52093025 on Build GRCh38
rs542828988 Chr.13: 52092848-52092848 on Build GRCh38
rs139319272 Chr.13: 52092860-52092860 on Build GRCh38
rs995791645 Chr.13: 52092994-52092994 on Build GRCh38
rs773987887 Chr.13: 52093004-52093004 on Build GRCh38
rs757402487 Chr.13: 52093080-52093080 on Build GRCh38
rs915933098 Chr.13: 52092656-52092656 on Build GRCh38
rs1035763986 Chr.13: 52092614-52092614 on Build GRCh38
rs896688853 Chr.13: 52092698-52092698 on Build GRCh38
rs548557599 Chr.13: 52092766-52092766 on Build GRCh38
rs191865099 Chr.13: 52092900-52092900 on Build GRCh38
rs544269657 Chr.13: 52092759-52092759 on Build GRCh38
rs896239732 Chr.13: 52092919-52092919 on Build GRCh38
rs960144772 Chr.13: 52092838-52092838 on Build GRCh38
rs751721894 Chr.13: 52093071-52093071 on Build GRCh38
rs903442839 Chr.13: 52093074-52093074 on Build GRCh38
rs1036635372 Chr.13: 52092758-52092758 on Build GRCh38
rs991950919 Chr.13: 52092839-52092839 on Build GRCh38
rs977570156 Chr.13: 52092605-52092605 on Build GRCh38
rs761357631 Chr.13: 52093009-52093009 on Build GRCh38
rs987965141 Chr.13: 52092817-52092817 on Build GRCh38
rs1037168610 Chr.13: 52092983-52092983 on Build GRCh38
rs926320868 Chr.13: 52092867-52092867 on Build GRCh38
rs1001976145 Chr.13: 52092630-52092630 on Build GRCh38
rs903472107 Chr.13: 52092794-52092794 on Build GRCh38
rs971913988 Chr.13: 52092896-52092896 on Build GRCh38
rs998959530 Chr.13: 52092801-52092801 on Build GRCh38
rs1056589384 Chr.13: 52092666-52092667 on Build GRCh38
rs759924241 Chr.13: 52093022-52093022 on Build GRCh38
rs946397400 Chr.13: 52092623-52092623 on Build GRCh38
rs888743391 Chr.13: 52092769-52092769 on Build GRCh38
rs954161371 Chr.13: 52092773-52092773 on Build GRCh38
rs562644687 Chr.13: 52092849-52092849 on Build GRCh38
rs560465062 Chr.13: 52092800-52092800 on Build GRCh38
rs947040110 Chr.13: 52093042-52093042 on Build GRCh38
rs965907407 Chr.13: 52092796-52092796 on Build GRCh38
rs1050575319 Chr.13: 52093006-52093006 on Build GRCh38
rs909147458 Chr.13: 52092719-52092719 on Build GRCh38
rs556370921 Chr.13: 52092731-52092731 on Build GRCh38
rs1056242170 Chr.13: 52092875-52092875 on Build GRCh38
rs142357047 Chr.13: 52093091-52093091 on Build GRCh38
rs368630433 Chr.13: 52092762-52092762 on Build GRCh38
rs539890804 Chr.13: 52092786-52092786 on Build GRCh38
rs149978343 Chr.13: 52092955-52092955 on Build GRCh38
rs181674367 Chr.13: 52092750-52092750 on Build GRCh38
rs1008641941 Chr.13: 52093034-52093034 on Build GRCh38
rs201813265 Chr.13: 52093013-52093013 on Build GRCh38
rs749188790 Chr.13: 52093102-52093102 on Build GRCh38
rs187587226 Chr.13: 52092776-52092776 on Build GRCh38
rs909116406 Chr.13: 52092868-52092868 on Build GRCh38
rs931311989 Chr.13: 52092663-52092663 on Build GRCh38
rs574012461 Chr.13: 52092833-52092833 on Build GRCh38
rs984669933 Chr.13: 52092699-52092699 on Build GRCh38
rs780082875 Chr.13: 52093097-52093097 on Build GRCh38
rs55715265 Chr.13: 52093092-52093092 on Build GRCh38
rs757646467 Chr.13: 52092901-52092901 on Build GRCh38
rs150842841 Chr.13: 52092624-52092624 on Build GRCh38
rs144410186 Chr.13: 52092618-52092618 on Build GRCh38
rs764074677 Chr.13: 52093063-52093063 on Build GRCh38
rs531407996 Chr.13: 52092853-52092853 on Build GRCh38
rs961980287 Chr.13: 52092879-52092879 on Build GRCh38
rs898629907 Chr.13: 52092993-52092993 on Build GRCh38
rs1021800758 Chr.13: 52092793-52092793 on Build GRCh38
rs984135407 Chr.13: 52092851-52092851 on Build GRCh38
rs568400564 Chr.13: 52093055-52093055 on Build GRCh38
rs1036309801 Chr.13: 52092811-52092811 on Build GRCh38

View Full Product Details