Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 52844704-52845210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDH8
Entrez Gene ID
5100
Gene Name
protocadherin 8
Gene Aliases
ARCADLIN, PAPC
UniGene
Hs.19492
Gene Chromosome Location
Chr.13: 52843974-52848640 on Build GRCh38
Transcripts
NM_002590.3 NM_032949.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5624853 c.3042G>T p.L1014L Chr.13: 52844731-52844731 on Build GRCh38
5628602 c.2927C>T p.P976L Chr.13: 52844846-52844846 on Build GRCh38
6268624 c.3040C>A p.L1014M Chr.13: 52844733-52844733 on Build GRCh38
1219512 c.2953T>C p.S985P Chr.13: 52844820-52844820 on Build GRCh38
1219511 c.2888G>A p.R963H Chr.13: 52844885-52844885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020304012 Chr.13: 52845145-52845146 on Build GRCh38
rs777349184 Chr.13: 52844819-52844819 on Build GRCh38
rs201055906 Chr.13: 52844817-52844817 on Build GRCh38
rs766623736 Chr.13: 52844763-52844763 on Build GRCh38
rs202162563 Chr.13: 52844928-52844928 on Build GRCh38
rs752537106 Chr.13: 52844909-52844909 on Build GRCh38
rs1009871656 Chr.13: 52845121-52845121 on Build GRCh38
rs751145802 Chr.13: 52844829-52844829 on Build GRCh38
rs926128347 Chr.13: 52844925-52844925 on Build GRCh38
rs868183205 Chr.13: 52844824-52844824 on Build GRCh38
rs994615187 Chr.13: 52844970-52844970 on Build GRCh38
rs887140956 Chr.13: 52844979-52844979 on Build GRCh38
rs927783933 Chr.13: 52844884-52844884 on Build GRCh38
rs267603851 Chr.13: 52844861-52844861 on Build GRCh38
rs899065406 Chr.13: 52844931-52844931 on Build GRCh38
rs950329376 Chr.13: 52844937-52844937 on Build GRCh38
rs978793132 Chr.13: 52844917-52844917 on Build GRCh38
rs753623139 Chr.13: 52844778-52844778 on Build GRCh38
rs577347803 Chr.13: 52844757-52844757 on Build GRCh38
rs776783055 Chr.13: 52844978-52844978 on Build GRCh38
rs770899664 Chr.13: 52844971-52844971 on Build GRCh38
rs3742299 Chr.13: 52844843-52844843 on Build GRCh38
rs867890787 Chr.13: 52844812-52844812 on Build GRCh38
rs867700520 Chr.13: 52844870-52844870 on Build GRCh38
rs888861014 Chr.13: 52845029-52845029 on Build GRCh38
rs201885126 Chr.13: 52844805-52844805 on Build GRCh38
rs926071028 Chr.13: 52845188-52845188 on Build GRCh38
rs1007438624 Chr.13: 52845039-52845039 on Build GRCh38
rs978825646 Chr.13: 52845155-52845155 on Build GRCh38
rs371976283 Chr.13: 52844941-52844941 on Build GRCh38
rs765142969 Chr.13: 52844799-52844799 on Build GRCh38
rs773294757 Chr.13: 52844738-52844738 on Build GRCh38
rs770082639 Chr.13: 52844984-52844984 on Build GRCh38
rs754728186 Chr.13: 52844738-52844739 on Build GRCh38
rs778233522 Chr.13: 52844966-52844966 on Build GRCh38
rs1055878926 Chr.13: 52844927-52844927 on Build GRCh38
rs762942607 Chr.13: 52844916-52844916 on Build GRCh38
rs1028512771 Chr.13: 52845124-52845124 on Build GRCh38
rs780823635 Chr.13: 52844929-52844929 on Build GRCh38
rs376578193 Chr.13: 52844842-52844842 on Build GRCh38
rs901259132 Chr.13: 52845054-52845054 on Build GRCh38
rs966017733 Chr.13: 52845097-52845097 on Build GRCh38
rs746006475 Chr.13: 52844980-52844980 on Build GRCh38
rs751594263 Chr.13: 52844924-52844924 on Build GRCh38
rs572138106 Chr.13: 52845049-52845049 on Build GRCh38
rs772523199 Chr.13: 52844944-52844944 on Build GRCh38
rs150492311 Chr.13: 52844895-52844895 on Build GRCh38
rs764878292 Chr.13: 52844904-52844904 on Build GRCh38
rs773239656 Chr.13: 52844862-52844862 on Build GRCh38
rs367908389 Chr.13: 52844932-52844932 on Build GRCh38
rs897454468 Chr.13: 52844963-52844963 on Build GRCh38
rs781112432 Chr.13: 52844741-52844741 on Build GRCh38
rs772574126 Chr.13: 52844854-52844854 on Build GRCh38
rs759857297 Chr.13: 52844901-52844901 on Build GRCh38
rs141509995 Chr.13: 52844853-52844853 on Build GRCh38
rs2236363 Chr.13: 52845004-52845004 on Build GRCh38
rs1048370311 Chr.13: 52844965-52844965 on Build GRCh38
rs747557465 Chr.13: 52844969-52844969 on Build GRCh38
rs745572281 Chr.13: 52844836-52844836 on Build GRCh38
rs764149792 Chr.13: 52844923-52844923 on Build GRCh38
rs779347497 Chr.13: 52844940-52844940 on Build GRCh38
rs759232639 Chr.13: 52844789-52844789 on Build GRCh38
rs967746384 Chr.13: 52845146-52845146 on Build GRCh38
rs760770328 Chr.13: 52844893-52844893 on Build GRCh38
rs937474226 Chr.13: 52844926-52844926 on Build GRCh38
rs377547518 Chr.13: 52844727-52844727 on Build GRCh38
rs374007225 Chr.13: 52844834-52844834 on Build GRCh38
rs776710357 Chr.13: 52844898-52844898 on Build GRCh38
rs867928404 Chr.13: 52845063-52845063 on Build GRCh38
rs780684382 Chr.13: 52844835-52844835 on Build GRCh38
rs945105819 Chr.13: 52844850-52844850 on Build GRCh38
rs913582252 Chr.13: 52844841-52844841 on Build GRCh38
rs996976671 Chr.13: 52845102-52845102 on Build GRCh38

View Full Product Details