Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 83879922-83880430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLITRK1
Entrez Gene ID
114798
Gene Name
SLIT and NTRK like family member 1
Gene Aliases
LRRC12, TTM
UniGene
Hs.415478
Gene Chromosome Location
Chr.13: 83877205-83882393 on Build GRCh38
Transcripts
NM_001281503.1 NM_052910.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1607253 c.1543C>A p.L515M Chr.13: 83879965-83879965 on Build GRCh38
459237 c.1428C>A p.L476L Chr.13: 83880080-83880080 on Build GRCh38
5653542 c.1315T>C p.S439P Chr.13: 83880193-83880193 on Build GRCh38
5362181 c.1217T>C p.L406P Chr.13: 83880291-83880291 on Build GRCh38
6261861 c.1495C>A p.L499I Chr.13: 83880013-83880013 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769689046 Chr.13: 83880236-83880236 on Build GRCh38
rs753451536 Chr.13: 83880392-83880392 on Build GRCh38
rs780649631 Chr.13: 83880072-83880072 on Build GRCh38
rs142639256 Chr.13: 83880080-83880080 on Build GRCh38
rs749904055 Chr.13: 83880386-83880386 on Build GRCh38
rs757819524 Chr.13: 83879979-83879979 on Build GRCh38
rs193302861 Chr.13: 83880244-83880244 on Build GRCh38
rs150504822 Chr.13: 83880256-83880256 on Build GRCh38
rs775804434 Chr.13: 83880351-83880351 on Build GRCh38
rs746196829 Chr.13: 83880338-83880338 on Build GRCh38
rs746718348 Chr.13: 83880044-83880044 on Build GRCh38
rs117961355 Chr.13: 83879989-83879989 on Build GRCh38
rs751501445 Chr.13: 83880260-83880260 on Build GRCh38
rs140899296 Chr.13: 83880343-83880343 on Build GRCh38
rs540539932 Chr.13: 83880247-83880247 on Build GRCh38
rs372083302 Chr.13: 83880301-83880301 on Build GRCh38
rs1042996810 Chr.13: 83880014-83880014 on Build GRCh38
rs767117381 Chr.13: 83880380-83880380 on Build GRCh38
rs1032347594 Chr.13: 83880137-83880137 on Build GRCh38
rs200447166 Chr.13: 83879970-83879970 on Build GRCh38
rs753343543 Chr.13: 83880103-83880103 on Build GRCh38
rs764910876 Chr.13: 83880107-83880107 on Build GRCh38
rs768539503 Chr.13: 83880371-83880371 on Build GRCh38
rs988197980 Chr.13: 83880402-83880402 on Build GRCh38
rs780928365 Chr.13: 83880161-83880161 on Build GRCh38
rs200924568 Chr.13: 83880224-83880224 on Build GRCh38
rs200160380 Chr.13: 83880232-83880232 on Build GRCh38
rs774671182 Chr.13: 83879959-83879959 on Build GRCh38
rs761426714 Chr.13: 83880266-83880266 on Build GRCh38
rs758398750 Chr.13: 83880124-83880124 on Build GRCh38
rs1001384905 Chr.13: 83880031-83880031 on Build GRCh38
rs758806174 Chr.13: 83880400-83880400 on Build GRCh38
rs747445549 Chr.13: 83880316-83880316 on Build GRCh38
rs569289544 Chr.13: 83880105-83880105 on Build GRCh38
rs549097396 Chr.13: 83880093-83880093 on Build GRCh38
rs752646104 Chr.13: 83880008-83880008 on Build GRCh38
rs866132504 Chr.13: 83880054-83880054 on Build GRCh38
rs750144491 Chr.13: 83880275-83880275 on Build GRCh38
rs755712979 Chr.13: 83880285-83880285 on Build GRCh38
rs752731962 Chr.13: 83880117-83880117 on Build GRCh38
rs756967655 Chr.13: 83880049-83880049 on Build GRCh38
rs766640879 Chr.13: 83880104-83880104 on Build GRCh38
rs778288574 Chr.13: 83880314-83880314 on Build GRCh38
rs921540492 Chr.13: 83880323-83880323 on Build GRCh38
rs760870601 Chr.13: 83879972-83879972 on Build GRCh38
rs760171615 Chr.13: 83880387-83880387 on Build GRCh38
rs761639713 Chr.13: 83880379-83880379 on Build GRCh38
rs772193157 Chr.13: 83880097-83880097 on Build GRCh38
rs766913591 Chr.13: 83880272-83880272 on Build GRCh38
rs375655554 Chr.13: 83880114-83880114 on Build GRCh38
rs769913716 Chr.13: 83880350-83880350 on Build GRCh38
rs773468859 Chr.13: 83879971-83879971 on Build GRCh38
rs759410083 Chr.13: 83880106-83880106 on Build GRCh38
rs886050334 Chr.13: 83880016-83880016 on Build GRCh38
rs779231728 Chr.13: 83880192-83880192 on Build GRCh38
rs980748911 Chr.13: 83880130-83880130 on Build GRCh38
rs761912581 Chr.13: 83879960-83879960 on Build GRCh38
rs888200525 Chr.13: 83879947-83879947 on Build GRCh38
rs775422257 Chr.13: 83880245-83880245 on Build GRCh38
rs550166518 Chr.13: 83880305-83880305 on Build GRCh38
rs969332686 Chr.13: 83880128-83880128 on Build GRCh38
rs557887167 Chr.13: 83880230-83880230 on Build GRCh38
rs766044466 Chr.13: 83880389-83880389 on Build GRCh38
rs751127410 Chr.13: 83880159-83880159 on Build GRCh38
rs866867094 Chr.13: 83880187-83880187 on Build GRCh38
rs769217224 Chr.13: 83879957-83879957 on Build GRCh38
rs773272574 Chr.13: 83880100-83880100 on Build GRCh38
rs745478522 Chr.13: 83880188-83880188 on Build GRCh38
rs868463485 Chr.13: 83880134-83880134 on Build GRCh38
rs748628299 Chr.13: 83880094-83880094 on Build GRCh38
rs749683191 Chr.13: 83880079-83880079 on Build GRCh38
rs141650772 Chr.13: 83880023-83880023 on Build GRCh38
rs1014587889 Chr.13: 83880121-83880121 on Build GRCh38
rs959775583 Chr.13: 83880092-83880092 on Build GRCh38
rs753070860 Chr.13: 83880286-83880286 on Build GRCh38
rs748718393 Chr.13: 83880196-83880196 on Build GRCh38
rs766151458 Chr.13: 83879975-83879975 on Build GRCh38
rs773655036 Chr.13: 83880222-83880222 on Build GRCh38
rs758716146 Chr.13: 83880312-83880312 on Build GRCh38
rs774477502 Chr.13: 83880374-83880374 on Build GRCh38
rs771092440 Chr.13: 83880322-83880322 on Build GRCh38
rs201016092 Chr.13: 83880370-83880370 on Build GRCh38
rs777400552 Chr.13: 83880030-83880030 on Build GRCh38
rs935404020 Chr.13: 83880390-83880390 on Build GRCh38
rs905653392 Chr.13: 83880024-83880024 on Build GRCh38
rs781770617 Chr.13: 83880191-83880191 on Build GRCh38

View Full Product Details