Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 99255413-99255910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR18
Entrez Gene ID
2841
Gene Name
G protein-coupled receptor 18
Gene Aliases
-
UniGene
Hs.741589
Gene Chromosome Location
Chr.13: 99254713-99262498 on Build GRCh38
Transcripts
NM_001098200.1 NM_005292.3 XM_006719946.3 NM_001144072.1 NM_177967.3 NR_026644.1 XM_006719948.3 XM_011521082.2 XM_011521083.2 XM_011521084.2

Gene Symbol
UBAC2
Entrez Gene ID
337867
Gene Name
UBA domain containing 2
Gene Aliases
PHGDHL1
UniGene
Hs.508545
Gene Chromosome Location
Chr.13: 99200425-99386499 on Build GRCh38
Transcripts
NM_001098200.1 NM_005292.3 XM_006719946.3 NM_001144072.1 NM_177967.3 NR_026644.1 XM_006719948.3 XM_011521082.2 XM_011521083.2 XM_011521084.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3987499 c.248C>T p.A83V Chr.13: 99255625-99255625 on Build GRCh38
6674002 c.391G>T p.E131* Chr.13: 99255482-99255482 on Build GRCh38
3987500 c.76C>G p.L26V Chr.13: 99255797-99255797 on Build GRCh38
3470240 c.180T>A p.Y60* Chr.13: 99255693-99255693 on Build GRCh38
3704750 c.421G>A p.A141T Chr.13: 99255452-99255452 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367651389 Chr.13: 99255489-99255489 on Build GRCh38
rs143171650 Chr.13: 99255468-99255468 on Build GRCh38
rs759661276 Chr.13: 99255473-99255473 on Build GRCh38
rs538186904 Chr.13: 99255542-99255542 on Build GRCh38
rs752254725 Chr.13: 99255815-99255815 on Build GRCh38
rs750736365 Chr.13: 99255744-99255744 on Build GRCh38
rs755059165 Chr.13: 99255765-99255765 on Build GRCh38
rs774847906 Chr.13: 99255805-99255805 on Build GRCh38
rs760398254 Chr.13: 99255678-99255679 on Build GRCh38
rs749880572 Chr.13: 99255796-99255796 on Build GRCh38
rs140539549 Chr.13: 99255740-99255740 on Build GRCh38
rs760134164 Chr.13: 99255614-99255614 on Build GRCh38
rs755502855 Chr.13: 99255452-99255452 on Build GRCh38
rs748032389 Chr.13: 99255773-99255773 on Build GRCh38
rs776777158 Chr.13: 99255451-99255451 on Build GRCh38
rs781365371 Chr.13: 99255537-99255537 on Build GRCh38
rs374546166 Chr.13: 99255504-99255504 on Build GRCh38
rs766645228 Chr.13: 99255851-99255851 on Build GRCh38
rs138394107 Chr.13: 99255821-99255821 on Build GRCh38
rs757618760 Chr.13: 99255735-99255735 on Build GRCh38
rs1039335150 Chr.13: 99255761-99255761 on Build GRCh38
rs1016659191 Chr.13: 99255839-99255839 on Build GRCh38
rs189039762 Chr.13: 99255645-99255645 on Build GRCh38
rs147490848 Chr.13: 99255693-99255693 on Build GRCh38
rs745952926 Chr.13: 99255539-99255539 on Build GRCh38
rs755112652 Chr.13: 99255865-99255865 on Build GRCh38
rs764658155 Chr.13: 99255802-99255802 on Build GRCh38
rs779442891 Chr.13: 99255456-99255456 on Build GRCh38
rs371398228 Chr.13: 99255769-99255769 on Build GRCh38
rs775281609 Chr.13: 99255637-99255638 on Build GRCh38
rs151309537 Chr.13: 99255474-99255474 on Build GRCh38
rs780790126 Chr.13: 99255449-99255449 on Build GRCh38
rs765936310 Chr.13: 99255705-99255705 on Build GRCh38
rs767327161 Chr.13: 99255592-99255592 on Build GRCh38
rs756424491 Chr.13: 99255745-99255745 on Build GRCh38
rs774352675 Chr.13: 99255570-99255570 on Build GRCh38
rs759073439 Chr.13: 99255799-99255799 on Build GRCh38
rs142984439 Chr.13: 99255523-99255523 on Build GRCh38
rs779079190 Chr.13: 99255871-99255871 on Build GRCh38
rs533640164 Chr.13: 99255778-99255778 on Build GRCh38
rs990432598 Chr.13: 99255697-99255697 on Build GRCh38
rs185252466 Chr.13: 99255442-99255442 on Build GRCh38
rs751964738 Chr.13: 99255630-99255630 on Build GRCh38
rs367795735 Chr.13: 99255640-99255640 on Build GRCh38
rs776752710 Chr.13: 99255469-99255469 on Build GRCh38
rs757600315 Chr.13: 99255636-99255636 on Build GRCh38
rs768700715 Chr.13: 99255556-99255556 on Build GRCh38
rs772744547 Chr.13: 99255683-99255683 on Build GRCh38
rs553909988 Chr.13: 99255727-99255727 on Build GRCh38
rs750044310 Chr.13: 99255601-99255601 on Build GRCh38
rs771953548 Chr.13: 99255673-99255673 on Build GRCh38
rs757027038 Chr.13: 99255498-99255498 on Build GRCh38
rs767809276 Chr.13: 99255743-99255743 on Build GRCh38
rs746778970 Chr.13: 99255785-99255785 on Build GRCh38
rs549443596 Chr.13: 99255443-99255443 on Build GRCh38
rs773134509 Chr.13: 99255550-99255550 on Build GRCh38
rs778250186 Chr.13: 99255628-99255628 on Build GRCh38
rs771212065 Chr.13: 99255536-99255536 on Build GRCh38
rs149113944 Chr.13: 99255458-99255458 on Build GRCh38
rs761722169 Chr.13: 99255662-99255662 on Build GRCh38
rs369303917 Chr.13: 99255854-99255854 on Build GRCh38
rs372739338 Chr.13: 99255883-99255883 on Build GRCh38
rs765850458 Chr.13: 99255615-99255615 on Build GRCh38
rs748648382 Chr.13: 99255512-99255512 on Build GRCh38
rs1038270775 Chr.13: 99255486-99255486 on Build GRCh38
rs750837913 Chr.13: 99255831-99255831 on Build GRCh38
rs761669083 Chr.13: 99255588-99255588 on Build GRCh38
rs771132347 Chr.13: 99255593-99255593 on Build GRCh38
rs539240344 Chr.13: 99255684-99255684 on Build GRCh38
rs764638150 Chr.13: 99255714-99255714 on Build GRCh38
rs766126078 Chr.13: 99255641-99255641 on Build GRCh38
rs775445604 Chr.13: 99255477-99255477 on Build GRCh38
rs773920225 Chr.13: 99255555-99255555 on Build GRCh38
rs371441036 Chr.13: 99255786-99255786 on Build GRCh38
rs753159651 Chr.13: 99255624-99255624 on Build GRCh38
rs747364454 Chr.13: 99255526-99255526 on Build GRCh38
rs780235298 Chr.13: 99255647-99255647 on Build GRCh38
rs758696917 Chr.13: 99255516-99255516 on Build GRCh38
rs370331461 Chr.13: 99255446-99255446 on Build GRCh38
rs753458670 Chr.13: 99255706-99255706 on Build GRCh38
rs776130635 Chr.13: 99255793-99255793 on Build GRCh38
rs759004749 Chr.13: 99255627-99255627 on Build GRCh38
rs374299759 Chr.13: 99255459-99255459 on Build GRCh38
rs111938999 Chr.13: 99255500-99255500 on Build GRCh38
rs865982902 Chr.13: 99255690-99255690 on Build GRCh38
rs61970277 Chr.13: 99255465-99255465 on Build GRCh38
rs745356452 Chr.13: 99255450-99255450 on Build GRCh38
rs768752139 Chr.13: 99255657-99255657 on Build GRCh38
rs376159827 Chr.13: 99255467-99255467 on Build GRCh38
rs759018484 Chr.13: 99255712-99255712 on Build GRCh38
rs756410859 Chr.13: 99255834-99255834 on Build GRCh38
rs755626668 Chr.13: 99255506-99255506 on Build GRCh38
rs571822303 Chr.13: 99255659-99255659 on Build GRCh38
rs889501425 Chr.13: 99255689-99255689 on Build GRCh38
rs780105154 Chr.13: 99255746-99255746 on Build GRCh38
rs775292571 Chr.13: 99255545-99255545 on Build GRCh38
rs749579391 Chr.13: 99255570-99255571 on Build GRCh38
rs762683808 Chr.13: 99255488-99255488 on Build GRCh38
rs749434511 Chr.13: 99255750-99255750 on Build GRCh38
rs749323566 Chr.13: 99255655-99255655 on Build GRCh38

View Full Product Details