Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 99984811-99985333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZIC2
Entrez Gene ID
7546
Gene Name
Zic family member 2
Gene Aliases
HPE5
UniGene
Hs.653700
Gene Chromosome Location
Chr.13: 99981772-99986765 on Build GRCh38
Transcripts
NM_007129.3 XM_011521110.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6311043 c.1098G>T p.E366D Chr.13: 99984968-99984968 on Build GRCh38
6827151 c.1156G>A p.V386I Chr.13: 99985026-99985026 on Build GRCh38
7403249 c.1141A>G p.K381E Chr.13: 99985011-99985011 on Build GRCh38
6344345 c.1152G>T p.M384I Chr.13: 99985022-99985022 on Build GRCh38
7100338 c.1214C>A p.P405H Chr.13: 99985084-99985084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1023524104 Chr.13: 99985159-99985159 on Build GRCh38
rs938662378 Chr.13: 99985020-99985020 on Build GRCh38
rs780380550 Chr.13: 99985152-99985152 on Build GRCh38
rs546620526 Chr.13: 99985261-99985261 on Build GRCh38
rs769776768 Chr.13: 99984925-99984925 on Build GRCh38
rs760314965 Chr.13: 99985016-99985016 on Build GRCh38
rs1057518503 Chr.13: 99984941-99984941 on Build GRCh38
rs750045150 Chr.13: 99985013-99985013 on Build GRCh38
rs950626015 Chr.13: 99984860-99984860 on Build GRCh38
rs762420178 Chr.13: 99985301-99985301 on Build GRCh38
rs752061109 Chr.13: 99985048-99985048 on Build GRCh38
rs1057444837 Chr.13: 99985026-99985026 on Build GRCh38
rs370509521 Chr.13: 99985137-99985137 on Build GRCh38
rs768752128 Chr.13: 99984937-99984937 on Build GRCh38
rs760273107 Chr.13: 99985117-99985117 on Build GRCh38
rs766107194 Chr.13: 99985118-99985118 on Build GRCh38
rs779073415 Chr.13: 99985161-99985161 on Build GRCh38
rs757712884 Chr.13: 99985140-99985140 on Build GRCh38
rs755110242 Chr.13: 99985155-99985155 on Build GRCh38
rs780106898 Chr.13: 99985065-99985065 on Build GRCh38
rs886041583 Chr.13: 99984967-99984968 on Build GRCh38
rs753432048 Chr.13: 99985031-99985031 on Build GRCh38
rs746035691 Chr.13: 99984923-99984923 on Build GRCh38
rs756410756 Chr.13: 99985151-99985151 on Build GRCh38
rs774495822 Chr.13: 99985104-99985104 on Build GRCh38
rs748028341 Chr.13: 99985115-99985115 on Build GRCh38
rs757624783 Chr.13: 99985052-99985052 on Build GRCh38
rs1038743512 Chr.13: 99985217-99985217 on Build GRCh38
rs747427835 Chr.13: 99984900-99984900 on Build GRCh38
rs1030021793 Chr.13: 99985253-99985253 on Build GRCh38
rs1030215955 Chr.13: 99984859-99984859 on Build GRCh38
rs996264199 Chr.13: 99984956-99984956 on Build GRCh38
rs983259108 Chr.13: 99984883-99984883 on Build GRCh38
rs750879393 Chr.13: 99985310-99985310 on Build GRCh38
rs749111385 Chr.13: 99984838-99984838 on Build GRCh38
rs770632004 Chr.13: 99985289-99985289 on Build GRCh38
rs771275199 Chr.13: 99984910-99984910 on Build GRCh38
rs772003781 Chr.13: 99985276-99985276 on Build GRCh38
rs778315160 Chr.13: 99985034-99985034 on Build GRCh38
rs773008224 Chr.13: 99985116-99985116 on Build GRCh38
rs369127058 Chr.13: 99985091-99985091 on Build GRCh38
rs928680757 Chr.13: 99984836-99984837 on Build GRCh38
rs748116312 Chr.13: 99985273-99985273 on Build GRCh38
rs997165301 Chr.13: 99985235-99985235 on Build GRCh38
rs963237180 Chr.13: 99984896-99984896 on Build GRCh38
rs370381240 Chr.13: 99985144-99985144 on Build GRCh38
rs746882309 Chr.13: 99985287-99985287 on Build GRCh38
rs776273987 Chr.13: 99985290-99985290 on Build GRCh38
rs927763124 Chr.13: 99985172-99985172 on Build GRCh38
rs950386908 Chr.13: 99985288-99985288 on Build GRCh38
rs981917769 Chr.13: 99985163-99985163 on Build GRCh38
rs9585308 Chr.13: 99984848-99984848 on Build GRCh38
rs765826453 Chr.13: 99985025-99985025 on Build GRCh38
rs750838088 Chr.13: 99985149-99985149 on Build GRCh38
rs113030947 Chr.13: 99985227-99985227 on Build GRCh38
rs1048221388 Chr.13: 99984852-99984852 on Build GRCh38
rs781563139 Chr.13: 99985054-99985054 on Build GRCh38
rs749517224 Chr.13: 99985153-99985153 on Build GRCh38
rs1006924617 Chr.13: 99984869-99984869 on Build GRCh38
rs759118402 Chr.13: 99985121-99985121 on Build GRCh38
rs909028447 Chr.13: 99984884-99984884 on Build GRCh38
rs774407989 Chr.13: 99984955-99984955 on Build GRCh38
rs1007511667 Chr.13: 99985186-99985186 on Build GRCh38
rs561915299 Chr.13: 99984878-99984878 on Build GRCh38
rs558250196 Chr.13: 99984894-99984895 on Build GRCh38
rs983546866 Chr.13: 99985191-99985191 on Build GRCh38
rs764814464 Chr.13: 99985049-99985050 on Build GRCh38
rs767413047 Chr.13: 99984984-99984984 on Build GRCh38
rs964407493 Chr.13: 99984841-99984841 on Build GRCh38
rs762688543 Chr.13: 99985136-99985137 on Build GRCh38
rs907904173 Chr.13: 99985199-99985199 on Build GRCh38
rs746102473 Chr.13: 99985059-99985059 on Build GRCh38
rs185662169 Chr.13: 99985073-99985073 on Build GRCh38
rs775592738 Chr.13: 99984928-99984928 on Build GRCh38
rs756320568 Chr.13: 99985060-99985060 on Build GRCh38
rs200208088 Chr.13: 99984980-99984980 on Build GRCh38
rs529436875 Chr.13: 99984917-99984917 on Build GRCh38
rs397515500 Chr.13: 99984961-99984962 on Build GRCh38
rs768117425 Chr.13: 99985306-99985306 on Build GRCh38
rs957726544 Chr.13: 99985147-99985147 on Build GRCh38
rs750137898 Chr.13: 99985117-99985118 on Build GRCh38
rs764664489 Chr.13: 99985126-99985126 on Build GRCh38
rs776137007 Chr.13: 99985119-99985119 on Build GRCh38
rs764749763 Chr.13: 99985294-99985294 on Build GRCh38
rs190038129 Chr.13: 99985256-99985256 on Build GRCh38
rs762829296 Chr.13: 99984929-99984929 on Build GRCh38
rs139312964 Chr.13: 99985127-99985127 on Build GRCh38
rs754464194 Chr.13: 99985033-99985033 on Build GRCh38
rs942000896 Chr.13: 99985217-99985218 on Build GRCh38
rs774943558 Chr.13: 99985300-99985300 on Build GRCh38
rs773003744 Chr.13: 99985279-99985279 on Build GRCh38

View Full Product Details