Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21520989-21521501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SALL2
Entrez Gene ID
6297
Gene Name
spalt like transcription factor 2
Gene Aliases
COLB, HSAL2, Sal-2, ZNF795, p150(Sal2)
UniGene
Hs.416358
Gene Chromosome Location
Chr.14: 21521080-21537216 on Build GRCh38
Transcripts
NM_001291446.1 NM_001291447.1 NM_005407.2 NR_111966.1 NR_111967.1 XM_011537064.1 XM_011537065.1

SNP Detail

SNP ID Chromosome Location
rs866725912 Chr.14: 21521359-21521359 on Build GRCh38
rs536254085 Chr.14: 21521197-21521197 on Build GRCh38
rs75678936 Chr.14: 21521340-21521341 on Build GRCh38
rs963422452 Chr.14: 21521072-21521072 on Build GRCh38
rs749346490 Chr.14: 21521303-21521303 on Build GRCh38
rs999518839 Chr.14: 21521034-21521034 on Build GRCh38
rs186693552 Chr.14: 21521390-21521390 on Build GRCh38
rs1009730266 Chr.14: 21521221-21521221 on Build GRCh38
rs1051843522 Chr.14: 21521344-21521344 on Build GRCh38
rs983119168 Chr.14: 21521129-21521129 on Build GRCh38
rs1034077130 Chr.14: 21521345-21521346 on Build GRCh38
rs770710161 Chr.14: 21521103-21521103 on Build GRCh38
rs781487609 Chr.14: 21521157-21521157 on Build GRCh38
rs1026175144 Chr.14: 21521428-21521428 on Build GRCh38
rs565972070 Chr.14: 21521181-21521181 on Build GRCh38
rs776596916 Chr.14: 21521193-21521193 on Build GRCh38
rs944079222 Chr.14: 21521099-21521099 on Build GRCh38
rs142086208 Chr.14: 21521337-21521338 on Build GRCh38
rs973467955 Chr.14: 21521080-21521080 on Build GRCh38
rs112977283 Chr.14: 21521173-21521173 on Build GRCh38
rs982720520 Chr.14: 21521418-21521425 on Build GRCh38
rs777780730 Chr.14: 21521243-21521243 on Build GRCh38
rs938520912 Chr.14: 21521174-21521174 on Build GRCh38
rs146140238 Chr.14: 21521035-21521035 on Build GRCh38
rs559663215 Chr.14: 21521312-21521312 on Build GRCh38
rs540657831 Chr.14: 21521386-21521386 on Build GRCh38
rs892740092 Chr.14: 21521220-21521220 on Build GRCh38
rs535252058 Chr.14: 21521111-21521111 on Build GRCh38
rs147552969 Chr.14: 21521311-21521312 on Build GRCh38
rs77544266 Chr.14: 21521059-21521059 on Build GRCh38
rs865836863 Chr.14: 21521342-21521342 on Build GRCh38
rs143426826 Chr.14: 21521385-21521385 on Build GRCh38
rs890133787 Chr.14: 21521357-21521357 on Build GRCh38
rs868832423 Chr.14: 21521321-21521321 on Build GRCh38
rs574186063 Chr.14: 21521290-21521290 on Build GRCh38
rs929920358 Chr.14: 21521417-21521417 on Build GRCh38
rs954892104 Chr.14: 21521040-21521040 on Build GRCh38
rs537571648 Chr.14: 21521254-21521254 on Build GRCh38
rs148905386 Chr.14: 21521069-21521069 on Build GRCh38
rs999141802 Chr.14: 21521343-21521343 on Build GRCh38
rs535066521 Chr.14: 21521182-21521182 on Build GRCh38
rs557128912 Chr.14: 21521261-21521261 on Build GRCh38
rs182797831 Chr.14: 21521218-21521218 on Build GRCh38
rs748301106 Chr.14: 21521188-21521188 on Build GRCh38
rs958343503 Chr.14: 21521345-21521345 on Build GRCh38
rs950673367 Chr.14: 21521431-21521431 on Build GRCh38
rs891206817 Chr.14: 21521073-21521076 on Build GRCh38
rs375943423 Chr.14: 21521310-21521310 on Build GRCh38
rs902807177 Chr.14: 21521028-21521028 on Build GRCh38
rs578169890 Chr.14: 21521226-21521226 on Build GRCh38
rs916044667 Chr.14: 21521293-21521293 on Build GRCh38
rs747208305 Chr.14: 21521083-21521083 on Build GRCh38
rs1032576907 Chr.14: 21521207-21521207 on Build GRCh38
rs542840896 Chr.14: 21521309-21521309 on Build GRCh38
rs970923266 Chr.14: 21521274-21521274 on Build GRCh38

View Full Product Details