Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 22775358-22775854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC7A7
Entrez Gene ID
9056
Gene Name
solute carrier family 7 member 7
Gene Aliases
LAT3, LPI, MOP-2, Y+LAT1, y+LAT-1
UniGene
Hs.513147
Gene Chromosome Location
Chr.14: 22773222-22819811 on Build GRCh38
Transcripts
NM_001126105.2 NM_001126106.2 NR_040448.1 XM_006720302.1 XM_011537298.2 XM_011537299.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3677929 c.999-2A>T p.? Chr.14: 22775542-22775542 on Build GRCh38
5815194 c.996T>C p.S332S Chr.14: 22775835-22775835 on Build GRCh38
4633883 c.1025G>A p.G342D Chr.14: 22775514-22775514 on Build GRCh38
3677930 c.999-2A>G p.? Chr.14: 22775542-22775542 on Build GRCh38
7310019 c.1044C>T p.I348I Chr.14: 22775495-22775495 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs386833797 Chr.14: 22775446-22775446 on Build GRCh38
rs760611547 Chr.14: 22775453-22775453 on Build GRCh38
rs11568423 Chr.14: 22775475-22775475 on Build GRCh38
rs1018799624 Chr.14: 22775391-22775391 on Build GRCh38
rs3751485 Chr.14: 22775720-22775720 on Build GRCh38
rs11568424 Chr.14: 22775535-22775535 on Build GRCh38
rs187221631 Chr.14: 22775697-22775697 on Build GRCh38
rs904360687 Chr.14: 22775702-22775702 on Build GRCh38
rs374581096 Chr.14: 22775585-22775585 on Build GRCh38
rs796228960 Chr.14: 22775736-22775736 on Build GRCh38
rs145897110 Chr.14: 22775750-22775750 on Build GRCh38
rs768481415 Chr.14: 22775456-22775456 on Build GRCh38
rs767798496 Chr.14: 22775814-22775814 on Build GRCh38
rs531862402 Chr.14: 22775438-22775438 on Build GRCh38
rs762508285 Chr.14: 22775793-22775793 on Build GRCh38
rs777732498 Chr.14: 22775543-22775543 on Build GRCh38
rs775426220 Chr.14: 22775445-22775445 on Build GRCh38
rs777873680 Chr.14: 22775423-22775423 on Build GRCh38
rs758005257 Chr.14: 22775396-22775396 on Build GRCh38
rs28365818 Chr.14: 22775734-22775734 on Build GRCh38
rs766087313 Chr.14: 22775509-22775509 on Build GRCh38
rs771215665 Chr.14: 22775444-22775444 on Build GRCh38
rs368801968 Chr.14: 22775405-22775405 on Build GRCh38
rs759190824 Chr.14: 22775809-22775809 on Build GRCh38
rs754387608 Chr.14: 22775829-22775829 on Build GRCh38
rs748294661 Chr.14: 22775807-22775807 on Build GRCh38
rs773129400 Chr.14: 22775788-22775788 on Build GRCh38
rs534044676 Chr.14: 22775613-22775613 on Build GRCh38
rs769713964 Chr.14: 22775787-22775787 on Build GRCh38
rs202099370 Chr.14: 22775584-22775584 on Build GRCh38
rs753043927 Chr.14: 22775823-22775823 on Build GRCh38
rs879129760 Chr.14: 22775600-22775600 on Build GRCh38
rs568001318 Chr.14: 22775411-22775411 on Build GRCh38
rs781609196 Chr.14: 22775409-22775409 on Build GRCh38
rs746063195 Chr.14: 22775575-22775575 on Build GRCh38
rs111266433 Chr.14: 22775615-22775615 on Build GRCh38
rs554612459 Chr.14: 22775828-22775828 on Build GRCh38
rs386833828 Chr.14: 22775832-22775832 on Build GRCh38
rs748058328 Chr.14: 22775591-22775591 on Build GRCh38
rs146720775 Chr.14: 22775540-22775540 on Build GRCh38
rs765855443 Chr.14: 22775805-22775805 on Build GRCh38
rs761778207 Chr.14: 22775476-22775476 on Build GRCh38
rs775431657 Chr.14: 22775745-22775745 on Build GRCh38
rs779332809 Chr.14: 22775574-22775574 on Build GRCh38
rs961921504 Chr.14: 22775768-22775768 on Build GRCh38
rs891675293 Chr.14: 22775602-22775602 on Build GRCh38
rs983482972 Chr.14: 22775735-22775735 on Build GRCh38
rs45467196 Chr.14: 22775729-22775729 on Build GRCh38
rs749310348 Chr.14: 22775555-22775555 on Build GRCh38
rs386833794 Chr.14: 22775531-22775534 on Build GRCh38
rs149755922 Chr.14: 22775530-22775530 on Build GRCh38
rs753178470 Chr.14: 22775531-22775531 on Build GRCh38
rs72552272 Chr.14: 22775538-22775538 on Build GRCh38
rs769807346 Chr.14: 22775420-22775420 on Build GRCh38
rs190239183 Chr.14: 22775416-22775416 on Build GRCh38
rs755387204 Chr.14: 22775527-22775527 on Build GRCh38
rs980824268 Chr.14: 22775401-22775401 on Build GRCh38
rs749885602 Chr.14: 22775500-22775500 on Build GRCh38
rs386833829 Chr.14: 22775833-22775833 on Build GRCh38
rs11568425 Chr.14: 22775406-22775406 on Build GRCh38
rs964451845 Chr.14: 22775392-22775392 on Build GRCh38
rs935978380 Chr.14: 22775550-22775550 on Build GRCh38
rs764683947 Chr.14: 22775485-22775485 on Build GRCh38
rs780419456 Chr.14: 22775403-22775403 on Build GRCh38
rs943138656 Chr.14: 22775645-22775645 on Build GRCh38
rs386833795 Chr.14: 22775526-22775526 on Build GRCh38
rs757404381 Chr.14: 22775565-22775565 on Build GRCh38
rs776446065 Chr.14: 22775588-22775588 on Build GRCh38
rs1018789932 Chr.14: 22775650-22775650 on Build GRCh38
rs762598998 Chr.14: 22775506-22775506 on Build GRCh38
rs267603944 Chr.14: 22775489-22775489 on Build GRCh38
rs919514252 Chr.14: 22775544-22775544 on Build GRCh38
rs987082748 Chr.14: 22775606-22775606 on Build GRCh38

View Full Product Details