Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23027757-23028301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMB5
Entrez Gene ID
5693
Gene Name
proteasome subunit beta 5
Gene Aliases
LMPX, MB1, X
UniGene
Hs.422990
Gene Chromosome Location
Chr.14: 23016543-23035220 on Build GRCh38
Transcripts
NM_001130725.1 NM_001144932.2 NM_002797.4 XM_005267871.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5005214 c.577G>A p.G193S Chr.14: 23027758-23027758 on Build GRCh38
954600 c.574A>G p.I192V Chr.14: 23027761-23027761 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs574671552 Chr.14: 23027961-23027961 on Build GRCh38
rs907756759 Chr.14: 23027936-23027936 on Build GRCh38
rs1009451280 Chr.14: 23027806-23027806 on Build GRCh38
rs559980291 Chr.14: 23028057-23028057 on Build GRCh38
rs562253292 Chr.14: 23028105-23028105 on Build GRCh38
rs983341316 Chr.14: 23027917-23027917 on Build GRCh38
rs991503976 Chr.14: 23027834-23027834 on Build GRCh38
rs138518815 Chr.14: 23028147-23028148 on Build GRCh38
rs60261589 Chr.14: 23028161-23028162 on Build GRCh38
rs1019046553 Chr.14: 23027829-23027829 on Build GRCh38
rs568620647 Chr.14: 23028193-23028193 on Build GRCh38
rs756273001 Chr.14: 23028265-23028265 on Build GRCh38
rs764346385 Chr.14: 23027843-23027843 on Build GRCh38
rs544115890 Chr.14: 23027886-23027886 on Build GRCh38
rs960896643 Chr.14: 23028163-23028163 on Build GRCh38
rs1027654287 Chr.14: 23028090-23028090 on Build GRCh38
rs750622250 Chr.14: 23028089-23028089 on Build GRCh38
rs887793613 Chr.14: 23028092-23028092 on Build GRCh38
rs916020460 Chr.14: 23027840-23027840 on Build GRCh38
rs1023888032 Chr.14: 23028220-23028220 on Build GRCh38
rs934949055 Chr.14: 23028014-23028014 on Build GRCh38
rs1043452329 Chr.14: 23028011-23028011 on Build GRCh38
rs532003129 Chr.14: 23028062-23028062 on Build GRCh38
rs559244793 Chr.14: 23027801-23027801 on Build GRCh38
rs901315374 Chr.14: 23027903-23027903 on Build GRCh38
rs114527385 Chr.14: 23028197-23028197 on Build GRCh38
rs529813747 Chr.14: 23028004-23028004 on Build GRCh38
rs891970310 Chr.14: 23027796-23027796 on Build GRCh38
rs764500350 Chr.14: 23027861-23027861 on Build GRCh38
rs1053376884 Chr.14: 23028015-23028015 on Build GRCh38
rs1003836143 Chr.14: 23028077-23028077 on Build GRCh38
rs576439979 Chr.14: 23027869-23027869 on Build GRCh38
rs113107529 Chr.14: 23028146-23028147 on Build GRCh38
rs752008113 Chr.14: 23028003-23028003 on Build GRCh38
rs548549722 Chr.14: 23028166-23028166 on Build GRCh38
rs952001505 Chr.14: 23027909-23027909 on Build GRCh38
rs916361470 Chr.14: 23027987-23027987 on Build GRCh38
rs915989438 Chr.14: 23028090-23028090 on Build GRCh38
rs545347493 Chr.14: 23028085-23028085 on Build GRCh38
rs976981123 Chr.14: 23028135-23028136 on Build GRCh38
rs979185562 Chr.14: 23028238-23028238 on Build GRCh38
rs138958911 Chr.14: 23028030-23028030 on Build GRCh38
rs1049485111 Chr.14: 23027960-23027960 on Build GRCh38
rs1009090536 Chr.14: 23028043-23028043 on Build GRCh38
rs940229643 Chr.14: 23027872-23027872 on Build GRCh38
rs193207409 Chr.14: 23027947-23027947 on Build GRCh38
rs1034913077 Chr.14: 23028081-23028081 on Build GRCh38
rs757534106 Chr.14: 23028067-23028067 on Build GRCh38
rs760874814 Chr.14: 23027842-23027842 on Build GRCh38
rs1041329435 Chr.14: 23027897-23027897 on Build GRCh38
rs924944521 Chr.14: 23028012-23028012 on Build GRCh38
rs531425820 Chr.14: 23028111-23028111 on Build GRCh38
rs867263099 Chr.14: 23027908-23027908 on Build GRCh38
rs149319993 Chr.14: 23028229-23028229 on Build GRCh38
rs893573671 Chr.14: 23028006-23028006 on Build GRCh38
rs952947217 Chr.14: 23028098-23028098 on Build GRCh38
rs940051881 Chr.14: 23028121-23028121 on Build GRCh38

View Full Product Details