Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23049508-23050013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH24
Entrez Gene ID
64403
Gene Name
cadherin 24
Gene Aliases
CDH11L
UniGene
Hs.155912
Gene Chromosome Location
Chr.14: 23047061-23057619 on Build GRCh38
Transcripts
NM_022478.3 NM_144985.3 XM_011537089.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6998419 c.1599+6C>A p.? Chr.14: 23049816-23049816 on Build GRCh38
1587338 c.1659C>A p.S553S Chr.14: 23049679-23049679 on Build GRCh38
3983181 c.1691T>A p.F564Y Chr.14: 23049647-23049647 on Build GRCh38
3983182 c.1577T>A p.F526Y Chr.14: 23049647-23049647 on Build GRCh38
697482 c.1387G>A p.V463M Chr.14: 23049920-23049920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759411852 Chr.14: 23049598-23049598 on Build GRCh38
rs769567390 Chr.14: 23049952-23049952 on Build GRCh38
rs762619392 Chr.14: 23049855-23049855 on Build GRCh38
rs763449945 Chr.14: 23049792-23049792 on Build GRCh38
rs763699965 Chr.14: 23049622-23049622 on Build GRCh38
rs370300266 Chr.14: 23049775-23049775 on Build GRCh38
rs749280643 Chr.14: 23049918-23049918 on Build GRCh38
rs201957516 Chr.14: 23049745-23049745 on Build GRCh38
rs752949288 Chr.14: 23049983-23049983 on Build GRCh38
rs190871483 Chr.14: 23049781-23049781 on Build GRCh38
rs757924327 Chr.14: 23049749-23049749 on Build GRCh38
rs769727328 Chr.14: 23049692-23049692 on Build GRCh38
rs775446662 Chr.14: 23049812-23049812 on Build GRCh38
rs765849346 Chr.14: 23049970-23049970 on Build GRCh38
rs773527989 Chr.14: 23049702-23049702 on Build GRCh38
rs564399448 Chr.14: 23049964-23049964 on Build GRCh38
rs760700341 Chr.14: 23049729-23049729 on Build GRCh38
rs977491107 Chr.14: 23049701-23049701 on Build GRCh38
rs776628061 Chr.14: 23049586-23049586 on Build GRCh38
rs761269290 Chr.14: 23049988-23049988 on Build GRCh38
rs55783852 Chr.14: 23049965-23049965 on Build GRCh38
rs1028358495 Chr.14: 23049529-23049529 on Build GRCh38
rs750022789 Chr.14: 23049655-23049655 on Build GRCh38
rs984046715 Chr.14: 23049565-23049565 on Build GRCh38
rs772057457 Chr.14: 23049715-23049715 on Build GRCh38
rs952315623 Chr.14: 23049541-23049541 on Build GRCh38
rs776221201 Chr.14: 23049924-23049924 on Build GRCh38
rs780248705 Chr.14: 23049984-23049987 on Build GRCh38
rs767191305 Chr.14: 23049604-23049604 on Build GRCh38
rs745629706 Chr.14: 23049607-23049607 on Build GRCh38
rs958828514 Chr.14: 23049866-23049866 on Build GRCh38
rs898157444 Chr.14: 23049773-23049773 on Build GRCh38
rs558066396 Chr.14: 23049687-23049687 on Build GRCh38
rs765979042 Chr.14: 23049857-23049857 on Build GRCh38
rs779479496 Chr.14: 23049967-23049977 on Build GRCh38
rs756508966 Chr.14: 23049896-23049896 on Build GRCh38
rs774048775 Chr.14: 23049593-23049593 on Build GRCh38
rs1039555352 Chr.14: 23049567-23049567 on Build GRCh38
rs550085379 Chr.14: 23049529-23049537 on Build GRCh38
rs749164584 Chr.14: 23049888-23049888 on Build GRCh38
rs145152245 Chr.14: 23049580-23049580 on Build GRCh38
rs933452073 Chr.14: 23049717-23049717 on Build GRCh38
rs145585228 Chr.14: 23049629-23049629 on Build GRCh38
rs914498749 Chr.14: 23049632-23049632 on Build GRCh38
rs372231304 Chr.14: 23049807-23049807 on Build GRCh38
rs761435199 Chr.14: 23049744-23049744 on Build GRCh38
rs2295689 Chr.14: 23049582-23049582 on Build GRCh38
rs770013051 Chr.14: 23049591-23049591 on Build GRCh38
rs767931473 Chr.14: 23049982-23049982 on Build GRCh38
rs768549590 Chr.14: 23049817-23049817 on Build GRCh38
rs778667953 Chr.14: 23049642-23049642 on Build GRCh38
rs751876578 Chr.14: 23049770-23049770 on Build GRCh38
rs750158749 Chr.14: 23049746-23049746 on Build GRCh38
rs757584877 Chr.14: 23049920-23049920 on Build GRCh38
rs748127257 Chr.14: 23049940-23049940 on Build GRCh38
rs146177987 Chr.14: 23049610-23049611 on Build GRCh38
rs368568578 Chr.14: 23049793-23049793 on Build GRCh38
rs34629744 Chr.14: 23049805-23049806 on Build GRCh38
rs768637763 Chr.14: 23049923-23049923 on Build GRCh38
rs746760919 Chr.14: 23049816-23049816 on Build GRCh38
rs746311128 Chr.14: 23049973-23049974 on Build GRCh38
rs367754433 Chr.14: 23049743-23049743 on Build GRCh38
rs149513148 Chr.14: 23049846-23049846 on Build GRCh38
rs761225766 Chr.14: 23049872-23049872 on Build GRCh38
rs184060458 Chr.14: 23049821-23049821 on Build GRCh38
rs769096080 Chr.14: 23049670-23049670 on Build GRCh38
rs751185577 Chr.14: 23049858-23049858 on Build GRCh38
rs548759407 Chr.14: 23049574-23049574 on Build GRCh38
rs780395008 Chr.14: 23049666-23049666 on Build GRCh38
rs528997143 Chr.14: 23049545-23049545 on Build GRCh38
rs908010960 Chr.14: 23049572-23049572 on Build GRCh38
rs373643580 Chr.14: 23049825-23049825 on Build GRCh38
rs749528489 Chr.14: 23049801-23049801 on Build GRCh38
rs758856229 Chr.14: 23049662-23049662 on Build GRCh38
rs200472203 Chr.14: 23049722-23049722 on Build GRCh38
rs371616736 Chr.14: 23049910-23049910 on Build GRCh38
rs747522364 Chr.14: 23049669-23049669 on Build GRCh38
rs11623976 Chr.14: 23049709-23049709 on Build GRCh38
rs371296538 Chr.14: 23049971-23049971 on Build GRCh38
rs1012828785 Chr.14: 23049909-23049909 on Build GRCh38
rs759188119 Chr.14: 23049972-23049972 on Build GRCh38
rs374685642 Chr.14: 23049617-23049617 on Build GRCh38
rs1051950574 Chr.14: 23049723-23049723 on Build GRCh38
rs796178457 Chr.14: 23049580-23049582 on Build GRCh38
rs764074182 Chr.14: 23049741-23049741 on Build GRCh38
rs144262661 Chr.14: 23049862-23049862 on Build GRCh38
rs755513709 Chr.14: 23049772-23049772 on Build GRCh38
rs766746350 Chr.14: 23049761-23049761 on Build GRCh38
rs1038397548 Chr.14: 23049752-23049752 on Build GRCh38
rs781276382 Chr.14: 23049679-23049679 on Build GRCh38
rs374162864 Chr.14: 23049840-23049840 on Build GRCh38
rs756882371 Chr.14: 23049630-23049630 on Build GRCh38
rs753311467 Chr.14: 23049891-23049891 on Build GRCh38
rs199721652 Chr.14: 23049900-23049900 on Build GRCh38
rs368443534 Chr.14: 23049884-23049884 on Build GRCh38
rs116261083 Chr.14: 23049922-23049922 on Build GRCh38
rs761652414 Chr.14: 23049834-23049834 on Build GRCh38
rs796100532 Chr.14: 23049973-23049974 on Build GRCh38
rs375343397 Chr.14: 23049939-23049939 on Build GRCh38
rs572219131 Chr.14: 23049921-23049921 on Build GRCh38

View Full Product Details