Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23323214-23323712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL2L2-PABPN1
Entrez Gene ID
100529063
Gene Name
BCL2L2-PABPN1 readthrough
Gene Aliases
-
UniGene
Hs.707712
Gene Chromosome Location
Chr.14: 23306762-23326185 on Build GRCh38
Transcripts
NM_001199864.1 NM_004643.3

Gene Symbol
PABPN1
Entrez Gene ID
8106
Gene Name
poly(A) binding protein nuclear 1
Gene Aliases
OPMD, PAB2, PABII, PABP-2, PABP2
UniGene
Hs.707712
Gene Chromosome Location
Chr.14: 23320188-23326185 on Build GRCh38
Transcripts
NM_001199864.1 NM_004643.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3793589 c.590C>T p.S197L Chr.14: 23323432-23323432 on Build GRCh38
5729540 c.561G>A p.E187E Chr.14: 23323403-23323403 on Build GRCh38
6140225 c.582C>G p.G194G Chr.14: 23323424-23323424 on Build GRCh38
6393943 c.540C>A p.D180E Chr.14: 23323382-23323382 on Build GRCh38
4150750 c.609A>G p.I203M Chr.14: 23323451-23323451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547257997 Chr.14: 23323533-23323533 on Build GRCh38
rs768480157 Chr.14: 23323333-23323333 on Build GRCh38
rs778932762 Chr.14: 23323493-23323493 on Build GRCh38
rs546717436 Chr.14: 23323681-23323681 on Build GRCh38
rs1014226955 Chr.14: 23323293-23323293 on Build GRCh38
rs750694446 Chr.14: 23323494-23323494 on Build GRCh38
rs56390381 Chr.14: 23323539-23323540 on Build GRCh38
rs905858671 Chr.14: 23323310-23323310 on Build GRCh38
rs966308605 Chr.14: 23323525-23323525 on Build GRCh38
rs949999321 Chr.14: 23323675-23323675 on Build GRCh38
rs777498186 Chr.14: 23323531-23323534 on Build GRCh38
rs895693418 Chr.14: 23323282-23323282 on Build GRCh38
rs900740535 Chr.14: 23323292-23323292 on Build GRCh38
rs747580389 Chr.14: 23323276-23323276 on Build GRCh38
rs757395524 Chr.14: 23323441-23323441 on Build GRCh38
rs895630660 Chr.14: 23323659-23323659 on Build GRCh38
rs767227035 Chr.14: 23323400-23323400 on Build GRCh38
rs551156807 Chr.14: 23323537-23323537 on Build GRCh38
rs753139856 Chr.14: 23323411-23323411 on Build GRCh38
rs1000269007 Chr.14: 23323313-23323313 on Build GRCh38
rs528706743 Chr.14: 23323300-23323300 on Build GRCh38
rs770557501 Chr.14: 23323369-23323369 on Build GRCh38
rs762608789 Chr.14: 23323367-23323367 on Build GRCh38
rs182822835 Chr.14: 23323255-23323255 on Build GRCh38
rs781020011 Chr.14: 23323344-23323344 on Build GRCh38
rs989358667 Chr.14: 23323498-23323498 on Build GRCh38
rs919552798 Chr.14: 23323539-23323539 on Build GRCh38
rs974980704 Chr.14: 23323528-23323528 on Build GRCh38
rs770612485 Chr.14: 23323531-23323531 on Build GRCh38
rs771616093 Chr.14: 23323567-23323567 on Build GRCh38
rs777830751 Chr.14: 23323516-23323516 on Build GRCh38
rs769774240 Chr.14: 23323364-23323364 on Build GRCh38
rs150035782 Chr.14: 23323424-23323424 on Build GRCh38
rs769580252 Chr.14: 23323513-23323517 on Build GRCh38
rs371658709 Chr.14: 23323436-23323436 on Build GRCh38
rs747808800 Chr.14: 23323508-23323508 on Build GRCh38
rs983874134 Chr.14: 23323605-23323605 on Build GRCh38
rs747857541 Chr.14: 23323360-23323360 on Build GRCh38
rs780748360 Chr.14: 23323503-23323503 on Build GRCh38
rs747988260 Chr.14: 23323506-23323506 on Build GRCh38
rs759237746 Chr.14: 23323385-23323385 on Build GRCh38
rs909589493 Chr.14: 23323615-23323615 on Build GRCh38
rs1040966039 Chr.14: 23323263-23323263 on Build GRCh38
rs373574548 Chr.14: 23323497-23323497 on Build GRCh38
rs1019214226 Chr.14: 23323512-23323512 on Build GRCh38
rs749202419 Chr.14: 23323517-23323517 on Build GRCh38
rs780071417 Chr.14: 23323500-23323500 on Build GRCh38
rs368017089 Chr.14: 23323336-23323336 on Build GRCh38
rs746941529 Chr.14: 23323352-23323352 on Build GRCh38
rs981278088 Chr.14: 23323468-23323468 on Build GRCh38
rs774184575 Chr.14: 23323374-23323374 on Build GRCh38
rs145369012 Chr.14: 23323452-23323452 on Build GRCh38
rs754485523 Chr.14: 23323493-23323495 on Build GRCh38
rs116983161 Chr.14: 23323427-23323427 on Build GRCh38
rs78971167 Chr.14: 23323646-23323646 on Build GRCh38
rs956233098 Chr.14: 23323488-23323488 on Build GRCh38
rs772962837 Chr.14: 23323366-23323366 on Build GRCh38
rs1046770119 Chr.14: 23323660-23323660 on Build GRCh38
rs185949472 Chr.14: 23323616-23323616 on Build GRCh38
rs769462736 Chr.14: 23323511-23323511 on Build GRCh38

View Full Product Details