Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24179912-24180413 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPO4
Entrez Gene ID
79711
Gene Name
importin 4
Gene Aliases
Imp4
UniGene
Hs.411865
Gene Chromosome Location
Chr.14: 24180219-24188918 on Build GRCh38
Transcripts
NM_024658.3 NR_051979.1 NM_001048205.1 NM_005132.2 XM_011537421.1 XM_017021841.1

Gene Symbol
REC8
Entrez Gene ID
9985
Gene Name
REC8 meiotic recombination protein
Gene Aliases
HR21spB, REC8L1, Rec8p
UniGene
Hs.419259
Gene Chromosome Location
Chr.14: 24171853-24180408 on Build GRCh38
Transcripts
NM_024658.3 NR_051979.1 NM_001048205.1 NM_005132.2 XM_011537421.1 XM_017021841.1

SNP Detail

SNP ID Chromosome Location
rs751554206 Chr.14: 24179991-24179991 on Build GRCh38
rs771590672 Chr.14: 24180024-24180024 on Build GRCh38
rs759743941 Chr.14: 24180180-24180180 on Build GRCh38
rs768750032 Chr.14: 24180147-24180148 on Build GRCh38
rs751313539 Chr.14: 24180218-24180218 on Build GRCh38
rs929736941 Chr.14: 24180370-24180370 on Build GRCh38
rs943522879 Chr.14: 24180335-24180335 on Build GRCh38
rs779919167 Chr.14: 24180106-24180107 on Build GRCh38
rs766464041 Chr.14: 24180089-24180089 on Build GRCh38
rs919340715 Chr.14: 24180269-24180269 on Build GRCh38
rs765558861 Chr.14: 24179974-24179974 on Build GRCh38
rs762210672 Chr.14: 24179969-24179969 on Build GRCh38
rs758061425 Chr.14: 24180204-24180204 on Build GRCh38
rs763270288 Chr.14: 24180077-24180077 on Build GRCh38
rs770387502 Chr.14: 24180161-24180161 on Build GRCh38
rs771263350 Chr.14: 24180391-24180391 on Build GRCh38
rs777211905 Chr.14: 24180060-24180060 on Build GRCh38
rs933772529 Chr.14: 24180198-24180198 on Build GRCh38
rs756099867 Chr.14: 24179995-24179995 on Build GRCh38
rs756176491 Chr.14: 24180012-24180013 on Build GRCh38
rs1038080102 Chr.14: 24180339-24180339 on Build GRCh38
rs757167272 Chr.14: 24180138-24180138 on Build GRCh38
rs746738329 Chr.14: 24180143-24180143 on Build GRCh38
rs780882093 Chr.14: 24180241-24180241 on Build GRCh38
rs777633723 Chr.14: 24179997-24179997 on Build GRCh38
rs758088389 Chr.14: 24180149-24180149 on Build GRCh38
rs754098387 Chr.14: 24180100-24180101 on Build GRCh38
rs547453462 Chr.14: 24180262-24180262 on Build GRCh38
rs994320459 Chr.14: 24179941-24179941 on Build GRCh38
rs377245718 Chr.14: 24180019-24180019 on Build GRCh38
rs771192524 Chr.14: 24180171-24180171 on Build GRCh38
rs747615290 Chr.14: 24180257-24180257 on Build GRCh38
rs750242580 Chr.14: 24180203-24180203 on Build GRCh38
rs766522572 Chr.14: 24179987-24179987 on Build GRCh38
rs754653566 Chr.14: 24180221-24180221 on Build GRCh38
rs757367742 Chr.14: 24180099-24180100 on Build GRCh38
rs780835045 Chr.14: 24180158-24180158 on Build GRCh38
rs556424331 Chr.14: 24179980-24179980 on Build GRCh38
rs568707414 Chr.14: 24180188-24180188 on Build GRCh38
rs765031853 Chr.14: 24180202-24180202 on Build GRCh38
rs773419032 Chr.14: 24180076-24180076 on Build GRCh38
rs764652784 Chr.14: 24179956-24179956 on Build GRCh38
rs748706917 Chr.14: 24180160-24180160 on Build GRCh38
rs750142900 Chr.14: 24180148-24180148 on Build GRCh38
rs374010286 Chr.14: 24180062-24180062 on Build GRCh38
rs769299580 Chr.14: 24180050-24180050 on Build GRCh38
rs532503774 Chr.14: 24180002-24180002 on Build GRCh38
rs183161120 Chr.14: 24180168-24180168 on Build GRCh38
rs779642513 Chr.14: 24180030-24180030 on Build GRCh38
rs757113882 Chr.14: 24180016-24180016 on Build GRCh38
rs1051791051 Chr.14: 24180301-24180301 on Build GRCh38
rs774507508 Chr.14: 24180174-24180174 on Build GRCh38
rs759549808 Chr.14: 24180123-24180123 on Build GRCh38
rs552406292 Chr.14: 24180020-24180020 on Build GRCh38
rs778717822 Chr.14: 24180144-24180144 on Build GRCh38
rs778041288 Chr.14: 24180023-24180024 on Build GRCh38
rs142040563 Chr.14: 24179964-24179964 on Build GRCh38
rs911170954 Chr.14: 24180263-24180263 on Build GRCh38
rs753639978 Chr.14: 24180003-24180003 on Build GRCh38
rs746532831 Chr.14: 24180152-24180152 on Build GRCh38
rs1048559 Chr.14: 24180349-24180349 on Build GRCh38
rs889311988 Chr.14: 24180313-24180313 on Build GRCh38
rs372394424 Chr.14: 24180097-24180097 on Build GRCh38
rs202088503 Chr.14: 24179981-24179981 on Build GRCh38
rs749801587 Chr.14: 24180276-24180276 on Build GRCh38
rs756702300 Chr.14: 24180259-24180259 on Build GRCh38
rs531823084 Chr.14: 24180165-24180165 on Build GRCh38
rs878856185 Chr.14: 24180105-24180105 on Build GRCh38
rs904744483 Chr.14: 24180385-24180385 on Build GRCh38
rs371180320 Chr.14: 24180182-24180199 on Build GRCh38
rs1048554 Chr.14: 24180366-24180366 on Build GRCh38
rs1034541946 Chr.14: 24180034-24180034 on Build GRCh38
rs761767696 Chr.14: 24180200-24180200 on Build GRCh38
rs200300723 Chr.14: 24179965-24179965 on Build GRCh38
rs775514345 Chr.14: 24180197-24180197 on Build GRCh38
rs779514559 Chr.14: 24180150-24180150 on Build GRCh38
rs368937002 Chr.14: 24180061-24180061 on Build GRCh38
rs770338744 Chr.14: 24180075-24180075 on Build GRCh38
rs768345916 Chr.14: 24180154-24180154 on Build GRCh38
rs1045420025 Chr.14: 24180374-24180374 on Build GRCh38
rs753833388 Chr.14: 24180130-24180130 on Build GRCh38
rs941257736 Chr.14: 24180271-24180271 on Build GRCh38
rs777157025 Chr.14: 24179962-24179962 on Build GRCh38
rs372852976 Chr.14: 24180129-24180129 on Build GRCh38
rs956448127 Chr.14: 24180101-24180101 on Build GRCh38
rs752823808 Chr.14: 24179976-24179976 on Build GRCh38
rs62001573 Chr.14: 24180208-24180208 on Build GRCh38
rs574899082 Chr.14: 24180035-24180035 on Build GRCh38

View Full Product Details