Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24316243-24316767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LTB4R
Entrez Gene ID
1241
Gene Name
leukotriene B4 receptor
Gene Aliases
BLT1, BLTR, CMKRL1, GPR16, LTB4R1, LTBR1, P2RY7, P2Y7
UniGene
Hs.567248
Gene Chromosome Location
Chr.14: 24311499-24318036 on Build GRCh38
Transcripts
NM_001143919.2 NM_181657.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6554255 c.813C>T p.I271I Chr.14: 24316464-24316464 on Build GRCh38
5770501 c.987C>T p.P329P Chr.14: 24316638-24316638 on Build GRCh38
6430259 c.892G>A p.V298M Chr.14: 24316543-24316543 on Build GRCh38
4988039 c.778G>C p.G260R Chr.14: 24316429-24316429 on Build GRCh38
6521384 c.1005C>A p.G335G Chr.14: 24316656-24316656 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1050073241 Chr.14: 24316640-24316640 on Build GRCh38
rs567553911 Chr.14: 24316268-24316268 on Build GRCh38
rs202233555 Chr.14: 24316429-24316429 on Build GRCh38
rs781570076 Chr.14: 24316649-24316649 on Build GRCh38
rs772461943 Chr.14: 24316326-24316326 on Build GRCh38
rs542278468 Chr.14: 24316738-24316738 on Build GRCh38
rs754794828 Chr.14: 24316299-24316299 on Build GRCh38
rs758960291 Chr.14: 24316696-24316696 on Build GRCh38
rs920018399 Chr.14: 24316536-24316536 on Build GRCh38
rs763877140 Chr.14: 24316735-24316735 on Build GRCh38
rs1035732098 Chr.14: 24316462-24316462 on Build GRCh38
rs892878354 Chr.14: 24316670-24316671 on Build GRCh38
rs761810548 Chr.14: 24316347-24316347 on Build GRCh38
rs17849863 Chr.14: 24316645-24316645 on Build GRCh38
rs758961667 Chr.14: 24316534-24316534 on Build GRCh38
rs1025111534 Chr.14: 24316624-24316624 on Build GRCh38
rs951923898 Chr.14: 24316344-24316344 on Build GRCh38
rs754775733 Chr.14: 24316366-24316366 on Build GRCh38
rs746277997 Chr.14: 24316324-24316324 on Build GRCh38
rs868745159 Chr.14: 24316430-24316430 on Build GRCh38
rs751311323 Chr.14: 24316292-24316292 on Build GRCh38
rs778518761 Chr.14: 24316531-24316531 on Build GRCh38
rs535078695 Chr.14: 24316468-24316468 on Build GRCh38
rs748414268 Chr.14: 24316660-24316660 on Build GRCh38
rs775554959 Chr.14: 24316603-24316603 on Build GRCh38
rs756373678 Chr.14: 24316661-24316661 on Build GRCh38
rs764735193 Chr.14: 24316628-24316628 on Build GRCh38
rs747444595 Chr.14: 24316543-24316543 on Build GRCh38
rs752042885 Chr.14: 24316641-24316641 on Build GRCh38
rs370069608 Chr.14: 24316315-24316315 on Build GRCh38
rs908703838 Chr.14: 24316584-24316584 on Build GRCh38
rs777510348 Chr.14: 24316383-24316383 on Build GRCh38
rs776495849 Chr.14: 24316717-24316717 on Build GRCh38
rs768875034 Chr.14: 24316451-24316451 on Build GRCh38
rs936913092 Chr.14: 24316659-24316659 on Build GRCh38
rs200462048 Chr.14: 24316678-24316678 on Build GRCh38
rs868502219 Chr.14: 24316488-24316488 on Build GRCh38
rs760921281 Chr.14: 24316332-24316332 on Build GRCh38
rs775731303 Chr.14: 24316330-24316330 on Build GRCh38
rs540931037 Chr.14: 24316606-24316606 on Build GRCh38
rs901539899 Chr.14: 24316620-24316620 on Build GRCh38
rs760479124 Chr.14: 24316506-24316506 on Build GRCh38
rs767311299 Chr.14: 24316368-24316368 on Build GRCh38
rs780476851 Chr.14: 24316540-24316540 on Build GRCh38
rs747778454 Chr.14: 24316309-24316309 on Build GRCh38
rs776709653 Chr.14: 24316345-24316345 on Build GRCh38
rs779338558 Chr.14: 24316402-24316402 on Build GRCh38
rs749855593 Chr.14: 24316532-24316532 on Build GRCh38
rs576783203 Chr.14: 24316535-24316535 on Build GRCh38
rs752499151 Chr.14: 24316374-24316374 on Build GRCh38
rs1041144853 Chr.14: 24316730-24316730 on Build GRCh38
rs1020033537 Chr.14: 24316406-24316406 on Build GRCh38
rs757895009 Chr.14: 24316639-24316639 on Build GRCh38
rs780421759 Chr.14: 24316425-24316425 on Build GRCh38
rs577864624 Chr.14: 24316524-24316524 on Build GRCh38
rs761547296 Chr.14: 24316625-24316625 on Build GRCh38
rs756779238 Chr.14: 24316392-24316392 on Build GRCh38
rs774917821 Chr.14: 24316317-24316317 on Build GRCh38
rs1014589629 Chr.14: 24316465-24316465 on Build GRCh38
rs772421776 Chr.14: 24316423-24316423 on Build GRCh38
rs769892544 Chr.14: 24316459-24316459 on Build GRCh38
rs369807879 Chr.14: 24316587-24316587 on Build GRCh38
rs765563648 Chr.14: 24316444-24316444 on Build GRCh38
rs755379086 Chr.14: 24316646-24316646 on Build GRCh38
rs761445609 Chr.14: 24316725-24316725 on Build GRCh38
rs766285559 Chr.14: 24316287-24316287 on Build GRCh38
rs772141314 Chr.14: 24316601-24316601 on Build GRCh38
rs867687139 Chr.14: 24316485-24316485 on Build GRCh38
rs748874512 Chr.14: 24316390-24316390 on Build GRCh38
rs776826117 Chr.14: 24316456-24316456 on Build GRCh38
rs901504514 Chr.14: 24316308-24316308 on Build GRCh38
rs759364766 Chr.14: 24316685-24316686 on Build GRCh38
rs144146055 Chr.14: 24316281-24316281 on Build GRCh38
rs780762185 Chr.14: 24316300-24316300 on Build GRCh38
rs200942984 Chr.14: 24316515-24316515 on Build GRCh38
rs749451264 Chr.14: 24316669-24316669 on Build GRCh38
rs142918543 Chr.14: 24316564-24316564 on Build GRCh38
rs751498547 Chr.14: 24316356-24316356 on Build GRCh38
rs748374182 Chr.14: 24316571-24316571 on Build GRCh38
rs549333517 Chr.14: 24316393-24316393 on Build GRCh38
rs753878361 Chr.14: 24316273-24316273 on Build GRCh38
rs768664022 Chr.14: 24316683-24316683 on Build GRCh38
rs17849864 Chr.14: 24316687-24316687 on Build GRCh38
rs774063017 Chr.14: 24316290-24316298 on Build GRCh38
rs755852752 Chr.14: 24316378-24316378 on Build GRCh38
rs752552509 Chr.14: 24316503-24316503 on Build GRCh38
rs1046584 Chr.14: 24316578-24316578 on Build GRCh38
rs771113104 Chr.14: 24316675-24316675 on Build GRCh38
rs769868736 Chr.14: 24316572-24316572 on Build GRCh38
rs771355497 Chr.14: 24316316-24316316 on Build GRCh38
rs774193539 Chr.14: 24316467-24316467 on Build GRCh38
rs749950313 Chr.14: 24316629-24316629 on Build GRCh38
rs766290486 Chr.14: 24316353-24316353 on Build GRCh38
rs755723597 Chr.14: 24316312-24316312 on Build GRCh38
rs753534160 Chr.14: 24316622-24316622 on Build GRCh38
rs781438982 Chr.14: 24316566-24316566 on Build GRCh38
rs767511518 Chr.14: 24316470-24316470 on Build GRCh38
rs763949514 Chr.14: 24316617-24316617 on Build GRCh38
rs981326485 Chr.14: 24316632-24316632 on Build GRCh38
rs777455355 Chr.14: 24316314-24316314 on Build GRCh38
rs901682345 Chr.14: 24316310-24316310 on Build GRCh38
rs756759693 Chr.14: 24316525-24316525 on Build GRCh38
rs746356935 Chr.14: 24316405-24316405 on Build GRCh38
rs961830871 Chr.14: 24316478-24316478 on Build GRCh38
rs761852671 Chr.14: 24316457-24316457 on Build GRCh38
rs374250559 Chr.14: 24316341-24316341 on Build GRCh38

View Full Product Details