Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 31447382-31447929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTD2
Entrez Gene ID
112487
Gene Name
D-tyrosyl-tRNA deacylase 2 (putative)
Gene Aliases
C14orf126
UniGene
Hs.116014
Gene Chromosome Location
Chr.14: 31446036-31457474 on Build GRCh38
Transcripts
NM_080664.2 NR_110045.1

Gene Symbol
LOC101927124
Entrez Gene ID
101927124
Gene Name
uncharacterized LOC101927124
Gene Aliases
-
UniGene
Hs.647561
Gene Chromosome Location
Chr.14: 31420758-31452883 on Build GRCh38
Transcripts
NM_080664.2 NR_110045.1

SNP Detail

SNP ID Chromosome Location
rs950028872 Chr.14: 31447655-31447655 on Build GRCh38
rs139220962 Chr.14: 31447879-31447887 on Build GRCh38
rs576774889 Chr.14: 31447884-31447884 on Build GRCh38
rs116799502 Chr.14: 31447474-31447474 on Build GRCh38
rs919873671 Chr.14: 31447807-31447807 on Build GRCh38
rs190105818 Chr.14: 31447754-31447754 on Build GRCh38
rs1033717058 Chr.14: 31447896-31447896 on Build GRCh38
rs1020519119 Chr.14: 31447568-31447568 on Build GRCh38
rs1049592724 Chr.14: 31447617-31447617 on Build GRCh38
rs538117607 Chr.14: 31447806-31447806 on Build GRCh38
rs1028093071 Chr.14: 31447539-31447539 on Build GRCh38
rs371063842 Chr.14: 31447693-31447693 on Build GRCh38
rs560322533 Chr.14: 31447517-31447517 on Build GRCh38
rs569130196 Chr.14: 31447662-31447662 on Build GRCh38
rs553575727 Chr.14: 31447904-31447904 on Build GRCh38
rs1046267788 Chr.14: 31447413-31447413 on Build GRCh38
rs201669408 Chr.14: 31447820-31447821 on Build GRCh38
rs1019942057 Chr.14: 31447831-31447831 on Build GRCh38
rs567096303 Chr.14: 31447720-31447720 on Build GRCh38
rs555236716 Chr.14: 31447613-31447613 on Build GRCh38
rs898996052 Chr.14: 31447769-31447769 on Build GRCh38
rs146946063 Chr.14: 31447777-31447777 on Build GRCh38
rs953811515 Chr.14: 31447712-31447712 on Build GRCh38
rs115511128 Chr.14: 31447455-31447455 on Build GRCh38
rs375407282 Chr.14: 31447623-31447623 on Build GRCh38
rs187691331 Chr.14: 31447824-31447824 on Build GRCh38
rs979445163 Chr.14: 31447602-31447602 on Build GRCh38
rs76229484 Chr.14: 31447837-31447837 on Build GRCh38
rs938056501 Chr.14: 31447846-31447846 on Build GRCh38
rs186173564 Chr.14: 31447622-31447622 on Build GRCh38
rs7159566 Chr.14: 31447586-31447586 on Build GRCh38
rs555760239 Chr.14: 31447809-31447809 on Build GRCh38
rs979561548 Chr.14: 31447521-31447521 on Build GRCh38
rs1009738892 Chr.14: 31447637-31447637 on Build GRCh38
rs561162696 Chr.14: 31447604-31447604 on Build GRCh38
rs71115009 Chr.14: 31447466-31447467 on Build GRCh38
rs17097896 Chr.14: 31447491-31447491 on Build GRCh38
rs376777791 Chr.14: 31447636-31447636 on Build GRCh38
rs879823635 Chr.14: 31447694-31447694 on Build GRCh38
rs953842195 Chr.14: 31447541-31447541 on Build GRCh38
rs1020998852 Chr.14: 31447485-31447485 on Build GRCh38
rs972678912 Chr.14: 31447596-31447596 on Build GRCh38
rs577056098 Chr.14: 31447616-31447616 on Build GRCh38
rs986663037 Chr.14: 31447556-31447556 on Build GRCh38
rs905314211 Chr.14: 31447621-31447621 on Build GRCh38
rs61001235 Chr.14: 31447882-31447890 on Build GRCh38
rs549338098 Chr.14: 31447595-31447595 on Build GRCh38
rs755151361 Chr.14: 31447819-31447820 on Build GRCh38
rs182706379 Chr.14: 31447778-31447778 on Build GRCh38
rs967474885 Chr.14: 31447882-31447883 on Build GRCh38
rs77908938 Chr.14: 31447829-31447829 on Build GRCh38
rs954322068 Chr.14: 31447542-31447542 on Build GRCh38
rs763653914 Chr.14: 31447737-31447737 on Build GRCh38
rs551569517 Chr.14: 31447701-31447701 on Build GRCh38
rs896776992 Chr.14: 31447473-31447473 on Build GRCh38
rs886373275 Chr.14: 31447442-31447442 on Build GRCh38
rs926707611 Chr.14: 31447832-31447832 on Build GRCh38
rs1047069664 Chr.14: 31447684-31447684 on Build GRCh38

View Full Product Details