Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 35534266-35534759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INSM2
Entrez Gene ID
84684
Gene Name
INSM transcriptional repressor 2
Gene Aliases
IA-6, IA6, mlt1
UniGene
Hs.62813
Gene Chromosome Location
Chr.14: 35534042-35537054 on Build GRCh38
Transcripts
NM_032594.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6554322 c.422G>T p.G141V Chr.14: 35534674-35534674 on Build GRCh38
394130 c.55G>T p.V19F Chr.14: 35534307-35534307 on Build GRCh38
6419246 c.320G>A p.S107N Chr.14: 35534572-35534572 on Build GRCh38
3667640 c.438C>A p.A146A Chr.14: 35534690-35534690 on Build GRCh38
6433488 c.107C>T p.P36L Chr.14: 35534359-35534359 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772567825 Chr.14: 35534531-35534531 on Build GRCh38
rs748375530 Chr.14: 35534419-35534419 on Build GRCh38
rs757095420 Chr.14: 35534478-35534478 on Build GRCh38
rs1034751542 Chr.14: 35534326-35534326 on Build GRCh38
rs1047963497 Chr.14: 35534705-35534705 on Build GRCh38
rs975411830 Chr.14: 35534524-35534524 on Build GRCh38
rs755088826 Chr.14: 35534416-35534416 on Build GRCh38
rs758341952 Chr.14: 35534489-35534489 on Build GRCh38
rs979280943 Chr.14: 35534535-35534535 on Build GRCh38
rs572000158 Chr.14: 35534528-35534528 on Build GRCh38
rs921226936 Chr.14: 35534525-35534525 on Build GRCh38
rs750157549 Chr.14: 35534625-35534625 on Build GRCh38
rs777624337 Chr.14: 35534310-35534310 on Build GRCh38
rs865871973 Chr.14: 35534632-35534632 on Build GRCh38
rs924708184 Chr.14: 35534574-35534574 on Build GRCh38
rs768891344 Chr.14: 35534338-35534338 on Build GRCh38
rs915163696 Chr.14: 35534393-35534393 on Build GRCh38
rs750425595 Chr.14: 35534488-35534488 on Build GRCh38
rs769123084 Chr.14: 35534562-35534567 on Build GRCh38
rs763762326 Chr.14: 35534470-35534470 on Build GRCh38
rs1048008130 Chr.14: 35534540-35534540 on Build GRCh38
rs757218258 Chr.14: 35534404-35534404 on Build GRCh38
rs765072091 Chr.14: 35534616-35534616 on Build GRCh38
rs771585438 Chr.14: 35534424-35534424 on Build GRCh38
rs567275553 Chr.14: 35534677-35534677 on Build GRCh38
rs754679890 Chr.14: 35534288-35534288 on Build GRCh38
rs780662602 Chr.14: 35534733-35534733 on Build GRCh38
rs1056999224 Chr.14: 35534413-35534413 on Build GRCh38
rs551773653 Chr.14: 35534519-35534519 on Build GRCh38
rs780113915 Chr.14: 35534320-35534320 on Build GRCh38
rs763490919 Chr.14: 35534605-35534605 on Build GRCh38
rs752740930 Chr.14: 35534651-35534651 on Build GRCh38
rs112804872 Chr.14: 35534492-35534492 on Build GRCh38
rs776308477 Chr.14: 35534609-35534609 on Build GRCh38
rs771491594 Chr.14: 35534517-35534517 on Build GRCh38
rs776826954 Chr.14: 35534340-35534340 on Build GRCh38
rs777998832 Chr.14: 35534658-35534658 on Build GRCh38
rs371623805 Chr.14: 35534305-35534305 on Build GRCh38
rs761448230 Chr.14: 35534612-35534612 on Build GRCh38
rs766299697 Chr.14: 35534648-35534648 on Build GRCh38
rs762800280 Chr.14: 35534641-35534641 on Build GRCh38
rs770797194 Chr.14: 35534318-35534318 on Build GRCh38
rs199890483 Chr.14: 35534370-35534370 on Build GRCh38
rs557031979 Chr.14: 35534467-35534467 on Build GRCh38
rs746199188 Chr.14: 35534711-35534711 on Build GRCh38
rs868255521 Chr.14: 35534296-35534296 on Build GRCh38
rs1026451359 Chr.14: 35534294-35534294 on Build GRCh38
rs760248717 Chr.14: 35534463-35534463 on Build GRCh38
rs375701927 Chr.14: 35534446-35534446 on Build GRCh38
rs780949616 Chr.14: 35534298-35534298 on Build GRCh38
rs778775186 Chr.14: 35534408-35534408 on Build GRCh38
rs367844606 Chr.14: 35534559-35534559 on Build GRCh38
rs760119142 Chr.14: 35534440-35534440 on Build GRCh38
rs752375416 Chr.14: 35534304-35534304 on Build GRCh38
rs948307541 Chr.14: 35534543-35534543 on Build GRCh38
rs370971573 Chr.14: 35534657-35534657 on Build GRCh38
rs753602904 Chr.14: 35534401-35534401 on Build GRCh38
rs747053722 Chr.14: 35534322-35534322 on Build GRCh38
rs978001818 Chr.14: 35534694-35534694 on Build GRCh38
rs766922721 Chr.14: 35534356-35534356 on Build GRCh38
rs752362643 Chr.14: 35534387-35534387 on Build GRCh38
rs1027168629 Chr.14: 35534502-35534502 on Build GRCh38
rs1004422206 Chr.14: 35534731-35534731 on Build GRCh38
rs200773714 Chr.14: 35534730-35534730 on Build GRCh38
rs755781823 Chr.14: 35534395-35534395 on Build GRCh38
rs751143766 Chr.14: 35534357-35534357 on Build GRCh38
rs1051824963 Chr.14: 35534442-35534442 on Build GRCh38
rs934524854 Chr.14: 35534596-35534596 on Build GRCh38
rs767025381 Chr.14: 35534374-35534374 on Build GRCh38
rs760184149 Chr.14: 35534587-35534587 on Build GRCh38
rs775011630 Chr.14: 35534461-35534461 on Build GRCh38
rs370970531 Chr.14: 35534422-35534422 on Build GRCh38
rs951661328 Chr.14: 35534503-35534503 on Build GRCh38
rs1042911545 Chr.14: 35534399-35534399 on Build GRCh38
rs748188322 Chr.14: 35534499-35534499 on Build GRCh38
rs921384700 Chr.14: 35534348-35534348 on Build GRCh38
rs773611857 Chr.14: 35534352-35534352 on Build GRCh38
rs774893950 Chr.14: 35534536-35534536 on Build GRCh38
rs1019919160 Chr.14: 35534514-35534514 on Build GRCh38
rs770122838 Chr.14: 35534350-35534350 on Build GRCh38
rs749246220 Chr.14: 35534317-35534317 on Build GRCh38
rs144835486 Chr.14: 35534412-35534412 on Build GRCh38
rs749497004 Chr.14: 35534513-35534513 on Build GRCh38
rs754173053 Chr.14: 35534660-35534660 on Build GRCh38
rs930307081 Chr.14: 35534673-35534673 on Build GRCh38
rs562932749 Chr.14: 35534477-35534477 on Build GRCh38
rs775041277 Chr.14: 35534430-35534430 on Build GRCh38
rs766997283 Chr.14: 35534455-35534455 on Build GRCh38
rs756318653 Chr.14: 35534504-35534504 on Build GRCh38
rs764984286 Chr.14: 35534484-35534484 on Build GRCh38
rs763813197 Chr.14: 35534398-35534398 on Build GRCh38
rs779425672 Chr.14: 35534681-35534681 on Build GRCh38
rs372506238 Chr.14: 35534341-35534341 on Build GRCh38
rs967734668 Chr.14: 35534692-35534692 on Build GRCh38

View Full Product Details