Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 35832837-35833336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRMS1L
Entrez Gene ID
84312
Gene Name
breast cancer metastasis-suppressor 1-like
Gene Aliases
BRMS1
UniGene
Hs.525299
Gene Chromosome Location
Chr.14: 35825596-35871963 on Build GRCh38
Transcripts
NM_032352.3 XM_005268128.1 XM_006720274.2 XM_006720275.2 XM_017021703.1 XM_017021704.1 XM_017021705.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4050697 c.247C>T p.R83* Chr.14: 35832991-35832991 on Build GRCh38
955562 c.346C>T p.R116C Chr.14: 35833090-35833090 on Build GRCh38
6580844 c.298C>G p.P100A Chr.14: 35833042-35833042 on Build GRCh38
4666188 c.359delC p.A120fs*9 Chr.14: 35833103-35833103 on Build GRCh38
4050699 c.361+2T>C p.? Chr.14: 35833107-35833107 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548116060 Chr.14: 35832954-35832954 on Build GRCh38
rs934309994 Chr.14: 35833262-35833262 on Build GRCh38
rs951351454 Chr.14: 35833157-35833157 on Build GRCh38
rs750852821 Chr.14: 35833100-35833100 on Build GRCh38
rs970915440 Chr.14: 35833175-35833178 on Build GRCh38
rs960969539 Chr.14: 35833059-35833059 on Build GRCh38
rs769438522 Chr.14: 35833148-35833148 on Build GRCh38
rs373097362 Chr.14: 35833156-35833156 on Build GRCh38
rs369086837 Chr.14: 35833151-35833151 on Build GRCh38
rs943964099 Chr.14: 35833210-35833210 on Build GRCh38
rs185682133 Chr.14: 35832878-35832878 on Build GRCh38
rs150038841 Chr.14: 35833293-35833293 on Build GRCh38
rs1031211224 Chr.14: 35833021-35833021 on Build GRCh38
rs774746348 Chr.14: 35833277-35833277 on Build GRCh38
rs747831272 Chr.14: 35832975-35832975 on Build GRCh38
rs760941857 Chr.14: 35833043-35833043 on Build GRCh38
rs375342798 Chr.14: 35833030-35833030 on Build GRCh38
rs769273904 Chr.14: 35833267-35833267 on Build GRCh38
rs772183126 Chr.14: 35833017-35833017 on Build GRCh38
rs966789523 Chr.14: 35833303-35833303 on Build GRCh38
rs781109822 Chr.14: 35832959-35832959 on Build GRCh38
rs746699961 Chr.14: 35832955-35832956 on Build GRCh38
rs768355530 Chr.14: 35833063-35833063 on Build GRCh38
rs200116011 Chr.14: 35832998-35832998 on Build GRCh38
rs35454164 Chr.14: 35832896-35832897 on Build GRCh38
rs759695899 Chr.14: 35833003-35833003 on Build GRCh38
rs754682205 Chr.14: 35833126-35833126 on Build GRCh38
rs746699662 Chr.14: 35832930-35832930 on Build GRCh38
rs765795127 Chr.14: 35833084-35833084 on Build GRCh38
rs776265731 Chr.14: 35833162-35833162 on Build GRCh38
rs969981102 Chr.14: 35833099-35833099 on Build GRCh38
rs565742753 Chr.14: 35832908-35832908 on Build GRCh38
rs189106569 Chr.14: 35833257-35833257 on Build GRCh38
rs1023777427 Chr.14: 35833091-35833091 on Build GRCh38
rs7141280 Chr.14: 35832891-35832891 on Build GRCh38
rs193039872 Chr.14: 35833291-35833291 on Build GRCh38
rs11625349 Chr.14: 35832898-35832898 on Build GRCh38
rs768400848 Chr.14: 35832940-35832940 on Build GRCh38
rs754231571 Chr.14: 35833066-35833066 on Build GRCh38
rs111940131 Chr.14: 35832987-35832987 on Build GRCh38
rs1005701793 Chr.14: 35833132-35833132 on Build GRCh38
rs989905524 Chr.14: 35833278-35833278 on Build GRCh38
rs150367786 Chr.14: 35833050-35833050 on Build GRCh38
rs868297994 Chr.14: 35833072-35833072 on Build GRCh38
rs536673341 Chr.14: 35833020-35833020 on Build GRCh38
rs539825723 Chr.14: 35832965-35832965 on Build GRCh38
rs368212603 Chr.14: 35833117-35833117 on Build GRCh38
rs1042805147 Chr.14: 35833244-35833244 on Build GRCh38
rs895500922 Chr.14: 35833266-35833266 on Build GRCh38
rs747998404 Chr.14: 35833141-35833141 on Build GRCh38
rs556560583 Chr.14: 35833166-35833166 on Build GRCh38
rs573398779 Chr.14: 35833270-35833270 on Build GRCh38
rs374993101 Chr.14: 35833125-35833125 on Build GRCh38
rs542526249 Chr.14: 35833297-35833297 on Build GRCh38
rs780653502 Chr.14: 35833136-35833136 on Build GRCh38
rs539167529 Chr.14: 35833264-35833264 on Build GRCh38
rs947211045 Chr.14: 35833238-35833238 on Build GRCh38
rs139654728 Chr.14: 35832923-35832923 on Build GRCh38
rs921083000 Chr.14: 35833226-35833226 on Build GRCh38
rs762022323 Chr.14: 35833071-35833071 on Build GRCh38
rs975748695 Chr.14: 35833219-35833219 on Build GRCh38
rs758942324 Chr.14: 35833112-35833112 on Build GRCh38
rs769786220 Chr.14: 35832993-35832993 on Build GRCh38
rs1002499589 Chr.14: 35833206-35833206 on Build GRCh38
rs982818512 Chr.14: 35833164-35833164 on Build GRCh38

View Full Product Details