Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 38255054-38255551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC14A
Entrez Gene ID
161198
Gene Name
C-type lectin domain family 14 member A
Gene Aliases
C14orf27, CEG1, EGFR-5
UniGene
Hs.525307
Gene Chromosome Location
Chr.14: 38254000-38256370 on Build GRCh38
Transcripts
NM_175060.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
289271 c.939C>T p.P313P Chr.14: 38255084-38255084 on Build GRCh38
1369756 c.808G>A p.A270T Chr.14: 38255215-38255215 on Build GRCh38
4641865 c.497C>T p.A166V Chr.14: 38255526-38255526 on Build GRCh38
7307441 c.494G>A p.R165H Chr.14: 38255529-38255529 on Build GRCh38
6630097 c.620C>A p.P207Q Chr.14: 38255403-38255403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938791279 Chr.14: 38255400-38255400 on Build GRCh38
rs747879149 Chr.14: 38255103-38255103 on Build GRCh38
rs997745372 Chr.14: 38255105-38255105 on Build GRCh38
rs913502016 Chr.14: 38255234-38255234 on Build GRCh38
rs139405012 Chr.14: 38255098-38255098 on Build GRCh38
rs763426275 Chr.14: 38255155-38255155 on Build GRCh38
rs200597035 Chr.14: 38255278-38255278 on Build GRCh38
rs778577791 Chr.14: 38255281-38255281 on Build GRCh38
rs749415136 Chr.14: 38255406-38255406 on Build GRCh38
rs758796396 Chr.14: 38255090-38255090 on Build GRCh38
rs779015523 Chr.14: 38255485-38255485 on Build GRCh38
rs753525493 Chr.14: 38255268-38255268 on Build GRCh38
rs375051083 Chr.14: 38255289-38255289 on Build GRCh38
rs779870626 Chr.14: 38255301-38255301 on Build GRCh38
rs145092944 Chr.14: 38255261-38255261 on Build GRCh38
rs751425995 Chr.14: 38255456-38255456 on Build GRCh38
rs749083132 Chr.14: 38255114-38255114 on Build GRCh38
rs769229740 Chr.14: 38255507-38255507 on Build GRCh38
rs144490241 Chr.14: 38255141-38255141 on Build GRCh38
rs368742520 Chr.14: 38255512-38255512 on Build GRCh38
rs374048754 Chr.14: 38255099-38255099 on Build GRCh38
rs938844491 Chr.14: 38255167-38255167 on Build GRCh38
rs770067046 Chr.14: 38255306-38255306 on Build GRCh38
rs1007348317 Chr.14: 38255391-38255391 on Build GRCh38
rs931349842 Chr.14: 38255133-38255133 on Build GRCh38
rs150836716 Chr.14: 38255092-38255092 on Build GRCh38
rs924741086 Chr.14: 38255140-38255140 on Build GRCh38
rs767944196 Chr.14: 38255254-38255254 on Build GRCh38
rs931273230 Chr.14: 38255361-38255361 on Build GRCh38
rs769179875 Chr.14: 38255433-38255433 on Build GRCh38
rs370261872 Chr.14: 38255438-38255438 on Build GRCh38
rs145671249 Chr.14: 38255115-38255115 on Build GRCh38
rs746094763 Chr.14: 38255414-38255414 on Build GRCh38
rs774084335 Chr.14: 38255523-38255523 on Build GRCh38
rs774326804 Chr.14: 38255125-38255125 on Build GRCh38
rs975427789 Chr.14: 38255121-38255121 on Build GRCh38
rs759900108 Chr.14: 38255248-38255248 on Build GRCh38
rs769937145 Chr.14: 38255233-38255233 on Build GRCh38
rs760232515 Chr.14: 38255256-38255256 on Build GRCh38
rs768950746 Chr.14: 38255503-38255503 on Build GRCh38
rs767108935 Chr.14: 38255199-38255199 on Build GRCh38
rs200240958 Chr.14: 38255397-38255397 on Build GRCh38
rs756879176 Chr.14: 38255277-38255277 on Build GRCh38
rs753937174 Chr.14: 38255282-38255282 on Build GRCh38
rs964554256 Chr.14: 38255311-38255311 on Build GRCh38
rs1020321189 Chr.14: 38255440-38255440 on Build GRCh38
rs755412440 Chr.14: 38255198-38255198 on Build GRCh38
rs368955506 Chr.14: 38255237-38255237 on Build GRCh38
rs199846060 Chr.14: 38255132-38255132 on Build GRCh38
rs890198014 Chr.14: 38255516-38255521 on Build GRCh38
rs1006053713 Chr.14: 38255093-38255093 on Build GRCh38
rs777358442 Chr.14: 38255110-38255110 on Build GRCh38
rs780001182 Chr.14: 38255230-38255230 on Build GRCh38
rs763265743 Chr.14: 38255244-38255244 on Build GRCh38
rs997479865 Chr.14: 38255371-38255371 on Build GRCh38
rs755614078 Chr.14: 38255207-38255207 on Build GRCh38
rs750901008 Chr.14: 38255084-38255084 on Build GRCh38
rs114622864 Chr.14: 38255448-38255448 on Build GRCh38
rs766836976 Chr.14: 38255185-38255185 on Build GRCh38
rs779422328 Chr.14: 38255444-38255444 on Build GRCh38
rs376906472 Chr.14: 38255317-38255317 on Build GRCh38
rs771303414 Chr.14: 38255310-38255310 on Build GRCh38
rs754782472 Chr.14: 38255462-38255462 on Build GRCh38
rs937080683 Chr.14: 38255302-38255302 on Build GRCh38
rs759827068 Chr.14: 38255321-38255321 on Build GRCh38
rs765743832 Chr.14: 38255084-38255084 on Build GRCh38
rs149066268 Chr.14: 38255419-38255419 on Build GRCh38
rs200773571 Chr.14: 38255340-38255340 on Build GRCh38
rs1055550848 Chr.14: 38255320-38255320 on Build GRCh38
rs144937961 Chr.14: 38255373-38255373 on Build GRCh38
rs201416778 Chr.14: 38255499-38255499 on Build GRCh38
rs879740161 Chr.14: 38255329-38255329 on Build GRCh38
rs748630764 Chr.14: 38255478-38255478 on Build GRCh38
rs761909687 Chr.14: 38255079-38255079 on Build GRCh38
rs570907333 Chr.14: 38255157-38255157 on Build GRCh38
rs776550947 Chr.14: 38255130-38255130 on Build GRCh38
rs1018256221 Chr.14: 38255173-38255173 on Build GRCh38
rs147835726 Chr.14: 38255416-38255416 on Build GRCh38
rs758143878 Chr.14: 38255288-38255288 on Build GRCh38
rs757342962 Chr.14: 38255407-38255407 on Build GRCh38
rs750035910 Chr.14: 38255374-38255374 on Build GRCh38
rs151154489 Chr.14: 38255501-38255501 on Build GRCh38
rs1017479871 Chr.14: 38255299-38255299 on Build GRCh38
rs746951876 Chr.14: 38255303-38255303 on Build GRCh38
rs558964656 Chr.14: 38255326-38255326 on Build GRCh38
rs745784338 Chr.14: 38255129-38255129 on Build GRCh38
rs766090486 Chr.14: 38255521-38255521 on Build GRCh38
rs755702621 Chr.14: 38255109-38255109 on Build GRCh38
rs761272956 Chr.14: 38255368-38255368 on Build GRCh38
rs367824493 Chr.14: 38255323-38255323 on Build GRCh38
rs748650609 Chr.14: 38255504-38255504 on Build GRCh38
rs747323797 Chr.14: 38255431-38255431 on Build GRCh38
rs908630005 Chr.14: 38255385-38255385 on Build GRCh38
rs772465937 Chr.14: 38255322-38255322 on Build GRCh38
rs758666016 Chr.14: 38255489-38255489 on Build GRCh38
rs1057271433 Chr.14: 38255176-38255176 on Build GRCh38
rs955638844 Chr.14: 38255120-38255120 on Build GRCh38
rs11553024 Chr.14: 38255530-38255530 on Build GRCh38
rs780566045 Chr.14: 38255101-38255101 on Build GRCh38
rs766139028 Chr.14: 38255454-38255454 on Build GRCh38
rs201561377 Chr.14: 38255255-38255255 on Build GRCh38
rs781229998 Chr.14: 38255402-38255402 on Build GRCh38
rs757345305 Chr.14: 38255481-38255481 on Build GRCh38
rs752177134 Chr.14: 38255186-38255186 on Build GRCh38
rs772807752 Chr.14: 38255449-38255449 on Build GRCh38
rs888961431 Chr.14: 38255380-38255380 on Build GRCh38
rs570149280 Chr.14: 38255307-38255307 on Build GRCh38
rs774560433 Chr.14: 38255246-38255246 on Build GRCh38
rs773628972 Chr.14: 38255243-38255243 on Build GRCh38
rs140940981 Chr.14: 38255200-38255200 on Build GRCh38
rs1008624220 Chr.14: 38255527-38255527 on Build GRCh38
rs1017385112 Chr.14: 38255531-38255531 on Build GRCh38
rs957363358 Chr.14: 38255193-38255193 on Build GRCh38
rs754946928 Chr.14: 38255398-38255398 on Build GRCh38
rs753367757 Chr.14: 38255346-38255346 on Build GRCh38
rs767570743 Chr.14: 38255477-38255477 on Build GRCh38
rs868217583 Chr.14: 38255495-38255495 on Build GRCh38
rs185135187 Chr.14: 38255451-38255451 on Build GRCh38
rs779058011 Chr.14: 38255411-38255411 on Build GRCh38
rs746123317 Chr.14: 38255487-38255487 on Build GRCh38
rs758185951 Chr.14: 38255376-38255376 on Build GRCh38
rs371729454 Chr.14: 38255117-38255117 on Build GRCh38
rs775897589 Chr.14: 38255425-38255425 on Build GRCh38
rs757107915 Chr.14: 38255225-38255225 on Build GRCh38
rs765585574 Chr.14: 38255134-38255134 on Build GRCh38
rs776977655 Chr.14: 38255078-38255078 on Build GRCh38
rs887200451 Chr.14: 38255143-38255143 on Build GRCh38
rs779896874 Chr.14: 38255382-38255382 on Build GRCh38
rs762324498 Chr.14: 38255513-38255513 on Build GRCh38
rs777412085 Chr.14: 38255210-38255210 on Build GRCh38
rs750838843 Chr.14: 38255087-38255087 on Build GRCh38
rs745866144 Chr.14: 38255227-38255227 on Build GRCh38
rs987286732 Chr.14: 38255270-38255270 on Build GRCh38
rs373687961 Chr.14: 38255228-38255228 on Build GRCh38
rs767000134 Chr.14: 38255096-38255096 on Build GRCh38

View Full Product Details