Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 39031634-39032133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEC23A
Entrez Gene ID
10484
Gene Name
Sec23 homolog A, coat complex II component
Gene Aliases
CLSD
UniGene
Hs.272927
Gene Chromosome Location
Chr.14: 39031919-39103528 on Build GRCh38
Transcripts
NM_006364.3 XM_005267262.1

SNP Detail

SNP ID Chromosome Location
rs778697491 Chr.14: 39031958-39031958 on Build GRCh38
rs986979777 Chr.14: 39031681-39031681 on Build GRCh38
rs537078590 Chr.14: 39031957-39031957 on Build GRCh38
rs530736959 Chr.14: 39031771-39031771 on Build GRCh38
rs879790541 Chr.14: 39031875-39031875 on Build GRCh38
rs796940940 Chr.14: 39032020-39032020 on Build GRCh38
rs751936323 Chr.14: 39032078-39032078 on Build GRCh38
rs1035734479 Chr.14: 39031817-39031817 on Build GRCh38
rs568098241 Chr.14: 39031853-39031853 on Build GRCh38
rs551932921 Chr.14: 39031822-39031822 on Build GRCh38
rs557399121 Chr.14: 39032012-39032012 on Build GRCh38
rs370269666 Chr.14: 39031769-39031769 on Build GRCh38
rs1002865913 Chr.14: 39031807-39031807 on Build GRCh38
rs563565537 Chr.14: 39031696-39031696 on Build GRCh38
rs532707236 Chr.14: 39031719-39031719 on Build GRCh38
rs968664017 Chr.14: 39032060-39032060 on Build GRCh38
rs1039895892 Chr.14: 39031831-39031831 on Build GRCh38
rs942988503 Chr.14: 39031744-39031744 on Build GRCh38
rs903149240 Chr.14: 39031800-39031800 on Build GRCh38
rs771255307 Chr.14: 39031748-39031748 on Build GRCh38
rs1026029266 Chr.14: 39031909-39031909 on Build GRCh38
rs995457831 Chr.14: 39031896-39031896 on Build GRCh38
rs552507199 Chr.14: 39031743-39031743 on Build GRCh38
rs887170329 Chr.14: 39031951-39031951 on Build GRCh38
rs972184639 Chr.14: 39031898-39031898 on Build GRCh38
rs963261720 Chr.14: 39031871-39031871 on Build GRCh38
rs1025647879 Chr.14: 39031678-39031678 on Build GRCh38
rs958541514 Chr.14: 39032047-39032047 on Build GRCh38
rs1041940332 Chr.14: 39032075-39032075 on Build GRCh38
rs894380495 Chr.14: 39031823-39031823 on Build GRCh38
rs981677965 Chr.14: 39031686-39031686 on Build GRCh38
rs531556694 Chr.14: 39032018-39032018 on Build GRCh38
rs891268980 Chr.14: 39031819-39031819 on Build GRCh38
rs764304291 Chr.14: 39031980-39031980 on Build GRCh38
rs924354704 Chr.14: 39031781-39031781 on Build GRCh38
rs947584905 Chr.14: 39032065-39032065 on Build GRCh38
rs1016484585 Chr.14: 39031869-39031869 on Build GRCh38
rs893317785 Chr.14: 39031764-39031764 on Build GRCh38
rs938774049 Chr.14: 39032079-39032079 on Build GRCh38
rs1025636441 Chr.14: 39031919-39031919 on Build GRCh38
rs954496906 Chr.14: 39031921-39031921 on Build GRCh38
rs1040263238 Chr.14: 39031757-39031757 on Build GRCh38
rs1021545767 Chr.14: 39032057-39032057 on Build GRCh38
rs1012866938 Chr.14: 39031861-39031861 on Build GRCh38
rs186108970 Chr.14: 39031808-39031808 on Build GRCh38
rs991740361 Chr.14: 39032049-39032049 on Build GRCh38
rs942917251 Chr.14: 39032010-39032010 on Build GRCh38

View Full Product Details