Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 39042444-39042964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEC23A
Entrez Gene ID
10484
Gene Name
Sec23 homolog A, coat complex II component
Gene Aliases
CLSD
UniGene
Hs.272927
Gene Chromosome Location
Chr.14: 39031919-39103528 on Build GRCh38
Transcripts
NM_006364.3 XM_005267262.1 XM_011536355.2 XM_017020928.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
955646 c.1937G>A p.R646H Chr.14: 39042835-39042835 on Build GRCh38
6234902 c.1621delC p.A542fs*14 Chr.14: 39042845-39042845 on Build GRCh38
6418675 c.1951G>T p.D651Y Chr.14: 39042821-39042821 on Build GRCh38
4779427 c.1672C>G p.H558D Chr.14: 39042794-39042794 on Build GRCh38
469951 c.1977T>G p.Y659* Chr.14: 39042795-39042795 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932064421 Chr.14: 39042560-39042560 on Build GRCh38
rs747317789 Chr.14: 39042808-39042808 on Build GRCh38
rs565186528 Chr.14: 39042473-39042473 on Build GRCh38
rs763224522 Chr.14: 39042709-39042709 on Build GRCh38
rs1052769954 Chr.14: 39042619-39042619 on Build GRCh38
rs917709158 Chr.14: 39042667-39042667 on Build GRCh38
rs766243948 Chr.14: 39042749-39042749 on Build GRCh38
rs374385312 Chr.14: 39042915-39042915 on Build GRCh38
rs923345461 Chr.14: 39042550-39042550 on Build GRCh38
rs775250176 Chr.14: 39042692-39042692 on Build GRCh38
rs533075090 Chr.14: 39042509-39042509 on Build GRCh38
rs759333766 Chr.14: 39042883-39042883 on Build GRCh38
rs892407769 Chr.14: 39042637-39042637 on Build GRCh38
rs769518920 Chr.14: 39042654-39042654 on Build GRCh38
rs759530040 Chr.14: 39042754-39042754 on Build GRCh38
rs752558821 Chr.14: 39042889-39042889 on Build GRCh38
rs140632104 Chr.14: 39042583-39042586 on Build GRCh38
rs371541535 Chr.14: 39042775-39042775 on Build GRCh38
rs936574246 Chr.14: 39042608-39042608 on Build GRCh38
rs767295894 Chr.14: 39042755-39042755 on Build GRCh38
rs575894530 Chr.14: 39042836-39042836 on Build GRCh38
rs751952409 Chr.14: 39042899-39042899 on Build GRCh38
rs949219477 Chr.14: 39042644-39042644 on Build GRCh38
rs59942239 Chr.14: 39042583-39042583 on Build GRCh38
rs531626411 Chr.14: 39042922-39042922 on Build GRCh38
rs573327355 Chr.14: 39042540-39042540 on Build GRCh38
rs1030509427 Chr.14: 39042871-39042871 on Build GRCh38
rs540704554 Chr.14: 39042886-39042886 on Build GRCh38
rs747715736 Chr.14: 39042853-39042853 on Build GRCh38
rs765230150 Chr.14: 39042893-39042893 on Build GRCh38
rs761996875 Chr.14: 39042891-39042891 on Build GRCh38
rs925295547 Chr.14: 39042606-39042606 on Build GRCh38
rs991873311 Chr.14: 39042652-39042652 on Build GRCh38
rs141375970 Chr.14: 39042910-39042910 on Build GRCh38
rs549164110 Chr.14: 39042908-39042908 on Build GRCh38
rs112923906 Chr.14: 39042623-39042623 on Build GRCh38
rs542424742 Chr.14: 39042563-39042563 on Build GRCh38
rs1048025083 Chr.14: 39042706-39042706 on Build GRCh38
rs769211083 Chr.14: 39042824-39042824 on Build GRCh38
rs3837584 Chr.14: 39042749-39042749 on Build GRCh38
rs988198524 Chr.14: 39042901-39042901 on Build GRCh38
rs189849576 Chr.14: 39042882-39042882 on Build GRCh38
rs758830794 Chr.14: 39042776-39042776 on Build GRCh38
rs4902326 Chr.14: 39042918-39042918 on Build GRCh38
rs961706449 Chr.14: 39042616-39042616 on Build GRCh38
rs553478682 Chr.14: 39042522-39042522 on Build GRCh38
rs562200303 Chr.14: 39042751-39042751 on Build GRCh38
rs1046249923 Chr.14: 39042651-39042651 on Build GRCh38
rs891811341 Chr.14: 39042465-39042465 on Build GRCh38
rs753953017 Chr.14: 39042760-39042760 on Build GRCh38
rs201214772 Chr.14: 39042870-39042870 on Build GRCh38
rs762889177 Chr.14: 39042746-39042746 on Build GRCh38
rs757240112 Chr.14: 39042771-39042771 on Build GRCh38
rs201330860 Chr.14: 39042878-39042878 on Build GRCh38
rs1001912529 Chr.14: 39042701-39042701 on Build GRCh38
rs141058371 Chr.14: 39042895-39042895 on Build GRCh38
rs950298260 Chr.14: 39042495-39042511 on Build GRCh38
rs779247642 Chr.14: 39042774-39042774 on Build GRCh38
rs144656181 Chr.14: 39042866-39042866 on Build GRCh38
rs544966037 Chr.14: 39042877-39042877 on Build GRCh38
rs1010225972 Chr.14: 39042835-39042835 on Build GRCh38
rs745669345 Chr.14: 39042562-39042562 on Build GRCh38
rs200822209 Chr.14: 39042894-39042894 on Build GRCh38
rs962507933 Chr.14: 39042508-39042508 on Build GRCh38
rs780386756 Chr.14: 39042778-39042778 on Build GRCh38
rs1003909140 Chr.14: 39042714-39042714 on Build GRCh38

View Full Product Details