Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 39045185-39045733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEC23A
Entrez Gene ID
10484
Gene Name
Sec23 homolog A, coat complex II component
Gene Aliases
CLSD
UniGene
Hs.272927
Gene Chromosome Location
Chr.14: 39031919-39103528 on Build GRCh38
Transcripts
NM_006364.3 XM_005267262.1 XM_011536355.2 XM_017020928.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7275718 c.1438T>C p.F480L Chr.14: 39045318-39045318 on Build GRCh38
4050778 c.1471T>C p.F491L Chr.14: 39045285-39045285 on Build GRCh38
4844048 c.1828C>G p.Q610E Chr.14: 39045234-39045234 on Build GRCh38
4472886 c.1504C>T p.R502C Chr.14: 39045252-39045252 on Build GRCh38
7275717 c.1744T>C p.F582L Chr.14: 39045318-39045318 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991493291 Chr.14: 39045585-39045585 on Build GRCh38
rs397689791 Chr.14: 39045530-39045531 on Build GRCh38
rs1003531177 Chr.14: 39045589-39045589 on Build GRCh38
rs540878796 Chr.14: 39045422-39045422 on Build GRCh38
rs910962227 Chr.14: 39045479-39045479 on Build GRCh38
rs1055506226 Chr.14: 39045613-39045613 on Build GRCh38
rs2415524 Chr.14: 39045559-39045559 on Build GRCh38
rs768611692 Chr.14: 39045330-39045330 on Build GRCh38
rs918667037 Chr.14: 39045599-39045599 on Build GRCh38
rs374718440 Chr.14: 39045346-39045346 on Build GRCh38
rs1038127463 Chr.14: 39045541-39045541 on Build GRCh38
rs763069578 Chr.14: 39045340-39045340 on Build GRCh38
rs771963211 Chr.14: 39045257-39045257 on Build GRCh38
rs1022062406 Chr.14: 39045267-39045268 on Build GRCh38
rs986757559 Chr.14: 39045614-39045614 on Build GRCh38
rs745505951 Chr.14: 39045247-39045247 on Build GRCh38
rs914535517 Chr.14: 39045692-39045692 on Build GRCh38
rs752165778 Chr.14: 39045359-39045359 on Build GRCh38
rs966509411 Chr.14: 39045670-39045671 on Build GRCh38
rs571879548 Chr.14: 39045376-39045376 on Build GRCh38
rs951450232 Chr.14: 39045611-39045611 on Build GRCh38
rs955067759 Chr.14: 39045461-39045461 on Build GRCh38
rs192692429 Chr.14: 39045477-39045477 on Build GRCh38
rs558653332 Chr.14: 39045319-39045319 on Build GRCh38
rs752079558 Chr.14: 39045561-39045561 on Build GRCh38
rs57657891 Chr.14: 39045522-39045523 on Build GRCh38
rs974307052 Chr.14: 39045596-39045596 on Build GRCh38
rs918701287 Chr.14: 39045321-39045321 on Build GRCh38
rs1040012229 Chr.14: 39045685-39045685 on Build GRCh38
rs776504661 Chr.14: 39045339-39045339 on Build GRCh38
rs942602572 Chr.14: 39045655-39045655 on Build GRCh38
rs941426640 Chr.14: 39045249-39045249 on Build GRCh38
rs60316211 Chr.14: 39045481-39045481 on Build GRCh38
rs1011433756 Chr.14: 39045506-39045506 on Build GRCh38
rs748835653 Chr.14: 39045236-39045236 on Build GRCh38
rs1051129267 Chr.14: 39045362-39045362 on Build GRCh38
rs1010907976 Chr.14: 39045623-39045623 on Build GRCh38
rs774620917 Chr.14: 39045368-39045368 on Build GRCh38
rs569140973 Chr.14: 39045621-39045621 on Build GRCh38
rs547674515 Chr.14: 39045649-39045649 on Build GRCh38
rs559503794 Chr.14: 39045595-39045595 on Build GRCh38
rs1023919724 Chr.14: 39045586-39045586 on Build GRCh38
rs902976790 Chr.14: 39045550-39045550 on Build GRCh38
rs1020452634 Chr.14: 39045521-39045521 on Build GRCh38
rs80163026 Chr.14: 39045554-39045554 on Build GRCh38
rs775163398 Chr.14: 39045278-39045278 on Build GRCh38
rs751117084 Chr.14: 39045209-39045209 on Build GRCh38
rs574641594 Chr.14: 39045440-39045440 on Build GRCh38
rs554803680 Chr.14: 39045425-39045425 on Build GRCh38
rs1010960359 Chr.14: 39045237-39045237 on Build GRCh38

View Full Product Details