Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 44924772-44925281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL28
Entrez Gene ID
54813
Gene Name
kelch like family member 28
Gene Aliases
BTBD5
UniGene
Hs.550906
Gene Chromosome Location
Chr.14: 44924310-44961978 on Build GRCh38
Transcripts
NM_001308112.1 NM_017658.4 XM_005267770.3 XM_011536847.2 XM_011536849.2

SNP Detail

SNP ID Chromosome Location
rs1027824490 Chr.14: 44924953-44924953 on Build GRCh38
rs969476753 Chr.14: 44925132-44925132 on Build GRCh38
rs57261684 Chr.14: 44924867-44924867 on Build GRCh38
rs530196508 Chr.14: 44924854-44924854 on Build GRCh38
rs1055659162 Chr.14: 44925088-44925088 on Build GRCh38
rs776505492 Chr.14: 44924851-44924851 on Build GRCh38
rs142921397 Chr.14: 44925181-44925181 on Build GRCh38
rs978831639 Chr.14: 44925046-44925046 on Build GRCh38
rs946041818 Chr.14: 44925036-44925036 on Build GRCh38
rs946419585 Chr.14: 44924888-44924888 on Build GRCh38
rs913841992 Chr.14: 44925213-44925213 on Build GRCh38
rs1052324218 Chr.14: 44924862-44924862 on Build GRCh38
rs913274407 Chr.14: 44925035-44925035 on Build GRCh38
rs759689847 Chr.14: 44924855-44924855 on Build GRCh38
rs976712775 Chr.14: 44925009-44925009 on Build GRCh38
rs1018464129 Chr.14: 44924989-44924989 on Build GRCh38
rs1027635276 Chr.14: 44925121-44925121 on Build GRCh38
rs142959695 Chr.14: 44924947-44924947 on Build GRCh38
rs185361638 Chr.14: 44925075-44925075 on Build GRCh38
rs994924120 Chr.14: 44925159-44925159 on Build GRCh38
rs181925280 Chr.14: 44924921-44924921 on Build GRCh38
rs151117139 Chr.14: 44924968-44924968 on Build GRCh38
rs561762867 Chr.14: 44924832-44924832 on Build GRCh38
rs1036285148 Chr.14: 44925148-44925148 on Build GRCh38
rs1009475648 Chr.14: 44924970-44924970 on Build GRCh38
rs763378716 Chr.14: 44925194-44925194 on Build GRCh38
rs560536128 Chr.14: 44924900-44924900 on Build GRCh38
rs553543314 Chr.14: 44925090-44925090 on Build GRCh38
rs965627736 Chr.14: 44925002-44925002 on Build GRCh38
rs868597856 Chr.14: 44925161-44925161 on Build GRCh38
rs949989272 Chr.14: 44925128-44925128 on Build GRCh38
rs73334108 Chr.14: 44924949-44924949 on Build GRCh38
rs937164439 Chr.14: 44925072-44925072 on Build GRCh38
rs888886869 Chr.14: 44925203-44925203 on Build GRCh38
rs1014633902 Chr.14: 44924945-44924945 on Build GRCh38
rs927792690 Chr.14: 44924825-44924825 on Build GRCh38
rs750300347 Chr.14: 44924845-44924845 on Build GRCh38
rs952882785 Chr.14: 44924962-44924962 on Build GRCh38
rs568603519 Chr.14: 44925051-44925051 on Build GRCh38
rs532063484 Chr.14: 44924959-44924959 on Build GRCh38
rs980620731 Chr.14: 44924804-44924804 on Build GRCh38
rs897463497 Chr.14: 44925155-44925155 on Build GRCh38
rs926041801 Chr.14: 44925062-44925062 on Build GRCh38

View Full Product Details