Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 49601421-49601820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRR1
Entrez Gene ID
122769
Gene Name
leucine rich repeat protein 1
Gene Aliases
4-1BBLRR, LRR-1, PPIL5
UniGene
Hs.451090
Gene Chromosome Location
Chr.14: 49598697-49614672 on Build GRCh38
Transcripts
NM_152329.3 NM_203467.1 NR_037792.1 NR_037793.1

SNP Detail

SNP ID Chromosome Location
rs1046031594 Chr.14: 49601546-49601546 on Build GRCh38
rs940853546 Chr.14: 49601765-49601765 on Build GRCh38
rs562713831 Chr.14: 49601757-49601757 on Build GRCh38
rs1050807138 Chr.14: 49601654-49601654 on Build GRCh38
rs778255970 Chr.14: 49601724-49601724 on Build GRCh38
rs566615156 Chr.14: 49601487-49601487 on Build GRCh38
rs1022150995 Chr.14: 49601620-49601620 on Build GRCh38
rs775703750 Chr.14: 49601483-49601483 on Build GRCh38
rs753938516 Chr.14: 49601625-49601625 on Build GRCh38
rs1000613711 Chr.14: 49601449-49601449 on Build GRCh38
rs1038638531 Chr.14: 49601461-49601461 on Build GRCh38
rs1005161407 Chr.14: 49601534-49601534 on Build GRCh38
rs770351472 Chr.14: 49601640-49601640 on Build GRCh38
rs965896661 Chr.14: 49601673-49601673 on Build GRCh38
rs555279159 Chr.14: 49601624-49601624 on Build GRCh38
rs977864652 Chr.14: 49601635-49601635 on Build GRCh38
rs931915998 Chr.14: 49601638-49601638 on Build GRCh38
rs1045899401 Chr.14: 49601791-49601791 on Build GRCh38
rs60910708 Chr.14: 49601464-49601464 on Build GRCh38
rs1049266874 Chr.14: 49601516-49601516 on Build GRCh38
rs982846141 Chr.14: 49601574-49601574 on Build GRCh38
rs745667427 Chr.14: 49601477-49601477 on Build GRCh38
rs920096712 Chr.14: 49601535-49601535 on Build GRCh38
rs907426497 Chr.14: 49601561-49601561 on Build GRCh38
rs770894959 Chr.14: 49601584-49601584 on Build GRCh38
rs574670768 Chr.14: 49601629-49601629 on Build GRCh38
rs760002479 Chr.14: 49601621-49601621 on Build GRCh38
rs182651729 Chr.14: 49601583-49601583 on Build GRCh38
rs974420994 Chr.14: 49601718-49601718 on Build GRCh38
rs930388137 Chr.14: 49601514-49601514 on Build GRCh38
rs534171072 Chr.14: 49601733-49601733 on Build GRCh38
rs546283705 Chr.14: 49601741-49601741 on Build GRCh38
rs114183039 Chr.14: 49601738-49601738 on Build GRCh38
rs113860742 Chr.14: 49601585-49601585 on Build GRCh38
rs1024601656 Chr.14: 49601500-49601500 on Build GRCh38
rs886595561 Chr.14: 49601527-49601527 on Build GRCh38
rs769228564 Chr.14: 49601524-49601524 on Build GRCh38
rs981467640 Chr.14: 49601519-49601519 on Build GRCh38
rs772878178 Chr.14: 49601749-49601749 on Build GRCh38
rs1013184280 Chr.14: 49601488-49601488 on Build GRCh38
rs953020680 Chr.14: 49601553-49601553 on Build GRCh38
rs375141598 Chr.14: 49601604-49601604 on Build GRCh38
rs921706535 Chr.14: 49601468-49601468 on Build GRCh38
rs545998023 Chr.14: 49601736-49601736 on Build GRCh38
rs774836848 Chr.14: 49601502-49601502 on Build GRCh38
rs11625804 Chr.14: 49601581-49601581 on Build GRCh38
rs944388052 Chr.14: 49601460-49601460 on Build GRCh38

View Full Product Details