Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 50996274-50996812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM9
Entrez Gene ID
114088
Gene Name
tripartite motif containing 9
Gene Aliases
RNF91, SPRING
UniGene
Hs.654750
Gene Chromosome Location
Chr.14: 50975261-51105173 on Build GRCh38
Transcripts
NM_015163.5 NM_052978.4 XM_005267301.3 XM_005267302.3 XM_006720018.3 XM_011536389.2 XM_017020943.1 XM_017020944.1 XM_017020945.1 XM_017020946.1 XM_017020947.1 XM_017020948.1 XM_017020949.1 XM_017020950.1

SNP Detail

SNP ID Chromosome Location
rs564522162 Chr.14: 50996508-50996508 on Build GRCh38
rs184021710 Chr.14: 50996362-50996362 on Build GRCh38
rs147925399 Chr.14: 50996299-50996299 on Build GRCh38
rs72687113 Chr.14: 50996640-50996640 on Build GRCh38
rs1055824558 Chr.14: 50996451-50996451 on Build GRCh38
rs1028927506 Chr.14: 50996526-50996526 on Build GRCh38
rs1049494982 Chr.14: 50996478-50996478 on Build GRCh38
rs17796649 Chr.14: 50996656-50996656 on Build GRCh38
rs751050160 Chr.14: 50996630-50996630 on Build GRCh38
rs991850128 Chr.14: 50996636-50996636 on Build GRCh38
rs928728961 Chr.14: 50996412-50996412 on Build GRCh38
rs928602987 Chr.14: 50996746-50996746 on Build GRCh38
rs930681714 Chr.14: 50996450-50996450 on Build GRCh38
rs908122961 Chr.14: 50996714-50996714 on Build GRCh38
rs11852198 Chr.14: 50996507-50996507 on Build GRCh38
rs1004963414 Chr.14: 50996485-50996485 on Build GRCh38
rs745923898 Chr.14: 50996388-50996388 on Build GRCh38
rs937445958 Chr.14: 50996413-50996413 on Build GRCh38
rs979127725 Chr.14: 50996599-50996599 on Build GRCh38
rs190530928 Chr.14: 50996489-50996489 on Build GRCh38
rs956491697 Chr.14: 50996605-50996605 on Build GRCh38
rs562617746 Chr.14: 50996641-50996641 on Build GRCh38
rs886531388 Chr.14: 50996480-50996480 on Build GRCh38
rs1049095496 Chr.14: 50996742-50996742 on Build GRCh38
rs1026750610 Chr.14: 50996668-50996668 on Build GRCh38
rs555220120 Chr.14: 50996464-50996464 on Build GRCh38
rs766031274 Chr.14: 50996703-50996703 on Build GRCh38
rs201591265 Chr.14: 50996360-50996361 on Build GRCh38
rs189813804 Chr.14: 50996364-50996364 on Build GRCh38
rs141871743 Chr.14: 50996632-50996632 on Build GRCh38
rs185206391 Chr.14: 50996486-50996486 on Build GRCh38
rs772323226 Chr.14: 50996389-50996389 on Build GRCh38
rs906081564 Chr.14: 50996448-50996448 on Build GRCh38
rs937371928 Chr.14: 50996753-50996753 on Build GRCh38
rs547946567 Chr.14: 50996787-50996787 on Build GRCh38
rs962642470 Chr.14: 50996741-50996741 on Build GRCh38
rs1044640565 Chr.14: 50996420-50996420 on Build GRCh38
rs751252827 Chr.14: 50996780-50996780 on Build GRCh38
rs771182118 Chr.14: 50996308-50996308 on Build GRCh38
rs71422010 Chr.14: 50996387-50996387 on Build GRCh38
rs981453607 Chr.14: 50996745-50996745 on Build GRCh38
rs749706159 Chr.14: 50996707-50996707 on Build GRCh38
rs532278534 Chr.14: 50996578-50996578 on Build GRCh38
rs935046682 Chr.14: 50996371-50996371 on Build GRCh38
rs1055772057 Chr.14: 50996755-50996755 on Build GRCh38
rs1041576066 Chr.14: 50996544-50996544 on Build GRCh38
rs952159046 Chr.14: 50996675-50996675 on Build GRCh38
rs967752540 Chr.14: 50996344-50996344 on Build GRCh38
rs775730775 Chr.14: 50996402-50996402 on Build GRCh38
rs10483607 Chr.14: 50996604-50996604 on Build GRCh38

View Full Product Details