Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 52859313-52859813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FERMT2
Entrez Gene ID
10979
Gene Name
fermitin family member 2
Gene Aliases
KIND2, MIG2, PLEKHC1, UNC112, UNC112B, mig-2
UniGene
Hs.509343
Gene Chromosome Location
Chr.14: 52857271-52951192 on Build GRCh38
Transcripts
NM_001134999.1 NM_001135000.1 NM_006832.2 XM_005267285.2 XM_006720008.2 XM_006720009.2 XM_006720010.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7140240 c.1851C>A p.V617V Chr.14: 52859591-52859591 on Build GRCh38
1370119 c.1863C>A p.I621I Chr.14: 52859579-52859579 on Build GRCh38
956260 c.1742A>C p.K581T Chr.14: 52859700-52859700 on Build GRCh38
4684348 c.1746delA p.E583fs*11 Chr.14: 52859696-52859696 on Build GRCh38
4606419 c.1823G>A p.R608H Chr.14: 52859619-52859619 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017341030 Chr.14: 52859500-52859500 on Build GRCh38
rs763266914 Chr.14: 52859690-52859692 on Build GRCh38
rs765531426 Chr.14: 52859634-52859634 on Build GRCh38
rs3818453 Chr.14: 52859736-52859736 on Build GRCh38
rs753136089 Chr.14: 52859703-52859703 on Build GRCh38
rs372955263 Chr.14: 52859606-52859606 on Build GRCh38
rs773647443 Chr.14: 52859763-52859763 on Build GRCh38
rs773603392 Chr.14: 52859559-52859563 on Build GRCh38
rs774332955 Chr.14: 52859610-52859610 on Build GRCh38
rs145441604 Chr.14: 52859533-52859533 on Build GRCh38
rs756402897 Chr.14: 52859711-52859711 on Build GRCh38
rs745803960 Chr.14: 52859619-52859619 on Build GRCh38
rs756829780 Chr.14: 52859722-52859722 on Build GRCh38
rs763151830 Chr.14: 52859731-52859731 on Build GRCh38
rs907310260 Chr.14: 52859650-52859650 on Build GRCh38
rs766580924 Chr.14: 52859717-52859717 on Build GRCh38
rs370413868 Chr.14: 52859717-52859717 on Build GRCh38
rs967691571 Chr.14: 52859478-52859478 on Build GRCh38
rs753674418 Chr.14: 52859753-52859753 on Build GRCh38
rs767586396 Chr.14: 52859748-52859749 on Build GRCh38
rs142651496 Chr.14: 52859633-52859633 on Build GRCh38
rs771841843 Chr.14: 52859624-52859624 on Build GRCh38
rs910580556 Chr.14: 52859537-52859537 on Build GRCh38
rs920679895 Chr.14: 52859438-52859438 on Build GRCh38
rs375267385 Chr.14: 52859539-52859539 on Build GRCh38
rs149668518 Chr.14: 52859341-52859341 on Build GRCh38
rs768210015 Chr.14: 52859762-52859762 on Build GRCh38
rs755812689 Chr.14: 52859545-52859545 on Build GRCh38
rs985725188 Chr.14: 52859488-52859488 on Build GRCh38
rs1024962770 Chr.14: 52859377-52859377 on Build GRCh38
rs752407695 Chr.14: 52859785-52859785 on Build GRCh38
rs773542950 Chr.14: 52859636-52859636 on Build GRCh38
rs1009911227 Chr.14: 52859402-52859402 on Build GRCh38
rs543698477 Chr.14: 52859448-52859448 on Build GRCh38
rs945906576 Chr.14: 52859640-52859640 on Build GRCh38
rs377196292 Chr.14: 52859658-52859658 on Build GRCh38
rs773491542 Chr.14: 52859714-52859714 on Build GRCh38
rs948735041 Chr.14: 52859570-52859570 on Build GRCh38
rs199835806 Chr.14: 52859526-52859526 on Build GRCh38
rs746628542 Chr.14: 52859752-52859752 on Build GRCh38
rs770406738 Chr.14: 52859594-52859594 on Build GRCh38
rs1022177080 Chr.14: 52859754-52859754 on Build GRCh38
rs751620780 Chr.14: 52859659-52859659 on Build GRCh38
rs375492748 Chr.14: 52859737-52859737 on Build GRCh38
rs772073376 Chr.14: 52859739-52859739 on Build GRCh38
rs144927888 Chr.14: 52859685-52859685 on Build GRCh38
rs202196410 Chr.14: 52859542-52859542 on Build GRCh38
rs763180941 Chr.14: 52859643-52859643 on Build GRCh38
rs966184124 Chr.14: 52859523-52859523 on Build GRCh38
rs892412539 Chr.14: 52859392-52859392 on Build GRCh38
rs577650375 Chr.14: 52859502-52859502 on Build GRCh38
rs771274367 Chr.14: 52859464-52859465 on Build GRCh38
rs137900215 Chr.14: 52859632-52859632 on Build GRCh38
rs114425935 Chr.14: 52859467-52859467 on Build GRCh38
rs140299918 Chr.14: 52859560-52859560 on Build GRCh38
rs755459645 Chr.14: 52859536-52859536 on Build GRCh38
rs958611324 Chr.14: 52859354-52859354 on Build GRCh38
rs1014530328 Chr.14: 52859372-52859372 on Build GRCh38
rs754990637 Chr.14: 52859744-52859746 on Build GRCh38
rs775255559 Chr.14: 52859751-52859751 on Build GRCh38
rs972618693 Chr.14: 52859412-52859412 on Build GRCh38
rs751835898 Chr.14: 52859719-52859721 on Build GRCh38
rs909422369 Chr.14: 52859567-52859567 on Build GRCh38
rs879447983 Chr.14: 52859347-52859347 on Build GRCh38
rs757237572 Chr.14: 52859765-52859766 on Build GRCh38
rs537045234 Chr.14: 52859564-52859564 on Build GRCh38
rs962248019 Chr.14: 52859379-52859379 on Build GRCh38
rs373085863 Chr.14: 52859718-52859718 on Build GRCh38
rs780753129 Chr.14: 52859525-52859525 on Build GRCh38
rs767862305 Chr.14: 52859691-52859691 on Build GRCh38
rs765480357 Chr.14: 52859535-52859537 on Build GRCh38

View Full Product Details