Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 54957166-54957664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDHD1
Entrez Gene ID
11169
Gene Name
WD repeat and HMG-box DNA binding protein 1
Gene Aliases
AND-1, AND1, CHTF4, CTF4
UniGene
Hs.385998
Gene Chromosome Location
Chr.14: 54938938-55027105 on Build GRCh38
Transcripts
NM_001008396.2 NM_007086.3 XM_006720012.2 XM_011536373.2 XM_017020940.1 XM_017020941.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4740828 c.2729G>A p.R910Q Chr.14: 54957608-54957608 on Build GRCh38
4883012 c.2752G>T p.A918S Chr.14: 54957198-54957198 on Build GRCh38
6409640 c.2705C>A p.A902E Chr.14: 54957632-54957632 on Build GRCh38
2245190 c.2728C>T p.R910* Chr.14: 54957609-54957609 on Build GRCh38
470032 c.2746G>A p.V916I Chr.14: 54957204-54957204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs894244028 Chr.14: 54957357-54957357 on Build GRCh38
rs971046744 Chr.14: 54957384-54957384 on Build GRCh38
rs745848052 Chr.14: 54957595-54957601 on Build GRCh38
rs560297838 Chr.14: 54957533-54957533 on Build GRCh38
rs940282008 Chr.14: 54957266-54957266 on Build GRCh38
rs759271733 Chr.14: 54957246-54957246 on Build GRCh38
rs750111288 Chr.14: 54957573-54957573 on Build GRCh38
rs918588482 Chr.14: 54957477-54957477 on Build GRCh38
rs755324832 Chr.14: 54957580-54957580 on Build GRCh38
rs753481218 Chr.14: 54957191-54957191 on Build GRCh38
rs764779956 Chr.14: 54957568-54957568 on Build GRCh38
rs117649156 Chr.14: 54957295-54957295 on Build GRCh38
rs999285668 Chr.14: 54957300-54957300 on Build GRCh38
rs778663933 Chr.14: 54957198-54957198 on Build GRCh38
rs756551964 Chr.14: 54957608-54957608 on Build GRCh38
rs930333500 Chr.14: 54957310-54957310 on Build GRCh38
rs774692841 Chr.14: 54957239-54957239 on Build GRCh38
rs756029375 Chr.14: 54957358-54957358 on Build GRCh38
rs574079419 Chr.14: 54957395-54957395 on Build GRCh38
rs753163628 Chr.14: 54957605-54957605 on Build GRCh38
rs137977906 Chr.14: 54957631-54957631 on Build GRCh38
rs544617357 Chr.14: 54957409-54957409 on Build GRCh38
rs540712687 Chr.14: 54957328-54957328 on Build GRCh38
rs371209985 Chr.14: 54957612-54957612 on Build GRCh38
rs756979571 Chr.14: 54957481-54957481 on Build GRCh38
rs912791697 Chr.14: 54957240-54957240 on Build GRCh38
rs912733252 Chr.14: 54957591-54957591 on Build GRCh38
rs749002719 Chr.14: 54957379-54957379 on Build GRCh38
rs367731286 Chr.14: 54957196-54957196 on Build GRCh38
rs545599988 Chr.14: 54957476-54957476 on Build GRCh38
rs573449017 Chr.14: 54957509-54957509 on Build GRCh38
rs567761842 Chr.14: 54957567-54957567 on Build GRCh38
rs778648933 Chr.14: 54957589-54957590 on Build GRCh38
rs751963710 Chr.14: 54957202-54957202 on Build GRCh38
rs760685400 Chr.14: 54957291-54957291 on Build GRCh38
rs1039967775 Chr.14: 54957233-54957233 on Build GRCh38
rs1023574396 Chr.14: 54957380-54957380 on Build GRCh38
rs746848106 Chr.14: 54957620-54957629 on Build GRCh38
rs771057475 Chr.14: 54957625-54957625 on Build GRCh38
rs201296875 Chr.14: 54957553-54957553 on Build GRCh38
rs777697391 Chr.14: 54957226-54957226 on Build GRCh38
rs376786739 Chr.14: 54957232-54957232 on Build GRCh38
rs1045992546 Chr.14: 54957367-54957367 on Build GRCh38
rs775365350 Chr.14: 54957552-54957552 on Build GRCh38
rs371258231 Chr.14: 54957205-54957205 on Build GRCh38
rs755449494 Chr.14: 54957204-54957204 on Build GRCh38
rs75164364 Chr.14: 54957445-54957445 on Build GRCh38
rs756228188 Chr.14: 54957245-54957245 on Build GRCh38
rs149926168 Chr.14: 54957301-54957301 on Build GRCh38
rs769947426 Chr.14: 54957220-54957220 on Build GRCh38
rs748698948 Chr.14: 54957219-54957219 on Build GRCh38
rs189692878 Chr.14: 54957420-54957420 on Build GRCh38
rs997216225 Chr.14: 54957259-54957259 on Build GRCh38
rs764142038 Chr.14: 54957565-54957565 on Build GRCh38
rs771713401 Chr.14: 54957601-54957602 on Build GRCh38
rs145015087 Chr.14: 54957539-54957539 on Build GRCh38
rs367634159 Chr.14: 54957548-54957548 on Build GRCh38
rs778414691 Chr.14: 54957609-54957609 on Build GRCh38
rs778456966 Chr.14: 54957507-54957507 on Build GRCh38
rs775240435 Chr.14: 54957603-54957604 on Build GRCh38
rs117103315 Chr.14: 54957378-54957378 on Build GRCh38
rs538516335 Chr.14: 54957598-54957598 on Build GRCh38
rs957360589 Chr.14: 54957532-54957532 on Build GRCh38

View Full Product Details