Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 55353406-55353935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO34
Entrez Gene ID
55030
Gene Name
F-box protein 34
Gene Aliases
CGI-301, Fbx34
UniGene
Hs.525348
Gene Chromosome Location
Chr.14: 55271303-55355222 on Build GRCh38
Transcripts
NM_017943.3 XM_005267791.4 XM_006720185.3 XM_017021391.1 XM_017021392.1 XM_017021393.1

SNP Detail

SNP ID Chromosome Location
rs778699977 Chr.14: 55353674-55353674 on Build GRCh38
rs749842266 Chr.14: 55353685-55353685 on Build GRCh38
rs1056774235 Chr.14: 55353617-55353618 on Build GRCh38
rs144060951 Chr.14: 55353808-55353808 on Build GRCh38
rs923939204 Chr.14: 55353549-55353549 on Build GRCh38
rs905660996 Chr.14: 55353598-55353598 on Build GRCh38
rs775178814 Chr.14: 55353905-55353905 on Build GRCh38
rs566849675 Chr.14: 55353851-55353851 on Build GRCh38
rs955444894 Chr.14: 55353738-55353738 on Build GRCh38
rs1045229551 Chr.14: 55353594-55353594 on Build GRCh38
rs576078082 Chr.14: 55353651-55353651 on Build GRCh38
rs770342528 Chr.14: 55353627-55353627 on Build GRCh38
rs773588673 Chr.14: 55353675-55353675 on Build GRCh38
rs757948485 Chr.14: 55353681-55353681 on Build GRCh38
rs1016618703 Chr.14: 55353638-55353638 on Build GRCh38
rs1009840410 Chr.14: 55353624-55353624 on Build GRCh38
rs540448117 Chr.14: 55353703-55353703 on Build GRCh38
rs963799622 Chr.14: 55353693-55353693 on Build GRCh38
rs938104679 Chr.14: 55353835-55353835 on Build GRCh38
rs7153110 Chr.14: 55353910-55353910 on Build GRCh38
rs1028821198 Chr.14: 55353680-55353680 on Build GRCh38
rs533135109 Chr.14: 55353758-55353758 on Build GRCh38
rs770296943 Chr.14: 55353797-55353797 on Build GRCh38
rs748539226 Chr.14: 55353509-55353509 on Build GRCh38
rs941411577 Chr.14: 55353501-55353501 on Build GRCh38
rs989485992 Chr.14: 55353734-55353734 on Build GRCh38
rs149531026 Chr.14: 55353721-55353721 on Build GRCh38
rs981392411 Chr.14: 55353760-55353760 on Build GRCh38
rs144497401 Chr.14: 55353496-55353496 on Build GRCh38
rs767167660 Chr.14: 55353856-55353856 on Build GRCh38
rs1056827945 Chr.14: 55353793-55353793 on Build GRCh38
rs1000314024 Chr.14: 55353659-55353659 on Build GRCh38
rs941787998 Chr.14: 55353794-55353794 on Build GRCh38
rs1056455834 Chr.14: 55353842-55353842 on Build GRCh38
rs1054466581 Chr.14: 55353558-55353558 on Build GRCh38
rs937991530 Chr.14: 55353555-55353555 on Build GRCh38
rs962576250 Chr.14: 55353788-55353788 on Build GRCh38
rs1016153733 Chr.14: 55353478-55353478 on Build GRCh38
rs370383928 Chr.14: 55353634-55353635 on Build GRCh38
rs939494149 Chr.14: 55353783-55353783 on Build GRCh38
rs551794889 Chr.14: 55353824-55353824 on Build GRCh38
rs1031061520 Chr.14: 55353711-55353711 on Build GRCh38
rs1056511008 Chr.14: 55353577-55353577 on Build GRCh38
rs528236471 Chr.14: 55353572-55353572 on Build GRCh38
rs575285790 Chr.14: 55353429-55353429 on Build GRCh38
rs567637075 Chr.14: 55353798-55353798 on Build GRCh38
rs376223034 Chr.14: 55353859-55353859 on Build GRCh38
rs80046528 Chr.14: 55353749-55353749 on Build GRCh38
rs781637669 Chr.14: 55353460-55353460 on Build GRCh38
rs956717673 Chr.14: 55353723-55353724 on Build GRCh38
rs928145145 Chr.14: 55353766-55353766 on Build GRCh38
rs954767730 Chr.14: 55353531-55353531 on Build GRCh38
rs373957058 Chr.14: 55353730-55353731 on Build GRCh38
rs1042922405 Chr.14: 55353644-55353644 on Build GRCh38
rs117256830 Chr.14: 55353469-55353469 on Build GRCh38
rs1004527949 Chr.14: 55353628-55353629 on Build GRCh38
rs545791855 Chr.14: 55353438-55353438 on Build GRCh38
rs550832802 Chr.14: 55353745-55353745 on Build GRCh38
rs896725738 Chr.14: 55353597-55353597 on Build GRCh38
rs950348838 Chr.14: 55353468-55353468 on Build GRCh38
rs926571145 Chr.14: 55353548-55353548 on Build GRCh38
rs756947245 Chr.14: 55353542-55353542 on Build GRCh38
rs922038242 Chr.14: 55353524-55353524 on Build GRCh38
rs938438276 Chr.14: 55353616-55353616 on Build GRCh38
rs948250360 Chr.14: 55353578-55353578 on Build GRCh38
rs750438865 Chr.14: 55353676-55353676 on Build GRCh38
rs561975935 Chr.14: 55353704-55353704 on Build GRCh38

View Full Product Details