Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 56605432-56605951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM260
Entrez Gene ID
54916
Gene Name
transmembrane protein 260
Gene Aliases
C14orf101
UniGene
Hs.497253
Gene Chromosome Location
Chr.14: 56579791-56660629 on Build GRCh38
Transcripts
NM_017799.3 XM_005267771.1 XM_006720176.1 XM_006720178.1 XM_011536851.1 XM_011536852.1 XM_011536853.1 XM_017021379.1 XM_017021380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4417654 c.573C>T p.H191H Chr.14: 56605620-56605620 on Build GRCh38
87967 c.535G>A p.G179S Chr.14: 56605582-56605582 on Build GRCh38
3987690 c.587A>C p.Y196S Chr.14: 56605634-56605634 on Build GRCh38
3386587 c.523-2A>G p.? Chr.14: 56605568-56605568 on Build GRCh38
6803989 c.570G>A p.Q190Q Chr.14: 56605617-56605617 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557004874 Chr.14: 56605803-56605803 on Build GRCh38
rs924810513 Chr.14: 56605804-56605804 on Build GRCh38
rs775622106 Chr.14: 56605618-56605618 on Build GRCh38
rs761785568 Chr.14: 56605624-56605624 on Build GRCh38
rs977531529 Chr.14: 56605688-56605688 on Build GRCh38
rs370555117 Chr.14: 56605514-56605514 on Build GRCh38
rs775349797 Chr.14: 56605542-56605542 on Build GRCh38
rs996498480 Chr.14: 56605915-56605915 on Build GRCh38
rs757755990 Chr.14: 56605670-56605670 on Build GRCh38
rs779795892 Chr.14: 56605725-56605725 on Build GRCh38
rs779435739 Chr.14: 56605673-56605673 on Build GRCh38
rs113870154 Chr.14: 56605545-56605545 on Build GRCh38
rs1055912231 Chr.14: 56605497-56605497 on Build GRCh38
rs111378942 Chr.14: 56605467-56605468 on Build GRCh38
rs3818777 Chr.14: 56605485-56605485 on Build GRCh38
rs372375253 Chr.14: 56605593-56605593 on Build GRCh38
rs12891203 Chr.14: 56605703-56605703 on Build GRCh38
rs749998387 Chr.14: 56605662-56605662 on Build GRCh38
rs769421400 Chr.14: 56605718-56605718 on Build GRCh38
rs936228684 Chr.14: 56605812-56605812 on Build GRCh38
rs977435796 Chr.14: 56605474-56605474 on Build GRCh38
rs749326862 Chr.14: 56605546-56605546 on Build GRCh38
rs887444106 Chr.14: 56605819-56605819 on Build GRCh38
rs895954099 Chr.14: 56605528-56605528 on Build GRCh38
rs202175823 Chr.14: 56605544-56605545 on Build GRCh38
rs78528113 Chr.14: 56605710-56605710 on Build GRCh38
rs762594468 Chr.14: 56605634-56605634 on Build GRCh38
rs760424506 Chr.14: 56605544-56605544 on Build GRCh38
rs1000450515 Chr.14: 56605587-56605587 on Build GRCh38
rs761771123 Chr.14: 56605720-56605720 on Build GRCh38
rs534919428 Chr.14: 56605795-56605795 on Build GRCh38
rs751129296 Chr.14: 56605651-56605651 on Build GRCh38
rs548127922 Chr.14: 56605703-56605703 on Build GRCh38
rs749724466 Chr.14: 56605608-56605608 on Build GRCh38
rs1003969972 Chr.14: 56605672-56605672 on Build GRCh38
rs3737170 Chr.14: 56605620-56605620 on Build GRCh38
rs199850880 Chr.14: 56605632-56605632 on Build GRCh38
rs190689121 Chr.14: 56605471-56605471 on Build GRCh38
rs754255546 Chr.14: 56605669-56605669 on Build GRCh38
rs533390901 Chr.14: 56605691-56605691 on Build GRCh38
rs769828447 Chr.14: 56605734-56605734 on Build GRCh38
rs746013784 Chr.14: 56605617-56605617 on Build GRCh38
rs1045858873 Chr.14: 56605514-56605514 on Build GRCh38
rs746914324 Chr.14: 56605702-56605703 on Build GRCh38
rs754661988 Chr.14: 56605663-56605663 on Build GRCh38
rs761239959 Chr.14: 56605475-56605475 on Build GRCh38
rs908440441 Chr.14: 56605469-56605469 on Build GRCh38
rs749968028 Chr.14: 56605557-56605557 on Build GRCh38
rs3737169 Chr.14: 56605925-56605925 on Build GRCh38
rs765327310 Chr.14: 56605626-56605626 on Build GRCh38
rs568262674 Chr.14: 56605845-56605845 on Build GRCh38
rs776491378 Chr.14: 56605622-56605622 on Build GRCh38
rs529425398 Chr.14: 56605614-56605614 on Build GRCh38
rs771995331 Chr.14: 56605687-56605687 on Build GRCh38
rs758113994 Chr.14: 56605589-56605589 on Build GRCh38
rs754897636 Chr.14: 56605884-56605884 on Build GRCh38
rs564477201 Chr.14: 56605556-56605556 on Build GRCh38
rs79990465 Chr.14: 56605712-56605712 on Build GRCh38
rs745918339 Chr.14: 56605776-56605786 on Build GRCh38
rs17091787 Chr.14: 56605476-56605476 on Build GRCh38
rs981653165 Chr.14: 56605781-56605781 on Build GRCh38
rs745924486 Chr.14: 56605684-56605684 on Build GRCh38
rs1047505027 Chr.14: 56605809-56605809 on Build GRCh38
rs369707939 Chr.14: 56605529-56605529 on Build GRCh38
rs141128740 Chr.14: 56605600-56605600 on Build GRCh38
rs759693047 Chr.14: 56605523-56605523 on Build GRCh38
rs920583726 Chr.14: 56605715-56605715 on Build GRCh38
rs747958399 Chr.14: 56605708-56605711 on Build GRCh38
rs59106153 Chr.14: 56605556-56605557 on Build GRCh38
rs948916929 Chr.14: 56605499-56605499 on Build GRCh38
rs777937475 Chr.14: 56605668-56605668 on Build GRCh38
rs768526529 Chr.14: 56605703-56605706 on Build GRCh38
rs570806546 Chr.14: 56605794-56605794 on Build GRCh38
rs962601104 Chr.14: 56605689-56605689 on Build GRCh38
rs900104425 Chr.14: 56605903-56605903 on Build GRCh38
rs376685584 Chr.14: 56605619-56605619 on Build GRCh38
rs920636195 Chr.14: 56605477-56605477 on Build GRCh38
rs747163313 Chr.14: 56605602-56605602 on Build GRCh38
rs75417163 Chr.14: 56605711-56605711 on Build GRCh38
rs941704891 Chr.14: 56605831-56605831 on Build GRCh38
rs182027035 Chr.14: 56605923-56605923 on Build GRCh38
rs1054739293 Chr.14: 56605897-56605897 on Build GRCh38
rs772864422 Chr.14: 56605731-56605732 on Build GRCh38
rs955315296 Chr.14: 56605656-56605656 on Build GRCh38
rs757436723 Chr.14: 56605545-56605546 on Build GRCh38
rs991916812 Chr.14: 56605768-56605768 on Build GRCh38
rs766035551 Chr.14: 56605640-56605640 on Build GRCh38
rs573194291 Chr.14: 56605584-56605584 on Build GRCh38
rs1054747594 Chr.14: 56605490-56605490 on Build GRCh38
rs983042141 Chr.14: 56605461-56605461 on Build GRCh38
rs779641681 Chr.14: 56605592-56605592 on Build GRCh38
rs776278372 Chr.14: 56605707-56605707 on Build GRCh38
rs200631835 Chr.14: 56605606-56605606 on Build GRCh38
rs541594173 Chr.14: 56605799-56605799 on Build GRCh38
rs753670504 Chr.14: 56605552-56605552 on Build GRCh38
rs577189355 Chr.14: 56605707-56605711 on Build GRCh38
rs138673596 Chr.14: 56605657-56605657 on Build GRCh38

View Full Product Details