Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 57409190-57409694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAA30
Entrez Gene ID
122830
Gene Name
N(alpha)-acetyltransferase 30, NatC catalytic subunit
Gene Aliases
C14orf35, MAK3, Mak3p, NAT12, NAT12P
UniGene
Hs.165465
Gene Chromosome Location
Chr.14: 57390553-57412748 on Build GRCh38
Transcripts
NM_001011713.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4799969 c.1072A>C p.K358Q Chr.14: 57409499-57409499 on Build GRCh38
289881 c.994C>G p.L332V Chr.14: 57409421-57409421 on Build GRCh38
390913 c.1018C>T p.R340* Chr.14: 57409445-57409445 on Build GRCh38
1678008 c.1019G>A p.R340Q Chr.14: 57409446-57409446 on Build GRCh38
1607767 c.1010G>T p.G337V Chr.14: 57409437-57409437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs532951433 Chr.14: 57409222-57409222 on Build GRCh38
rs1019254018 Chr.14: 57409629-57409629 on Build GRCh38
rs757667781 Chr.14: 57409459-57409459 on Build GRCh38
rs1055743358 Chr.14: 57409270-57409270 on Build GRCh38
rs770923060 Chr.14: 57409402-57409405 on Build GRCh38
rs765247624 Chr.14: 57409360-57409360 on Build GRCh38
rs545938313 Chr.14: 57409293-57409293 on Build GRCh38
rs368332192 Chr.14: 57409564-57409564 on Build GRCh38
rs761744610 Chr.14: 57409344-57409344 on Build GRCh38
rs957863898 Chr.14: 57409468-57409468 on Build GRCh38
rs988009238 Chr.14: 57409294-57409294 on Build GRCh38
rs780855072 Chr.14: 57409400-57409400 on Build GRCh38
rs916796708 Chr.14: 57409662-57409662 on Build GRCh38
rs557936194 Chr.14: 57409653-57409653 on Build GRCh38
rs564480039 Chr.14: 57409303-57409303 on Build GRCh38
rs11620639 Chr.14: 57409547-57409547 on Build GRCh38
rs752402671 Chr.14: 57409565-57409565 on Build GRCh38
rs77940271 Chr.14: 57409474-57409474 on Build GRCh38
rs528783212 Chr.14: 57409337-57409337 on Build GRCh38
rs748909868 Chr.14: 57409556-57409556 on Build GRCh38
rs911713901 Chr.14: 57409237-57409237 on Build GRCh38
rs772634919 Chr.14: 57409513-57409513 on Build GRCh38
rs756618193 Chr.14: 57409511-57409511 on Build GRCh38
rs768190446 Chr.14: 57409292-57409292 on Build GRCh38
rs148748825 Chr.14: 57409411-57409411 on Build GRCh38
rs367603636 Chr.14: 57409333-57409333 on Build GRCh38
rs771974311 Chr.14: 57409634-57409634 on Build GRCh38
rs779376883 Chr.14: 57409471-57409471 on Build GRCh38
rs367695961 Chr.14: 57409354-57409354 on Build GRCh38
rs539120878 Chr.14: 57409615-57409615 on Build GRCh38
rs757612706 Chr.14: 57409566-57409566 on Build GRCh38
rs1029295892 Chr.14: 57409530-57409530 on Build GRCh38
rs760738220 Chr.14: 57409335-57409335 on Build GRCh38
rs528520377 Chr.14: 57409620-57409620 on Build GRCh38
rs772099582 Chr.14: 57409328-57409328 on Build GRCh38
rs1037484535 Chr.14: 57409228-57409228 on Build GRCh38
rs142565418 Chr.14: 57409371-57409371 on Build GRCh38
rs576200591 Chr.14: 57409535-57409535 on Build GRCh38
rs779938134 Chr.14: 57409534-57409534 on Build GRCh38
rs527809583 Chr.14: 57409289-57409292 on Build GRCh38
rs762743023 Chr.14: 57409550-57409550 on Build GRCh38
rs750446409 Chr.14: 57409366-57409366 on Build GRCh38
rs768716763 Chr.14: 57409540-57409540 on Build GRCh38
rs913624779 Chr.14: 57409301-57409301 on Build GRCh38
rs940402729 Chr.14: 57409227-57409227 on Build GRCh38
rs776514888 Chr.14: 57409341-57409341 on Build GRCh38
rs1014817243 Chr.14: 57409479-57409479 on Build GRCh38
rs895969219 Chr.14: 57409412-57409412 on Build GRCh38
rs115958610 Chr.14: 57409255-57409255 on Build GRCh38
rs567732974 Chr.14: 57409542-57409542 on Build GRCh38
rs902548600 Chr.14: 57409304-57409304 on Build GRCh38
rs150950524 Chr.14: 57409557-57409557 on Build GRCh38
rs778444961 Chr.14: 57409575-57409575 on Build GRCh38
rs766363326 Chr.14: 57409551-57409551 on Build GRCh38
rs1055312235 Chr.14: 57409635-57409635 on Build GRCh38
rs996027637 Chr.14: 57409489-57409489 on Build GRCh38
rs747130140 Chr.14: 57409538-57409538 on Build GRCh38
rs372759259 Chr.14: 57409373-57409373 on Build GRCh38
rs761691487 Chr.14: 57409545-57409545 on Build GRCh38
rs535065694 Chr.14: 57409546-57409546 on Build GRCh38
rs754538525 Chr.14: 57409393-57409393 on Build GRCh38
rs920081142 Chr.14: 57409233-57409233 on Build GRCh38
rs986485272 Chr.14: 57409218-57409218 on Build GRCh38
rs77432220 Chr.14: 57409460-57409460 on Build GRCh38
rs966001638 Chr.14: 57409644-57409644 on Build GRCh38

View Full Product Details