Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 57411179-57411728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAA30
Entrez Gene ID
122830
Gene Name
N(alpha)-acetyltransferase 30, NatC catalytic subunit
Gene Aliases
C14orf35, MAK3, Mak3p, NAT12, NAT12P
UniGene
Hs.165465
Gene Chromosome Location
Chr.14: 57390553-57412748 on Build GRCh38
Transcripts
NM_001011713.2

SNP Detail

SNP ID Chromosome Location
rs750127921 Chr.14: 57411678-57411678 on Build GRCh38
rs1008217919 Chr.14: 57411689-57411689 on Build GRCh38
rs978198672 Chr.14: 57411233-57411237 on Build GRCh38
rs368953048 Chr.14: 57411617-57411617 on Build GRCh38
rs892899597 Chr.14: 57411630-57411630 on Build GRCh38
rs529546602 Chr.14: 57411290-57411290 on Build GRCh38
rs1049383176 Chr.14: 57411487-57411487 on Build GRCh38
rs915808218 Chr.14: 57411519-57411519 on Build GRCh38
rs1030499598 Chr.14: 57411584-57411584 on Build GRCh38
rs544139912 Chr.14: 57411250-57411250 on Build GRCh38
rs953775521 Chr.14: 57411328-57411328 on Build GRCh38
rs1011050805 Chr.14: 57411651-57411651 on Build GRCh38
rs376225587 Chr.14: 57411562-57411562 on Build GRCh38
rs966889163 Chr.14: 57411664-57411664 on Build GRCh38
rs371698192 Chr.14: 57411557-57411557 on Build GRCh38
rs760490892 Chr.14: 57411305-57411305 on Build GRCh38
rs900715790 Chr.14: 57411564-57411564 on Build GRCh38
rs562084264 Chr.14: 57411275-57411275 on Build GRCh38
rs188454194 Chr.14: 57411295-57411295 on Build GRCh38
rs986422050 Chr.14: 57411348-57411348 on Build GRCh38
rs549205481 Chr.14: 57411473-57411473 on Build GRCh38
rs551484759 Chr.14: 57411405-57411405 on Build GRCh38
rs953601589 Chr.14: 57411685-57411685 on Build GRCh38
rs566443933 Chr.14: 57411431-57411431 on Build GRCh38
rs184210584 Chr.14: 57411441-57411441 on Build GRCh38
rs764966818 Chr.14: 57411337-57411337 on Build GRCh38
rs571074563 Chr.14: 57411542-57411542 on Build GRCh38
rs1019247307 Chr.14: 57411696-57411696 on Build GRCh38
rs925253747 Chr.14: 57411243-57411243 on Build GRCh38
rs573127541 Chr.14: 57411222-57411222 on Build GRCh38
rs866964878 Chr.14: 57411529-57411529 on Build GRCh38
rs911884624 Chr.14: 57411380-57411380 on Build GRCh38
rs929714094 Chr.14: 57411508-57411508 on Build GRCh38
rs1059481 Chr.14: 57411516-57411516 on Build GRCh38
rs956829217 Chr.14: 57411272-57411272 on Build GRCh38
rs966068845 Chr.14: 57411396-57411396 on Build GRCh38
rs972140597 Chr.14: 57411400-57411400 on Build GRCh38
rs998218107 Chr.14: 57411574-57411574 on Build GRCh38
rs991240061 Chr.14: 57411322-57411322 on Build GRCh38
rs551457598 Chr.14: 57411535-57411535 on Build GRCh38
rs1052769599 Chr.14: 57411576-57411576 on Build GRCh38
rs1031420019 Chr.14: 57411241-57411241 on Build GRCh38
rs948629095 Chr.14: 57411527-57411527 on Build GRCh38
rs181300393 Chr.14: 57411401-57411401 on Build GRCh38
rs138775751 Chr.14: 57411503-57411503 on Build GRCh38
rs998477081 Chr.14: 57411551-57411551 on Build GRCh38
rs562520874 Chr.14: 57411368-57411368 on Build GRCh38
rs888215884 Chr.14: 57411531-57411531 on Build GRCh38
rs897027194 Chr.14: 57411483-57411483 on Build GRCh38
rs1022865882 Chr.14: 57411676-57411676 on Build GRCh38
rs916466066 Chr.14: 57411428-57411428 on Build GRCh38
rs747944275 Chr.14: 57411417-57411417 on Build GRCh38

View Full Product Details