Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 58012032-58012534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf37
Entrez Gene ID
145407
Gene Name
chromosome 14 open reading frame 37
Gene Aliases
c14_5376
UniGene
Hs.26766
Gene Chromosome Location
Chr.14: 57993536-58152305 on Build GRCh38
Transcripts
NM_001001872.3 XM_017021029.1 XM_017021030.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5435640 c.2091G>A p.V697V Chr.14: 58012149-58012149 on Build GRCh38
4929137 c.2108A>G p.K703R Chr.14: 58012132-58012132 on Build GRCh38
6614316 c.2096G>T p.S699I Chr.14: 58012144-58012144 on Build GRCh38
7228124 c.2113A>T p.K705* Chr.14: 58012127-58012127 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs563779299 Chr.14: 58012192-58012192 on Build GRCh38
rs368482151 Chr.14: 58012142-58012142 on Build GRCh38
rs929413175 Chr.14: 58012405-58012405 on Build GRCh38
rs973482803 Chr.14: 58012226-58012226 on Build GRCh38
rs773963051 Chr.14: 58012166-58012168 on Build GRCh38
rs760920004 Chr.14: 58012474-58012474 on Build GRCh38
rs917605455 Chr.14: 58012056-58012056 on Build GRCh38
rs528459408 Chr.14: 58012270-58012270 on Build GRCh38
rs756723324 Chr.14: 58012107-58012107 on Build GRCh38
rs3783689 Chr.14: 58012272-58012272 on Build GRCh38
rs568619580 Chr.14: 58012375-58012375 on Build GRCh38
rs1007966198 Chr.14: 58012164-58012164 on Build GRCh38
rs398077780 Chr.14: 58012161-58012161 on Build GRCh38
rs781668450 Chr.14: 58012122-58012122 on Build GRCh38
rs536093000 Chr.14: 58012283-58012283 on Build GRCh38
rs774572558 Chr.14: 58012076-58012076 on Build GRCh38
rs777415867 Chr.14: 58012109-58012110 on Build GRCh38
rs1026342642 Chr.14: 58012500-58012502 on Build GRCh38
rs371099544 Chr.14: 58012103-58012103 on Build GRCh38
rs150566056 Chr.14: 58012116-58012116 on Build GRCh38
rs1040565102 Chr.14: 58012367-58012367 on Build GRCh38
rs72714763 Chr.14: 58012279-58012279 on Build GRCh38
rs770376556 Chr.14: 58012155-58012156 on Build GRCh38
rs542155112 Chr.14: 58012465-58012465 on Build GRCh38
rs748572304 Chr.14: 58012123-58012123 on Build GRCh38
rs775250185 Chr.14: 58012185-58012185 on Build GRCh38
rs371174030 Chr.14: 58012186-58012186 on Build GRCh38
rs892277392 Chr.14: 58012113-58012113 on Build GRCh38
rs763410602 Chr.14: 58012414-58012414 on Build GRCh38
rs746054614 Chr.14: 58012171-58012171 on Build GRCh38
rs756574204 Chr.14: 58012124-58012124 on Build GRCh38
rs3783687 Chr.14: 58012223-58012223 on Build GRCh38
rs1044853424 Chr.14: 58012087-58012087 on Build GRCh38
rs3783688 Chr.14: 58012261-58012261 on Build GRCh38
rs759774886 Chr.14: 58012081-58012081 on Build GRCh38
rs1037716598 Chr.14: 58012340-58012340 on Build GRCh38
rs899141556 Chr.14: 58012460-58012460 on Build GRCh38
rs375045147 Chr.14: 58012163-58012163 on Build GRCh38
rs981229598 Chr.14: 58012222-58012222 on Build GRCh38
rs764670139 Chr.14: 58012108-58012108 on Build GRCh38
rs969890811 Chr.14: 58012201-58012201 on Build GRCh38
rs373329436 Chr.14: 58012084-58012084 on Build GRCh38
rs753286790 Chr.14: 58012104-58012104 on Build GRCh38
rs749770477 Chr.14: 58012152-58012152 on Build GRCh38
rs770992308 Chr.14: 58012166-58012166 on Build GRCh38
rs892329752 Chr.14: 58012371-58012371 on Build GRCh38
rs78167789 Chr.14: 58012287-58012287 on Build GRCh38
rs887816399 Chr.14: 58012438-58012438 on Build GRCh38
rs371729614 Chr.14: 58012162-58012162 on Build GRCh38
rs774105240 Chr.14: 58012501-58012501 on Build GRCh38
rs10141224 Chr.14: 58012097-58012097 on Build GRCh38
rs1026459074 Chr.14: 58012236-58012236 on Build GRCh38
rs539110889 Chr.14: 58012420-58012420 on Build GRCh38
rs1022868268 Chr.14: 58012471-58012471 on Build GRCh38
rs936434407 Chr.14: 58012343-58012343 on Build GRCh38
rs3837616 Chr.14: 58012156-58012156 on Build GRCh38
rs988928929 Chr.14: 58012263-58012263 on Build GRCh38
rs772327822 Chr.14: 58012172-58012172 on Build GRCh38
rs750050143 Chr.14: 58012109-58012109 on Build GRCh38
rs182392105 Chr.14: 58012268-58012268 on Build GRCh38
rs397841275 Chr.14: 58012162-58012162 on Build GRCh38
rs3783690 Chr.14: 58012437-58012437 on Build GRCh38
rs201263862 Chr.14: 58012187-58012187 on Build GRCh38
rs1051717057 Chr.14: 58012413-58012413 on Build GRCh38
rs775301936 Chr.14: 58012086-58012086 on Build GRCh38

View Full Product Details