Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 58638365-58638858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DACT1
Entrez Gene ID
51339
Gene Name
dishevelled binding antagonist of beta catenin 1
Gene Aliases
DAPPER, DAPPER1, DPR1, FRODO, HDPR1, THYEX3
UniGene
Hs.48950
Gene Chromosome Location
Chr.14: 58634068-58648321 on Build GRCh38
Transcripts
NM_001079520.1 NM_016651.5 NR_046093.1 NR_046095.1 XM_006720167.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
956580 c.287G>A p.R96H Chr.14: 58638489-58638489 on Build GRCh38
6399979 c.296T>A p.L99Q Chr.14: 58638498-58638498 on Build GRCh38
394993 c.230G>T p.G77V Chr.14: 58638432-58638432 on Build GRCh38
3749277 c.268C>T p.L90L Chr.14: 58638470-58638470 on Build GRCh38
4676156 c.297_299delGGA p.E101delE Chr.14: 58638499-58638501 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973585715 Chr.14: 58638737-58638737 on Build GRCh38
rs770910849 Chr.14: 58638748-58638748 on Build GRCh38
rs1011452623 Chr.14: 58638576-58638576 on Build GRCh38
rs796860556 Chr.14: 58638643-58638643 on Build GRCh38
rs369739247 Chr.14: 58638409-58638409 on Build GRCh38
rs757357488 Chr.14: 58638445-58638445 on Build GRCh38
rs1043270684 Chr.14: 58638810-58638812 on Build GRCh38
rs943708631 Chr.14: 58638637-58638637 on Build GRCh38
rs764715977 Chr.14: 58638502-58638502 on Build GRCh38
rs975596200 Chr.14: 58638450-58638450 on Build GRCh38
rs908821481 Chr.14: 58638719-58638719 on Build GRCh38
rs373342908 Chr.14: 58638493-58638493 on Build GRCh38
rs767731225 Chr.14: 58638431-58638431 on Build GRCh38
rs774087743 Chr.14: 58638504-58638506 on Build GRCh38
rs199621547 Chr.14: 58638508-58638508 on Build GRCh38
rs759360093 Chr.14: 58638514-58638516 on Build GRCh38
rs921178086 Chr.14: 58638810-58638810 on Build GRCh38
rs563921752 Chr.14: 58638803-58638803 on Build GRCh38
rs1043882349 Chr.14: 58638395-58638395 on Build GRCh38
rs912191335 Chr.14: 58638625-58638625 on Build GRCh38
rs774092710 Chr.14: 58638580-58638580 on Build GRCh38
rs774410049 Chr.14: 58638778-58638778 on Build GRCh38
rs772946799 Chr.14: 58638490-58638490 on Build GRCh38
rs1000685661 Chr.14: 58638653-58638653 on Build GRCh38
rs773576064 Chr.14: 58638695-58638695 on Build GRCh38
rs748203756 Chr.14: 58638455-58638455 on Build GRCh38
rs1039614674 Chr.14: 58638641-58638641 on Build GRCh38
rs371248818 Chr.14: 58638661-58638661 on Build GRCh38
rs770591746 Chr.14: 58638495-58638495 on Build GRCh38
rs1026458104 Chr.14: 58638755-58638755 on Build GRCh38
rs769117476 Chr.14: 58638483-58638483 on Build GRCh38
rs750573800 Chr.14: 58638453-58638453 on Build GRCh38
rs963572144 Chr.14: 58638541-58638541 on Build GRCh38
rs759559327 Chr.14: 58638809-58638809 on Build GRCh38
rs765185034 Chr.14: 58638511-58638511 on Build GRCh38
rs984686711 Chr.14: 58638717-58638717 on Build GRCh38
rs1025017184 Chr.14: 58638672-58638672 on Build GRCh38
rs768648129 Chr.14: 58638468-58638468 on Build GRCh38
rs761281299 Chr.14: 58638413-58638413 on Build GRCh38
rs2003021 Chr.14: 58638470-58638470 on Build GRCh38
rs746776399 Chr.14: 58638465-58638465 on Build GRCh38
rs754709809 Chr.14: 58638547-58638547 on Build GRCh38
rs1032423630 Chr.14: 58638452-58638452 on Build GRCh38
rs777195162 Chr.14: 58638571-58638571 on Build GRCh38
rs376256989 Chr.14: 58638512-58638512 on Build GRCh38
rs772693125 Chr.14: 58638419-58638419 on Build GRCh38
rs761112902 Chr.14: 58638501-58638501 on Build GRCh38
rs961911793 Chr.14: 58638731-58638731 on Build GRCh38
rs746862608 Chr.14: 58638400-58638400 on Build GRCh38
rs780319203 Chr.14: 58638526-58638526 on Build GRCh38
rs563143762 Chr.14: 58638676-58638676 on Build GRCh38
rs781110719 Chr.14: 58638557-58638557 on Build GRCh38
rs1002650934 Chr.14: 58638679-58638679 on Build GRCh38
rs747669823 Chr.14: 58638473-58638473 on Build GRCh38
rs996561262 Chr.14: 58638701-58638701 on Build GRCh38
rs753266802 Chr.14: 58638432-58638432 on Build GRCh38
rs762806147 Chr.14: 58638431-58638431 on Build GRCh38
rs184788885 Chr.14: 58638820-58638820 on Build GRCh38
rs748037387 Chr.14: 58638560-58638560 on Build GRCh38
rs747386506 Chr.14: 58638596-58638596 on Build GRCh38
rs754332840 Chr.14: 58638506-58638506 on Build GRCh38
rs981009327 Chr.14: 58638476-58638476 on Build GRCh38
rs764661038 Chr.14: 58638440-58638440 on Build GRCh38
rs779204323 Chr.14: 58638451-58638451 on Build GRCh38
rs912347705 Chr.14: 58638801-58638801 on Build GRCh38
rs755669640 Chr.14: 58638569-58638569 on Build GRCh38
rs767258847 Chr.14: 58638564-58638564 on Build GRCh38
rs770552272 Chr.14: 58638578-58638578 on Build GRCh38
rs938105703 Chr.14: 58638788-58638788 on Build GRCh38
rs1055095783 Chr.14: 58638793-58638793 on Build GRCh38
rs974021529 Chr.14: 58638556-58638556 on Build GRCh38
rs199649683 Chr.14: 58638462-58638462 on Build GRCh38
rs892261681 Chr.14: 58638828-58638828 on Build GRCh38
rs766320869 Chr.14: 58638499-58638501 on Build GRCh38
rs952896037 Chr.14: 58638709-58638709 on Build GRCh38
rs1009583459 Chr.14: 58638670-58638670 on Build GRCh38
rs545731479 Chr.14: 58638794-58638794 on Build GRCh38
rs1040028427 Chr.14: 58638391-58638391 on Build GRCh38
rs927940392 Chr.14: 58638768-58638768 on Build GRCh38
rs762018488 Chr.14: 58638507-58638507 on Build GRCh38
rs963992421 Chr.14: 58638736-58638736 on Build GRCh38
rs999147567 Chr.14: 58638816-58638816 on Build GRCh38
rs768566527 Chr.14: 58638403-58638403 on Build GRCh38
rs899629060 Chr.14: 58638422-58638422 on Build GRCh38
rs769486207 Chr.14: 58638418-58638418 on Build GRCh38
rs373138468 Chr.14: 58638439-58638439 on Build GRCh38
rs892312668 Chr.14: 58638669-58638669 on Build GRCh38
rs919659151 Chr.14: 58638577-58638577 on Build GRCh38
rs193284405 Chr.14: 58638667-58638667 on Build GRCh38
rs751414282 Chr.14: 58638533-58638533 on Build GRCh38
rs1019673080 Chr.14: 58638673-58638673 on Build GRCh38
rs758589000 Chr.14: 58638514-58638514 on Build GRCh38
rs929742948 Chr.14: 58638581-58638581 on Build GRCh38
rs148938592 Chr.14: 58638704-58638704 on Build GRCh38
rs754034136 Chr.14: 58638443-58638443 on Build GRCh38
rs1053648413 Chr.14: 58638823-58638823 on Build GRCh38
rs759948805 Chr.14: 58638428-58638428 on Build GRCh38
rs574279826 Chr.14: 58638602-58638602 on Build GRCh38
rs1055567140 Chr.14: 58638559-58638559 on Build GRCh38

View Full Product Details