Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 59521551-59522060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC175
Entrez Gene ID
729665
Gene Name
coiled-coil domain containing 175
Gene Aliases
C14orf38, c14_5395
UniGene
Hs.29706
Gene Chromosome Location
Chr.14: 59504212-59577276 on Build GRCh38
Transcripts
NM_001164399.1 XM_011537120.2 XM_011537122.2 XM_011537124.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3956113 c.2098G>T p.A700S Chr.14: 59521574-59521574 on Build GRCh38
3956115 c.2097G>T p.E699D Chr.14: 59521575-59521575 on Build GRCh38
5425550 c.2066G>A p.S689N Chr.14: 59521606-59521606 on Build GRCh38
3956114 c.2098G>T p.A700S Chr.14: 59521574-59521574 on Build GRCh38
5425549 c.2066G>A p.S689N Chr.14: 59521606-59521606 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs930949570 Chr.14: 59521855-59521855 on Build GRCh38
rs547685148 Chr.14: 59521992-59521992 on Build GRCh38
rs565206117 Chr.14: 59521821-59521821 on Build GRCh38
rs774473757 Chr.14: 59521658-59521658 on Build GRCh38
rs1010850248 Chr.14: 59522004-59522004 on Build GRCh38
rs751396780 Chr.14: 59521687-59521687 on Build GRCh38
rs944828255 Chr.14: 59521884-59521884 on Build GRCh38
rs901112118 Chr.14: 59521944-59521944 on Build GRCh38
rs569278905 Chr.14: 59521988-59521988 on Build GRCh38
rs913716991 Chr.14: 59521589-59521589 on Build GRCh38
rs529783686 Chr.14: 59521895-59521895 on Build GRCh38
rs996697729 Chr.14: 59521965-59521965 on Build GRCh38
rs1028251073 Chr.14: 59521756-59521756 on Build GRCh38
rs1021260687 Chr.14: 59522011-59522011 on Build GRCh38
rs1006749317 Chr.14: 59522007-59522007 on Build GRCh38
rs12887189 Chr.14: 59521606-59521606 on Build GRCh38
rs75578137 Chr.14: 59521654-59521654 on Build GRCh38
rs182811879 Chr.14: 59521774-59521774 on Build GRCh38
rs4394993 Chr.14: 59521605-59521605 on Build GRCh38
rs938324928 Chr.14: 59521583-59521583 on Build GRCh38
rs73305628 Chr.14: 59521594-59521594 on Build GRCh38
rs148869013 Chr.14: 59521866-59521866 on Build GRCh38
rs752225076 Chr.14: 59521713-59521713 on Build GRCh38
rs763372372 Chr.14: 59521635-59521635 on Build GRCh38
rs1009010280 Chr.14: 59521716-59521716 on Build GRCh38
rs981838719 Chr.14: 59521781-59521781 on Build GRCh38
rs966532656 Chr.14: 59522024-59522024 on Build GRCh38
rs927695198 Chr.14: 59521800-59521800 on Build GRCh38
rs192960993 Chr.14: 59521722-59521722 on Build GRCh38
rs556834271 Chr.14: 59521593-59521593 on Build GRCh38
rs754559515 Chr.14: 59521690-59521690 on Build GRCh38
rs767096280 Chr.14: 59521696-59521696 on Build GRCh38
rs759609240 Chr.14: 59521760-59521760 on Build GRCh38
rs909376826 Chr.14: 59521769-59521769 on Build GRCh38
rs908064788 Chr.14: 59521873-59521873 on Build GRCh38
rs886605188 Chr.14: 59521695-59521695 on Build GRCh38
rs531746140 Chr.14: 59521782-59521782 on Build GRCh38
rs532184658 Chr.14: 59521840-59521840 on Build GRCh38
rs192460689 Chr.14: 59521893-59521893 on Build GRCh38
rs900553357 Chr.14: 59521945-59521945 on Build GRCh38
rs920249672 Chr.14: 59521830-59521830 on Build GRCh38
rs748572105 Chr.14: 59521595-59521595 on Build GRCh38
rs776867675 Chr.14: 59521588-59521588 on Build GRCh38
rs928852714 Chr.14: 59521777-59521777 on Build GRCh38
rs547881574 Chr.14: 59521947-59521947 on Build GRCh38
rs996254175 Chr.14: 59521987-59521987 on Build GRCh38
rs958015290 Chr.14: 59521759-59521759 on Build GRCh38
rs981806129 Chr.14: 59521604-59521604 on Build GRCh38
rs187488250 Chr.14: 59521775-59521775 on Build GRCh38
rs966930596 Chr.14: 59521780-59521780 on Build GRCh38
rs894108717 Chr.14: 59521802-59521802 on Build GRCh38
rs1047926531 Chr.14: 59521691-59521691 on Build GRCh38
rs768670805 Chr.14: 59521584-59521584 on Build GRCh38
rs900520848 Chr.14: 59521724-59521724 on Build GRCh38
rs751261957 Chr.14: 59521737-59521737 on Build GRCh38
rs996139382 Chr.14: 59521755-59521755 on Build GRCh38
rs17096349 Chr.14: 59521622-59521622 on Build GRCh38
rs1055481457 Chr.14: 59521791-59521791 on Build GRCh38
rs1040483741 Chr.14: 59521941-59521941 on Build GRCh38
rs529339067 Chr.14: 59521811-59521811 on Build GRCh38
rs202077551 Chr.14: 59521623-59521623 on Build GRCh38

View Full Product Details