Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 64211870-64212368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNE2
Entrez Gene ID
23224
Gene Name
spectrin repeat containing nuclear envelope protein 2
Gene Aliases
EDMD5, NUA, NUANCE, Nesp2, Nesprin-2, SYNE-2, TROPH
UniGene
Hs.745014
Gene Chromosome Location
Chr.14: 63761904-64226449 on Build GRCh38
Transcripts
NM_015180.4 NM_182914.2 XM_005267454.1 XM_005267456.1 XM_005267457.1 XM_005267458.1 XM_005267459.1 XM_011536574.1 XM_011536575.2 XM_011536576.2 XM_011536577.2 XM_011536578.1 XM_011536579.1 XM_011536580.1 XM_011536581.1 XM_011536582.1 XM_017021101.1 XM_017021102.1 XM_017021103.1 XM_017021104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5661469 c.7900G>A p.E2634K Chr.14: 64211982-64211982 on Build GRCh38
3815086 c.18851G>A p.R6284H Chr.14: 64212088-64212088 on Build GRCh38
4532349 c.18775G>A p.V6259M Chr.14: 64212012-64212012 on Build GRCh38
4926819 c.18729C>A p.F6243L Chr.14: 64211966-64211966 on Build GRCh38
3370018 c.18826G>C p.E6276Q Chr.14: 64212063-64212063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs923446863 Chr.14: 64211956-64211956 on Build GRCh38
rs775495959 Chr.14: 64212128-64212129 on Build GRCh38
rs184676859 Chr.14: 64212113-64212113 on Build GRCh38
rs757626548 Chr.14: 64211944-64211944 on Build GRCh38
rs759249878 Chr.14: 64212128-64212128 on Build GRCh38
rs932035992 Chr.14: 64212011-64212011 on Build GRCh38
rs112482233 Chr.14: 64212157-64212157 on Build GRCh38
rs777458871 Chr.14: 64212275-64212275 on Build GRCh38
rs747042989 Chr.14: 64211975-64211975 on Build GRCh38
rs563426023 Chr.14: 64212074-64212074 on Build GRCh38
rs149842821 Chr.14: 64211911-64211911 on Build GRCh38
rs1048705133 Chr.14: 64212314-64212314 on Build GRCh38
rs370847169 Chr.14: 64211981-64211981 on Build GRCh38
rs757742735 Chr.14: 64212071-64212071 on Build GRCh38
rs1015249801 Chr.14: 64211912-64211912 on Build GRCh38
rs866146605 Chr.14: 64212226-64212226 on Build GRCh38
rs765976678 Chr.14: 64212033-64212033 on Build GRCh38
rs767073602 Chr.14: 64212050-64212050 on Build GRCh38
rs754364107 Chr.14: 64212066-64212066 on Build GRCh38
rs947463668 Chr.14: 64212244-64212244 on Build GRCh38
rs755106430 Chr.14: 64212098-64212098 on Build GRCh38
rs773232957 Chr.14: 64212119-64212119 on Build GRCh38
rs369110774 Chr.14: 64211919-64211919 on Build GRCh38
rs532495528 Chr.14: 64212075-64212075 on Build GRCh38
rs202157687 Chr.14: 64212088-64212088 on Build GRCh38
rs781780825 Chr.14: 64212006-64212006 on Build GRCh38
rs948577796 Chr.14: 64212312-64212312 on Build GRCh38
rs754926501 Chr.14: 64212272-64212272 on Build GRCh38
rs148547914 Chr.14: 64212179-64212179 on Build GRCh38
rs575529846 Chr.14: 64212336-64212336 on Build GRCh38
rs780352939 Chr.14: 64212087-64212087 on Build GRCh38
rs370689732 Chr.14: 64212321-64212321 on Build GRCh38
rs376208379 Chr.14: 64211950-64211950 on Build GRCh38
rs762833113 Chr.14: 64212023-64212023 on Build GRCh38
rs1053902330 Chr.14: 64212204-64212204 on Build GRCh38
rs145871645 Chr.14: 64212072-64212072 on Build GRCh38
rs781173263 Chr.14: 64212105-64212105 on Build GRCh38
rs894416025 Chr.14: 64212290-64212290 on Build GRCh38
rs762557529 Chr.14: 64211917-64211917 on Build GRCh38
rs778188442 Chr.14: 64211930-64211930 on Build GRCh38
rs1056867267 Chr.14: 64212084-64212084 on Build GRCh38
rs1056963141 Chr.14: 64212281-64212281 on Build GRCh38
rs201046983 Chr.14: 64212126-64212126 on Build GRCh38
rs188319000 Chr.14: 64212254-64212254 on Build GRCh38
rs936648242 Chr.14: 64212189-64212189 on Build GRCh38
rs754515394 Chr.14: 64211921-64211921 on Build GRCh38
rs1030614075 Chr.14: 64212168-64212168 on Build GRCh38
rs772471257 Chr.14: 64211964-64211964 on Build GRCh38
rs552893876 Chr.14: 64212198-64212198 on Build GRCh38
rs750950612 Chr.14: 64212047-64212047 on Build GRCh38
rs772933807 Chr.14: 64212012-64212012 on Build GRCh38
rs75672086 Chr.14: 64212171-64212171 on Build GRCh38
rs556957036 Chr.14: 64212276-64212276 on Build GRCh38
rs754317609 Chr.14: 64211936-64211936 on Build GRCh38
rs947404239 Chr.14: 64212053-64212053 on Build GRCh38
rs372991467 Chr.14: 64211925-64211925 on Build GRCh38
rs746070450 Chr.14: 64211958-64211958 on Build GRCh38
rs1003978758 Chr.14: 64212322-64212322 on Build GRCh38
rs748103462 Chr.14: 64211995-64211995 on Build GRCh38
rs776571728 Chr.14: 64211984-64211984 on Build GRCh38
rs199652652 Chr.14: 64212124-64212124 on Build GRCh38
rs952398626 Chr.14: 64211901-64211901 on Build GRCh38
rs188125599 Chr.14: 64212146-64212146 on Build GRCh38
rs769603327 Chr.14: 64212008-64212008 on Build GRCh38
rs758990007 Chr.14: 64212048-64212048 on Build GRCh38
rs372881760 Chr.14: 64212147-64212147 on Build GRCh38
rs183603811 Chr.14: 64212220-64212220 on Build GRCh38
rs574447077 Chr.14: 64212165-64212165 on Build GRCh38
rs749016712 Chr.14: 64212121-64212121 on Build GRCh38
rs765896826 Chr.14: 64212327-64212327 on Build GRCh38
rs774910294 Chr.14: 64212136-64212136 on Build GRCh38
rs780442522 Chr.14: 64211969-64211969 on Build GRCh38
rs1003841629 Chr.14: 64212140-64212140 on Build GRCh38
rs374498035 Chr.14: 64212133-64212133 on Build GRCh38
rs780565577 Chr.14: 64212106-64212106 on Build GRCh38
rs976215020 Chr.14: 64211922-64211922 on Build GRCh38
rs541855640 Chr.14: 64212243-64212243 on Build GRCh38

View Full Product Details