Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 64410958-64411374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTHFD1
Entrez Gene ID
4522
Gene Name
methylenetetrahydrofolate dehydrogenase, cyclohydrolase and formyltetrahydrofolate synthetase 1
Gene Aliases
MTHFC, MTHFD
UniGene
Hs.652308
Gene Chromosome Location
Chr.14: 64388041-64460007 on Build GRCh38
Transcripts
NM_005956.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7167519 c.305G>C p.R102T Chr.14: 64411100-64411100 on Build GRCh38
7159225 c.339G>T p.L113L Chr.14: 64411134-64411134 on Build GRCh38
1665946 c.186+2T>A p.? Chr.14: 64411151-64411151 on Build GRCh38
7167520 c.137G>C p.R46T Chr.14: 64411100-64411100 on Build GRCh38
7159226 c.171G>T p.L57L Chr.14: 64411134-64411134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748129032 Chr.14: 64411057-64411057 on Build GRCh38
rs1031156925 Chr.14: 64411287-64411287 on Build GRCh38
rs931643937 Chr.14: 64411326-64411326 on Build GRCh38
rs555622752 Chr.14: 64411266-64411266 on Build GRCh38
rs781332569 Chr.14: 64411172-64411172 on Build GRCh38
rs368039115 Chr.14: 64411209-64411209 on Build GRCh38
rs370659538 Chr.14: 64411204-64411204 on Build GRCh38
rs756099557 Chr.14: 64411190-64411190 on Build GRCh38
rs924887170 Chr.14: 64410999-64410999 on Build GRCh38
rs750839500 Chr.14: 64411155-64411155 on Build GRCh38
rs901159754 Chr.14: 64411182-64411182 on Build GRCh38
rs201428609 Chr.14: 64411136-64411136 on Build GRCh38
rs759027931 Chr.14: 64411074-64411074 on Build GRCh38
rs373024383 Chr.14: 64411072-64411072 on Build GRCh38
rs539712991 Chr.14: 64411151-64411152 on Build GRCh38
rs187912122 Chr.14: 64411254-64411254 on Build GRCh38
rs747933171 Chr.14: 64411115-64411116 on Build GRCh38
rs751623217 Chr.14: 64411164-64411164 on Build GRCh38
rs938896026 Chr.14: 64411023-64411023 on Build GRCh38
rs769407733 Chr.14: 64411060-64411060 on Build GRCh38
rs1022016042 Chr.14: 64410987-64410987 on Build GRCh38
rs375760111 Chr.14: 64411040-64411040 on Build GRCh38
rs894496998 Chr.14: 64411210-64411210 on Build GRCh38
rs781120932 Chr.14: 64411048-64411048 on Build GRCh38
rs771627697 Chr.14: 64411095-64411095 on Build GRCh38
rs977745867 Chr.14: 64410992-64410992 on Build GRCh38
rs987378565 Chr.14: 64411347-64411347 on Build GRCh38
rs528618090 Chr.14: 64410989-64410989 on Build GRCh38
rs377397939 Chr.14: 64411098-64411098 on Build GRCh38
rs552514537 Chr.14: 64411325-64411325 on Build GRCh38
rs777380840 Chr.14: 64411176-64411176 on Build GRCh38
rs748047276 Chr.14: 64411205-64411205 on Build GRCh38
rs989644757 Chr.14: 64410986-64410986 on Build GRCh38
rs766656603 Chr.14: 64411160-64411160 on Build GRCh38
rs978209842 Chr.14: 64411034-64411034 on Build GRCh38
rs923745898 Chr.14: 64411036-64411036 on Build GRCh38
rs370120826 Chr.14: 64411063-64411063 on Build GRCh38
rs957785508 Chr.14: 64411292-64411292 on Build GRCh38
rs199580141 Chr.14: 64411129-64411129 on Build GRCh38
rs756962415 Chr.14: 64411052-64411052 on Build GRCh38
rs1022088336 Chr.14: 64411242-64411242 on Build GRCh38
rs539465811 Chr.14: 64411201-64411201 on Build GRCh38
rs373682324 Chr.14: 64411154-64411154 on Build GRCh38
rs201222323 Chr.14: 64411133-64411133 on Build GRCh38
rs370444838 Chr.14: 64411109-64411109 on Build GRCh38
rs777688362 Chr.14: 64411196-64411196 on Build GRCh38
rs184760932 Chr.14: 64411251-64411251 on Build GRCh38
rs1036852797 Chr.14: 64411181-64411181 on Build GRCh38
rs936318324 Chr.14: 64411038-64411038 on Build GRCh38
rs899269680 Chr.14: 64411197-64411204 on Build GRCh38
rs147438541 Chr.14: 64411231-64411231 on Build GRCh38
rs977643365 Chr.14: 64411287-64411288 on Build GRCh38
rs567786659 Chr.14: 64411044-64411044 on Build GRCh38
rs989655997 Chr.14: 64411302-64411302 on Build GRCh38
rs966407580 Chr.14: 64410988-64410988 on Build GRCh38
rs192490131 Chr.14: 64411067-64411067 on Build GRCh38
rs781048041 Chr.14: 64411054-64411054 on Build GRCh38
rs754952437 Chr.14: 64411167-64411167 on Build GRCh38
rs752532529 Chr.14: 64411178-64411178 on Build GRCh38

View Full Product Details