Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 68789705-68790193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFP36L1
Entrez Gene ID
677
Gene Name
ZFP36 ring finger protein like 1
Gene Aliases
BRF1, Berg36, ERF-1, ERF1, RNF162B, TIS11B, cMG1
UniGene
Hs.707091
Gene Chromosome Location
Chr.14: 68787655-68796243 on Build GRCh38
Transcripts
NM_001244698.1 NM_001244701.1 NM_004926.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5346698 c.411C>A p.F137L Chr.14: 68790139-68790139 on Build GRCh38
7080196 c.386G>C p.C129S Chr.14: 68790164-68790164 on Build GRCh38
416629 c.556G>A p.A186T Chr.14: 68789994-68789994 on Build GRCh38
1745537 c.453_454insC p.K152fs*29 Chr.14: 68790096-68790097 on Build GRCh38
433281 c.526_528delATC p.I176delI Chr.14: 68790022-68790024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049829110 Chr.14: 68789920-68789920 on Build GRCh38
rs369501004 Chr.14: 68790088-68790088 on Build GRCh38
rs776253509 Chr.14: 68790072-68790072 on Build GRCh38
rs763608011 Chr.14: 68789804-68789804 on Build GRCh38
rs780614338 Chr.14: 68789972-68789972 on Build GRCh38
rs866482568 Chr.14: 68790003-68790003 on Build GRCh38
rs192307797 Chr.14: 68789819-68789819 on Build GRCh38
rs776814611 Chr.14: 68789791-68789791 on Build GRCh38
rs747171551 Chr.14: 68789761-68789761 on Build GRCh38
rs758395092 Chr.14: 68789749-68789749 on Build GRCh38
rs202040968 Chr.14: 68789746-68789746 on Build GRCh38
rs549262013 Chr.14: 68789818-68789818 on Build GRCh38
rs758405640 Chr.14: 68789887-68789887 on Build GRCh38
rs766076255 Chr.14: 68789838-68789838 on Build GRCh38
rs749727355 Chr.14: 68789988-68789988 on Build GRCh38
rs764990481 Chr.14: 68790100-68790100 on Build GRCh38
rs770415552 Chr.14: 68790070-68790070 on Build GRCh38
rs1008332001 Chr.14: 68789926-68789926 on Build GRCh38
rs746245985 Chr.14: 68789794-68789794 on Build GRCh38
rs867766935 Chr.14: 68789882-68789882 on Build GRCh38
rs150284721 Chr.14: 68790169-68790169 on Build GRCh38
rs928821903 Chr.14: 68789822-68789822 on Build GRCh38
rs772546250 Chr.14: 68790006-68790006 on Build GRCh38
rs201731801 Chr.14: 68789974-68789974 on Build GRCh38
rs898495175 Chr.14: 68789911-68789911 on Build GRCh38
rs889842333 Chr.14: 68789921-68789921 on Build GRCh38
rs770355047 Chr.14: 68789799-68789799 on Build GRCh38
rs753564768 Chr.14: 68789842-68789842 on Build GRCh38
rs747138115 Chr.14: 68789899-68789899 on Build GRCh38
rs567307143 Chr.14: 68789901-68789901 on Build GRCh38
rs776133275 Chr.14: 68789801-68789801 on Build GRCh38
rs1008315301 Chr.14: 68790040-68790040 on Build GRCh38
rs781476840 Chr.14: 68790151-68790151 on Build GRCh38
rs762753405 Chr.14: 68790115-68790115 on Build GRCh38
rs777962695 Chr.14: 68789755-68789755 on Build GRCh38
rs746155673 Chr.14: 68789966-68789966 on Build GRCh38
rs755839037 Chr.14: 68789742-68789742 on Build GRCh38
rs199510843 Chr.14: 68790107-68790107 on Build GRCh38
rs766913691 Chr.14: 68789813-68789813 on Build GRCh38
rs750342662 Chr.14: 68789851-68789851 on Build GRCh38
rs757350812 Chr.14: 68790133-68790133 on Build GRCh38
rs765865743 Chr.14: 68789724-68789724 on Build GRCh38
rs756444666 Chr.14: 68790160-68790160 on Build GRCh38
rs770380051 Chr.14: 68789778-68789778 on Build GRCh38
rs760791768 Chr.14: 68789898-68789909 on Build GRCh38
rs547380298 Chr.14: 68789729-68789729 on Build GRCh38
rs750030944 Chr.14: 68789739-68789739 on Build GRCh38
rs760239208 Chr.14: 68790043-68790043 on Build GRCh38
rs769235506 Chr.14: 68789991-68789991 on Build GRCh38
rs773937725 Chr.14: 68789728-68789728 on Build GRCh38
rs750648602 Chr.14: 68790157-68790157 on Build GRCh38
rs765872866 Chr.14: 68790019-68790019 on Build GRCh38
rs948509662 Chr.14: 68789735-68789735 on Build GRCh38
rs764108567 Chr.14: 68790121-68790121 on Build GRCh38
rs754764949 Chr.14: 68789848-68789848 on Build GRCh38
rs757366032 Chr.14: 68789914-68789914 on Build GRCh38
rs775692081 Chr.14: 68789806-68789807 on Build GRCh38
rs765076175 Chr.14: 68789849-68789849 on Build GRCh38
rs755097253 Chr.14: 68789869-68789869 on Build GRCh38
rs1011551304 Chr.14: 68789908-68789908 on Build GRCh38
rs754854659 Chr.14: 68789743-68789743 on Build GRCh38
rs537181795 Chr.14: 68789971-68789971 on Build GRCh38
rs752523276 Chr.14: 68790108-68790108 on Build GRCh38
rs760203539 Chr.14: 68789832-68789832 on Build GRCh38
rs751523830 Chr.14: 68790129-68790129 on Build GRCh38
rs569750320 Chr.14: 68789938-68789938 on Build GRCh38
rs1037318578 Chr.14: 68789896-68789896 on Build GRCh38

View Full Product Details