Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 69209493-69209995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXD2
Entrez Gene ID
55218
Gene Name
exonuclease 3'-5' domain containing 2
Gene Aliases
C14orf114, EXDL2
UniGene
Hs.533878
Gene Chromosome Location
Chr.14: 69191477-69244020 on Build GRCh38
Transcripts
NM_001193360.1 NM_001193361.1 NM_001193362.1 NM_001193363.1 NM_018199.3 NR_034164.1 NR_034165.1 XM_011536908.2 XM_017021421.1

Gene Symbol
LOC105370550
Entrez Gene ID
105370550
Gene Name
uncharacterized LOC105370550
Gene Aliases
-
UniGene
Hs.667208
Gene Chromosome Location
Chr.14: 69183018-69260567 on Build GRCh38
Transcripts
NM_001193360.1 NM_001193361.1 NM_001193362.1 NM_001193363.1 NM_018199.3 NR_034164.1 NR_034165.1 XM_011536908.2 XM_017021421.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6194857 c.318A>C p.G106G Chr.14: 69209788-69209788 on Build GRCh38
239753 c.125A>G p.Q42R Chr.14: 69209595-69209595 on Build GRCh38
6333400 c.261G>T p.E87D Chr.14: 69209731-69209731 on Build GRCh38
5522611 c.301G>A p.D101N Chr.14: 69209771-69209771 on Build GRCh38
5740148 c.113C>A p.P38H Chr.14: 69209583-69209583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977885505 Chr.14: 69209882-69209882 on Build GRCh38
rs143159826 Chr.14: 69209843-69209843 on Build GRCh38
rs760048337 Chr.14: 69209569-69209569 on Build GRCh38
rs114632394 Chr.14: 69209893-69209893 on Build GRCh38
rs973104436 Chr.14: 69209878-69209878 on Build GRCh38
rs757533207 Chr.14: 69209825-69209827 on Build GRCh38
rs1032568684 Chr.14: 69209825-69209832 on Build GRCh38
rs763872672 Chr.14: 69209640-69209640 on Build GRCh38
rs770418703 Chr.14: 69209577-69209577 on Build GRCh38
rs779199657 Chr.14: 69209777-69209777 on Build GRCh38
rs1004874222 Chr.14: 69209676-69209676 on Build GRCh38
rs199733370 Chr.14: 69209825-69209825 on Build GRCh38
rs766419235 Chr.14: 69209658-69209658 on Build GRCh38
rs772603010 Chr.14: 69209809-69209809 on Build GRCh38
rs752980100 Chr.14: 69209822-69209823 on Build GRCh38
rs537950944 Chr.14: 69209669-69209669 on Build GRCh38
rs895674480 Chr.14: 69209715-69209715 on Build GRCh38
rs141827613 Chr.14: 69209823-69209823 on Build GRCh38
rs778963511 Chr.14: 69209845-69209846 on Build GRCh38
rs1014072150 Chr.14: 69209721-69209721 on Build GRCh38
rs763017009 Chr.14: 69209771-69209771 on Build GRCh38
rs757322538 Chr.14: 69209659-69209659 on Build GRCh38
rs570508901 Chr.14: 69209579-69209579 on Build GRCh38
rs118014825 Chr.14: 69209874-69209874 on Build GRCh38
rs774929784 Chr.14: 69209564-69209564 on Build GRCh38
rs573129686 Chr.14: 69209924-69209924 on Build GRCh38
rs752389207 Chr.14: 69209598-69209598 on Build GRCh38
rs987626120 Chr.14: 69209842-69209843 on Build GRCh38
rs750579457 Chr.14: 69209688-69209688 on Build GRCh38
rs769075965 Chr.14: 69209749-69209749 on Build GRCh38
rs764922126 Chr.14: 69209590-69209590 on Build GRCh38
rs776101197 Chr.14: 69209816-69209816 on Build GRCh38
rs751529492 Chr.14: 69209656-69209656 on Build GRCh38
rs749547416 Chr.14: 69209747-69209747 on Build GRCh38
rs918261119 Chr.14: 69209843-69209843 on Build GRCh38
rs940408789 Chr.14: 69209581-69209581 on Build GRCh38
rs902507379 Chr.14: 69209790-69209790 on Build GRCh38
rs775215929 Chr.14: 69209837-69209837 on Build GRCh38
rs748680924 Chr.14: 69209808-69209808 on Build GRCh38
rs954892447 Chr.14: 69209839-69209839 on Build GRCh38
rs1016650883 Chr.14: 69209705-69209705 on Build GRCh38
rs912299612 Chr.14: 69209860-69209860 on Build GRCh38
rs953856649 Chr.14: 69209840-69209840 on Build GRCh38
rs769400238 Chr.14: 69209836-69209836 on Build GRCh38
rs958297225 Chr.14: 69209825-69209825 on Build GRCh38
rs917071549 Chr.14: 69209957-69209957 on Build GRCh38
rs564921902 Chr.14: 69209956-69209956 on Build GRCh38
rs556197203 Chr.14: 69209677-69209677 on Build GRCh38
rs761048890 Chr.14: 69209825-69209826 on Build GRCh38
rs182466488 Chr.14: 69209562-69209562 on Build GRCh38
rs756354734 Chr.14: 69209743-69209743 on Build GRCh38
rs949171963 Chr.14: 69209817-69209817 on Build GRCh38
rs776194325 Chr.14: 69209578-69209578 on Build GRCh38
rs1028277326 Chr.14: 69209762-69209762 on Build GRCh38
rs762841088 Chr.14: 69209636-69209636 on Build GRCh38
rs1013743084 Chr.14: 69209565-69209565 on Build GRCh38
rs540149225 Chr.14: 69209955-69209955 on Build GRCh38
rs75083088 Chr.14: 69209744-69209744 on Build GRCh38
rs770207941 Chr.14: 69209561-69209561 on Build GRCh38
rs963430013 Chr.14: 69209713-69209713 on Build GRCh38
rs754003799 Chr.14: 69209824-69209825 on Build GRCh38
rs745408047 Chr.14: 69209822-69209822 on Build GRCh38
rs533779294 Chr.14: 69209838-69209838 on Build GRCh38

View Full Product Details