Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 69328366-69328866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT16
Entrez Gene ID
57452
Gene Name
polypeptide N-acetylgalactosaminyltransferase 16
Gene Aliases
GALNACT16, GALNTL1, GalNAc-T16
UniGene
Hs.21035
Gene Chromosome Location
Chr.14: 69259546-69379904 on Build GRCh38
Transcripts
NM_001168368.1 NM_020692.2 XM_011537005.1 XM_011537006.2 XM_011537007.2 XM_017021498.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6669064 c.615C>T p.T205T Chr.14: 69328496-69328496 on Build GRCh38
4686325 c.597G>A p.A199A Chr.14: 69328478-69328478 on Build GRCh38
3497367 c.688G>A p.E230K Chr.14: 69328569-69328569 on Build GRCh38
5828154 c.643G>A p.E215K Chr.14: 69328524-69328524 on Build GRCh38
3497363 c.594G>A p.G198G Chr.14: 69328475-69328475 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765779123 Chr.14: 69328399-69328399 on Build GRCh38
rs200438165 Chr.14: 69328619-69328619 on Build GRCh38
rs147516752 Chr.14: 69328497-69328497 on Build GRCh38
rs893139522 Chr.14: 69328738-69328738 on Build GRCh38
rs75562906 Chr.14: 69328622-69328622 on Build GRCh38
rs8004934 Chr.14: 69328808-69328808 on Build GRCh38
rs372330048 Chr.14: 69328524-69328524 on Build GRCh38
rs890163789 Chr.14: 69328694-69328694 on Build GRCh38
rs560466824 Chr.14: 69328477-69328477 on Build GRCh38
rs769322189 Chr.14: 69328544-69328544 on Build GRCh38
rs147334035 Chr.14: 69328472-69328472 on Build GRCh38
rs766050730 Chr.14: 69328547-69328547 on Build GRCh38
rs750975590 Chr.14: 69328406-69328406 on Build GRCh38
rs61748871 Chr.14: 69328471-69328471 on Build GRCh38
rs1050580196 Chr.14: 69328652-69328652 on Build GRCh38
rs998838509 Chr.14: 69328465-69328465 on Build GRCh38
rs373893534 Chr.14: 69328430-69328430 on Build GRCh38
rs183196628 Chr.14: 69328816-69328816 on Build GRCh38
rs752464899 Chr.14: 69328561-69328561 on Build GRCh38
rs931182073 Chr.14: 69328628-69328628 on Build GRCh38
rs894618836 Chr.14: 69328812-69328812 on Build GRCh38
rs199936468 Chr.14: 69328452-69328452 on Build GRCh38
rs767116865 Chr.14: 69328478-69328478 on Build GRCh38
rs764978774 Chr.14: 69328505-69328505 on Build GRCh38
rs551513124 Chr.14: 69328773-69328773 on Build GRCh38
rs752655414 Chr.14: 69328520-69328520 on Build GRCh38
rs775769291 Chr.14: 69328539-69328539 on Build GRCh38
rs12879377 Chr.14: 69328482-69328482 on Build GRCh38
rs1011694448 Chr.14: 69328564-69328564 on Build GRCh38
rs778892705 Chr.14: 69328442-69328442 on Build GRCh38
rs954173509 Chr.14: 69328792-69328792 on Build GRCh38
rs961155807 Chr.14: 69328819-69328819 on Build GRCh38
rs1000381767 Chr.14: 69328725-69328725 on Build GRCh38
rs190621989 Chr.14: 69328634-69328634 on Build GRCh38
rs1033151175 Chr.14: 69328751-69328751 on Build GRCh38
rs907755818 Chr.14: 69328415-69328415 on Build GRCh38
rs776961764 Chr.14: 69328596-69328596 on Build GRCh38
rs972511941 Chr.14: 69328829-69328829 on Build GRCh38
rs144789007 Chr.14: 69328496-69328496 on Build GRCh38
rs750215619 Chr.14: 69328775-69328775 on Build GRCh38
rs767204698 Chr.14: 69328418-69328418 on Build GRCh38
rs919849195 Chr.14: 69328596-69328596 on Build GRCh38
rs757325902 Chr.14: 69328587-69328587 on Build GRCh38
rs971370392 Chr.14: 69328546-69328546 on Build GRCh38
rs998804388 Chr.14: 69328707-69328707 on Build GRCh38
rs765030633 Chr.14: 69328560-69328560 on Build GRCh38
rs771173266 Chr.14: 69328463-69328463 on Build GRCh38
rs531337764 Chr.14: 69328535-69328535 on Build GRCh38
rs972426785 Chr.14: 69328593-69328593 on Build GRCh38
rs149859357 Chr.14: 69328579-69328579 on Build GRCh38
rs776793934 Chr.14: 69328467-69328467 on Build GRCh38
rs764032316 Chr.14: 69328569-69328569 on Build GRCh38
rs747548379 Chr.14: 69328403-69328403 on Build GRCh38
rs547543538 Chr.14: 69328624-69328624 on Build GRCh38
rs200530972 Chr.14: 69328562-69328562 on Build GRCh38
rs61746597 Chr.14: 69328553-69328553 on Build GRCh38
rs749767663 Chr.14: 69328541-69328541 on Build GRCh38
rs750072404 Chr.14: 69328422-69328422 on Build GRCh38
rs752091675 Chr.14: 69328420-69328420 on Build GRCh38
rs576508972 Chr.14: 69328394-69328394 on Build GRCh38
rs201138904 Chr.14: 69328551-69328551 on Build GRCh38
rs1012209578 Chr.14: 69328770-69328770 on Build GRCh38
rs766138593 Chr.14: 69328495-69328495 on Build GRCh38
rs758352543 Chr.14: 69328523-69328523 on Build GRCh38
rs779949674 Chr.14: 69328429-69328429 on Build GRCh38
rs756314914 Chr.14: 69328620-69328620 on Build GRCh38
rs770164612 Chr.14: 69328538-69328538 on Build GRCh38
rs780429319 Chr.14: 69328623-69328623 on Build GRCh38
rs1041999234 Chr.14: 69328712-69328712 on Build GRCh38
rs991910123 Chr.14: 69328536-69328536 on Build GRCh38
rs1023055156 Chr.14: 69328573-69328573 on Build GRCh38
rs779815207 Chr.14: 69328672-69328672 on Build GRCh38
rs747094920 Chr.14: 69328528-69328528 on Build GRCh38
rs903394339 Chr.14: 69328715-69328715 on Build GRCh38
rs944346342 Chr.14: 69328700-69328700 on Build GRCh38
rs757413840 Chr.14: 69328534-69328534 on Build GRCh38
rs140463814 Chr.14: 69328526-69328527 on Build GRCh38
rs572293760 Chr.14: 69328481-69328481 on Build GRCh38
rs931780124 Chr.14: 69328635-69328635 on Build GRCh38
rs79469907 Chr.14: 69328545-69328545 on Build GRCh38
rs371722697 Chr.14: 69328702-69328702 on Build GRCh38
rs781171324 Chr.14: 69328600-69328600 on Build GRCh38
rs747182949 Chr.14: 69328458-69328458 on Build GRCh38
rs1045776560 Chr.14: 69328667-69328667 on Build GRCh38
rs769195978 Chr.14: 69328436-69328437 on Build GRCh38
rs761375542 Chr.14: 69328410-69328410 on Build GRCh38
rs907328798 Chr.14: 69328667-69328667 on Build GRCh38
rs759959307 Chr.14: 69328470-69328470 on Build GRCh38
rs139971588 Chr.14: 69328445-69328445 on Build GRCh38
rs370181273 Chr.14: 69328423-69328423 on Build GRCh38
rs754810423 Chr.14: 69328441-69328441 on Build GRCh38
rs12100911 Chr.14: 69328451-69328451 on Build GRCh38
rs368466926 Chr.14: 69328640-69328640 on Build GRCh38

View Full Product Details