Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 70325275-70325820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX16
Entrez Gene ID
51241
Gene Name
COX16, cytochrome c oxidase assembly homolog
Gene Aliases
C14orf112, HSPC203
UniGene
Hs.709581
Gene Chromosome Location
Chr.14: 70325081-70359731 on Build GRCh38
Transcripts
NM_001204090.1 NM_016468.6 NM_001202547.2 NM_001202548.2 NM_001202549.2

Gene Symbol
SYNJ2BP-COX16
Entrez Gene ID
100529257
Gene Name
SYNJ2BP-COX16 readthrough
Gene Aliases
-
UniGene
Hs.443661
Gene Chromosome Location
Chr.14: 70325081-70417090 on Build GRCh38
Transcripts
NM_001204090.1 NM_016468.6 NM_001202547.2 NM_001202548.2 NM_001202549.2

SNP Detail

SNP ID Chromosome Location
rs897247221 Chr.14: 70325570-70325570 on Build GRCh38
rs888322583 Chr.14: 70325604-70325604 on Build GRCh38
rs57037789 Chr.14: 70325503-70325503 on Build GRCh38
rs954240770 Chr.14: 70325721-70325723 on Build GRCh38
rs534426374 Chr.14: 70325750-70325754 on Build GRCh38
rs59033684 Chr.14: 70325557-70325557 on Build GRCh38
rs576322224 Chr.14: 70325535-70325535 on Build GRCh38
rs895534844 Chr.14: 70325615-70325615 on Build GRCh38
rs141342347 Chr.14: 70325708-70325708 on Build GRCh38
rs997820174 Chr.14: 70325678-70325678 on Build GRCh38
rs1013147982 Chr.14: 70325400-70325400 on Build GRCh38
rs528321754 Chr.14: 70325610-70325610 on Build GRCh38
rs574842787 Chr.14: 70325702-70325702 on Build GRCh38
rs968897242 Chr.14: 70325756-70325756 on Build GRCh38
rs530293644 Chr.14: 70325348-70325348 on Build GRCh38
rs1042663082 Chr.14: 70325558-70325558 on Build GRCh38
rs927259733 Chr.14: 70325563-70325563 on Build GRCh38
rs141862353 Chr.14: 70325681-70325687 on Build GRCh38
rs934311095 Chr.14: 70325579-70325583 on Build GRCh38
rs896972034 Chr.14: 70325310-70325310 on Build GRCh38
rs980334680 Chr.14: 70325757-70325757 on Build GRCh38
rs762929354 Chr.14: 70325357-70325357 on Build GRCh38
rs766634880 Chr.14: 70325636-70325636 on Build GRCh38
rs924172462 Chr.14: 70325786-70325787 on Build GRCh38
rs754042874 Chr.14: 70325723-70325723 on Build GRCh38
rs984012027 Chr.14: 70325423-70325423 on Build GRCh38
rs111806712 Chr.14: 70325730-70325730 on Build GRCh38
rs965001619 Chr.14: 70325654-70325654 on Build GRCh38
rs989277868 Chr.14: 70325721-70325721 on Build GRCh38
rs961673939 Chr.14: 70325436-70325436 on Build GRCh38
rs1044175488 Chr.14: 70325646-70325647 on Build GRCh38
rs932775319 Chr.14: 70325532-70325532 on Build GRCh38
rs774235692 Chr.14: 70325504-70325504 on Build GRCh38
rs1048713323 Chr.14: 70325589-70325589 on Build GRCh38
rs181434008 Chr.14: 70325422-70325422 on Build GRCh38
rs947609447 Chr.14: 70325545-70325545 on Build GRCh38
rs989388069 Chr.14: 70325542-70325542 on Build GRCh38
rs888283163 Chr.14: 70325669-70325669 on Build GRCh38
rs914795548 Chr.14: 70325543-70325543 on Build GRCh38
rs189837760 Chr.14: 70325416-70325416 on Build GRCh38
rs915370874 Chr.14: 70325749-70325751 on Build GRCh38
rs993978464 Chr.14: 70325571-70325571 on Build GRCh38
rs956205039 Chr.14: 70325534-70325534 on Build GRCh38
rs9788528 Chr.14: 70325448-70325448 on Build GRCh38
rs978287270 Chr.14: 70325782-70325782 on Build GRCh38
rs770828385 Chr.14: 70325754-70325755 on Build GRCh38
rs548516943 Chr.14: 70325533-70325533 on Build GRCh38
rs965055927 Chr.14: 70325495-70325495 on Build GRCh38
rs1015511260 Chr.14: 70325432-70325432 on Build GRCh38
rs572046431 Chr.14: 70325397-70325397 on Build GRCh38
rs952743784 Chr.14: 70325401-70325401 on Build GRCh38
rs1017890022 Chr.14: 70325485-70325485 on Build GRCh38
rs1018170952 Chr.14: 70325643-70325643 on Build GRCh38
rs1029681299 Chr.14: 70325713-70325713 on Build GRCh38
rs562877126 Chr.14: 70325308-70325308 on Build GRCh38
rs759320655 Chr.14: 70325780-70325782 on Build GRCh38
rs199886061 Chr.14: 70325680-70325680 on Build GRCh38
rs938691389 Chr.14: 70325566-70325566 on Build GRCh38
rs555701658 Chr.14: 70325567-70325567 on Build GRCh38
rs1035842961 Chr.14: 70325313-70325313 on Build GRCh38

View Full Product Details