Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73276209-73276707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUMB
Entrez Gene ID
8650
Gene Name
NUMB, endocytic adaptor protein
Gene Aliases
C14orf41, S171, c14_5527
UniGene
Hs.525443
Gene Chromosome Location
Chr.14: 73275210-73458580 on Build GRCh38
Transcripts
NM_001005743.1 NM_001005744.1 NM_001005745.1 NM_001320114.1 NM_003744.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1370956 c.1938G>A p.T646T Chr.14: 73276596-73276596 on Build GRCh38
4052271 c.1857G>A p.Q619Q Chr.14: 73276677-73276677 on Build GRCh38
2137069 c.1889G>A p.R630H Chr.14: 73276645-73276645 on Build GRCh38
1477747 c.1876A>C p.K626Q Chr.14: 73276658-73276658 on Build GRCh38
957633 c.1917C>T p.F639F Chr.14: 73276617-73276617 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535477285 Chr.14: 73276267-73276267 on Build GRCh38
rs769689392 Chr.14: 73276684-73276684 on Build GRCh38
rs1006751430 Chr.14: 73276308-73276308 on Build GRCh38
rs751764764 Chr.14: 73276352-73276352 on Build GRCh38
rs902178355 Chr.14: 73276254-73276254 on Build GRCh38
rs11625196 Chr.14: 73276497-73276497 on Build GRCh38
rs567649839 Chr.14: 73276394-73276394 on Build GRCh38
rs533363786 Chr.14: 73276680-73276680 on Build GRCh38
rs553303989 Chr.14: 73276466-73276466 on Build GRCh38
rs746722095 Chr.14: 73276572-73276572 on Build GRCh38
rs200927213 Chr.14: 73276674-73276674 on Build GRCh38
rs758548890 Chr.14: 73276529-73276529 on Build GRCh38
rs367917511 Chr.14: 73276600-73276600 on Build GRCh38
rs1000362942 Chr.14: 73276555-73276555 on Build GRCh38
rs756600948 Chr.14: 73276549-73276549 on Build GRCh38
rs73306841 Chr.14: 73276480-73276480 on Build GRCh38
rs145338656 Chr.14: 73276673-73276673 on Build GRCh38
rs762037345 Chr.14: 73276614-73276614 on Build GRCh38
rs757467050 Chr.14: 73276533-73276533 on Build GRCh38
rs981375049 Chr.14: 73276358-73276360 on Build GRCh38
rs781543237 Chr.14: 73276543-73276543 on Build GRCh38
rs1007459035 Chr.14: 73276510-73276510 on Build GRCh38
rs942146645 Chr.14: 73276381-73276381 on Build GRCh38
rs749857685 Chr.14: 73276617-73276618 on Build GRCh38
rs781188773 Chr.14: 73276424-73276424 on Build GRCh38
rs549384821 Chr.14: 73276468-73276468 on Build GRCh38
rs547663063 Chr.14: 73276235-73276235 on Build GRCh38
rs549364703 Chr.14: 73276360-73276360 on Build GRCh38
rs374579310 Chr.14: 73276645-73276645 on Build GRCh38
rs775145232 Chr.14: 73276685-73276685 on Build GRCh38
rs772714277 Chr.14: 73276427-73276427 on Build GRCh38
rs776594669 Chr.14: 73276575-73276575 on Build GRCh38
rs933886642 Chr.14: 73276259-73276259 on Build GRCh38
rs759340298 Chr.14: 73276595-73276595 on Build GRCh38
rs35625053 Chr.14: 73276608-73276608 on Build GRCh38
rs149085247 Chr.14: 73276265-73276265 on Build GRCh38
rs541552944 Chr.14: 73276531-73276531 on Build GRCh38
rs773558195 Chr.14: 73276603-73276604 on Build GRCh38
rs959516024 Chr.14: 73276491-73276491 on Build GRCh38
rs368343455 Chr.14: 73276613-73276613 on Build GRCh38
rs770743201 Chr.14: 73276670-73276670 on Build GRCh38
rs750782987 Chr.14: 73276640-73276640 on Build GRCh38
rs769409332 Chr.14: 73276570-73276571 on Build GRCh38
rs757332138 Chr.14: 73276373-73276373 on Build GRCh38
rs177381 Chr.14: 73276659-73276659 on Build GRCh38
rs754423641 Chr.14: 73276654-73276654 on Build GRCh38
rs745873728 Chr.14: 73276433-73276433 on Build GRCh38
rs772832892 Chr.14: 73276567-73276567 on Build GRCh38
rs759982467 Chr.14: 73276673-73276673 on Build GRCh38
rs771585707 Chr.14: 73276654-73276655 on Build GRCh38
rs767676611 Chr.14: 73276630-73276630 on Build GRCh38
rs761443820 Chr.14: 73276238-73276239 on Build GRCh38
rs889599680 Chr.14: 73276268-73276268 on Build GRCh38
rs147475968 Chr.14: 73276646-73276646 on Build GRCh38
rs755532608 Chr.14: 73276657-73276657 on Build GRCh38
rs949590919 Chr.14: 73276311-73276311 on Build GRCh38
rs780907508 Chr.14: 73276532-73276533 on Build GRCh38
rs45624841 Chr.14: 73276528-73276528 on Build GRCh38
rs551591893 Chr.14: 73276271-73276271 on Build GRCh38
rs764139002 Chr.14: 73276530-73276530 on Build GRCh38
rs534948337 Chr.14: 73276441-73276441 on Build GRCh38
rs770637219 Chr.14: 73276574-73276574 on Build GRCh38
rs143147360 Chr.14: 73276524-73276524 on Build GRCh38
rs995244457 Chr.14: 73276372-73276372 on Build GRCh38
rs537074549 Chr.14: 73276341-73276341 on Build GRCh38
rs747594632 Chr.14: 73276553-73276553 on Build GRCh38
rs771754970 Chr.14: 73276562-73276562 on Build GRCh38
rs570086899 Chr.14: 73276303-73276303 on Build GRCh38
rs117957039 Chr.14: 73276596-73276596 on Build GRCh38
rs369532050 Chr.14: 73276558-73276558 on Build GRCh38
rs372727231 Chr.14: 73276457-73276457 on Build GRCh38
rs1042092529 Chr.14: 73276253-73276253 on Build GRCh38
rs897836161 Chr.14: 73276452-73276452 on Build GRCh38
rs779949869 Chr.14: 73276470-73276470 on Build GRCh38
rs746185165 Chr.14: 73276545-73276545 on Build GRCh38
rs1050480 Chr.14: 73276503-73276503 on Build GRCh38
rs780568361 Chr.14: 73276553-73276553 on Build GRCh38
rs774814763 Chr.14: 73276660-73276661 on Build GRCh38
rs1017688052 Chr.14: 73276325-73276325 on Build GRCh38
rs375897614 Chr.14: 73276662-73276662 on Build GRCh38
rs371320723 Chr.14: 73276660-73276660 on Build GRCh38
rs762788053 Chr.14: 73276601-73276601 on Build GRCh38

View Full Product Details