Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73697843-73698393 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAL1
Entrez Gene ID
83544
Gene Name
dynein axonemal light chain 1
Gene Aliases
C14orf168, CILD16
UniGene
Hs.271270
Gene Chromosome Location
Chr.14: 73644875-73703728 on Build GRCh38
Transcripts
NM_001201366.1 NM_031427.3

SNP Detail

SNP ID Chromosome Location
rs200947750 Chr.14: 73697899-73697899 on Build GRCh38
rs1043844742 Chr.14: 73698116-73698116 on Build GRCh38
rs560647853 Chr.14: 73698337-73698337 on Build GRCh38
rs1017893551 Chr.14: 73698301-73698301 on Build GRCh38
rs376454067 Chr.14: 73698276-73698276 on Build GRCh38
rs760581796 Chr.14: 73697986-73697986 on Build GRCh38
rs115026744 Chr.14: 73698036-73698036 on Build GRCh38
rs1030845019 Chr.14: 73697910-73697910 on Build GRCh38
rs886050698 Chr.14: 73697963-73697965 on Build GRCh38
rs541092674 Chr.14: 73698047-73698047 on Build GRCh38
rs1008136200 Chr.14: 73698296-73698296 on Build GRCh38
rs908075309 Chr.14: 73698356-73698356 on Build GRCh38
rs867930153 Chr.14: 73698064-73698064 on Build GRCh38
rs955312370 Chr.14: 73697915-73697915 on Build GRCh38
rs7152537 Chr.14: 73698055-73698055 on Build GRCh38
rs903947308 Chr.14: 73698124-73698124 on Build GRCh38
rs944955694 Chr.14: 73698347-73698347 on Build GRCh38
rs577696114 Chr.14: 73698066-73698066 on Build GRCh38
rs984039953 Chr.14: 73698339-73698339 on Build GRCh38
rs886050697 Chr.14: 73697944-73697944 on Build GRCh38
rs893538516 Chr.14: 73698149-73698149 on Build GRCh38
rs189469446 Chr.14: 73697919-73697919 on Build GRCh38
rs974217601 Chr.14: 73698325-73698325 on Build GRCh38
rs951207222 Chr.14: 73698341-73698341 on Build GRCh38
rs17182488 Chr.14: 73698015-73698015 on Build GRCh38
rs1009842504 Chr.14: 73698162-73698162 on Build GRCh38
rs531036854 Chr.14: 73698192-73698192 on Build GRCh38
rs973889938 Chr.14: 73697989-73697989 on Build GRCh38
rs1047873647 Chr.14: 73698025-73698025 on Build GRCh38
rs758786439 Chr.14: 73698349-73698349 on Build GRCh38
rs973401690 Chr.14: 73698297-73698297 on Build GRCh38
rs192948772 Chr.14: 73698006-73698006 on Build GRCh38
rs1021314156 Chr.14: 73697918-73697918 on Build GRCh38
rs765460583 Chr.14: 73698220-73698220 on Build GRCh38
rs563516136 Chr.14: 73698273-73698273 on Build GRCh38
rs530268865 Chr.14: 73698011-73698011 on Build GRCh38
rs562566868 Chr.14: 73697959-73697961 on Build GRCh38
rs999834743 Chr.14: 73697901-73697901 on Build GRCh38
rs948114795 Chr.14: 73698108-73698108 on Build GRCh38
rs557158339 Chr.14: 73697940-73697940 on Build GRCh38
rs963770573 Chr.14: 73698311-73698311 on Build GRCh38
rs183492735 Chr.14: 73698275-73698275 on Build GRCh38
rs926620072 Chr.14: 73697944-73697944 on Build GRCh38
rs142744279 Chr.14: 73698178-73698178 on Build GRCh38
rs944949569 Chr.14: 73698051-73698051 on Build GRCh38
rs536587090 Chr.14: 73698014-73698014 on Build GRCh38
rs761957988 Chr.14: 73698062-73698062 on Build GRCh38
rs886050699 Chr.14: 73698341-73698341 on Build GRCh38
rs886050696 Chr.14: 73697910-73697910 on Build GRCh38

View Full Product Details