Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 74101013-74101426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIN52
Entrez Gene ID
91750
Gene Name
lin-52 DREAM MuvB core complex component
Gene Aliases
C14orf46, c14_5549
UniGene
Hs.612866
Gene Chromosome Location
Chr.14: 74084831-74201255 on Build GRCh38
Transcripts
NM_001024674.2 XM_011537320.2 XM_011537321.2 XM_011537322.2 XM_017021763.1 XM_017021764.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7149375 c.254G>A p.R85Q Chr.14: 74101197-74101197 on Build GRCh38
4757549 c.224T>G p.L75R Chr.14: 74101167-74101167 on Build GRCh38
1300820 c.276T>C p.Y92Y Chr.14: 74101219-74101219 on Build GRCh38
957732 c.289G>T p.D97Y Chr.14: 74101232-74101232 on Build GRCh38
7178917 c.225C>A p.L75L Chr.14: 74101168-74101168 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs963723107 Chr.14: 74101309-74101309 on Build GRCh38
rs745548148 Chr.14: 74101282-74101282 on Build GRCh38
rs1007883215 Chr.14: 74101293-74101293 on Build GRCh38
rs753999253 Chr.14: 74101264-74101264 on Build GRCh38
rs766274035 Chr.14: 74101270-74101308 on Build GRCh38
rs920883278 Chr.14: 74101330-74101330 on Build GRCh38
rs777197282 Chr.14: 74101168-74101168 on Build GRCh38
rs567630548 Chr.14: 74101103-74101103 on Build GRCh38
rs781344690 Chr.14: 74101361-74101361 on Build GRCh38
rs201061535 Chr.14: 74101173-74101173 on Build GRCh38
rs17096314 Chr.14: 74101249-74101249 on Build GRCh38
rs190542767 Chr.14: 74101279-74101279 on Build GRCh38
rs1017922648 Chr.14: 74101298-74101298 on Build GRCh38
rs907938841 Chr.14: 74101399-74101399 on Build GRCh38
rs754370048 Chr.14: 74101141-74101141 on Build GRCh38
rs2300193 Chr.14: 74101033-74101033 on Build GRCh38
rs200491155 Chr.14: 74101254-74101254 on Build GRCh38
rs974959517 Chr.14: 74101319-74101319 on Build GRCh38
rs761951918 Chr.14: 74101204-74101204 on Build GRCh38
rs186139881 Chr.14: 74101043-74101043 on Build GRCh38
rs760843850 Chr.14: 74101246-74101246 on Build GRCh38
rs114005617 Chr.14: 74101148-74101148 on Build GRCh38
rs920735523 Chr.14: 74101049-74101049 on Build GRCh38
rs774355295 Chr.14: 74101202-74101202 on Build GRCh38
rs371223670 Chr.14: 74101276-74101276 on Build GRCh38
rs148902734 Chr.14: 74101087-74101087 on Build GRCh38
rs952310862 Chr.14: 74101336-74101336 on Build GRCh38
rs767432549 Chr.14: 74101210-74101210 on Build GRCh38
rs752138169 Chr.14: 74101060-74101060 on Build GRCh38
rs930837160 Chr.14: 74101079-74101079 on Build GRCh38
rs375130778 Chr.14: 74101176-74101176 on Build GRCh38
rs931952220 Chr.14: 74101380-74101380 on Build GRCh38
rs746514417 Chr.14: 74101172-74101172 on Build GRCh38
rs780344218 Chr.14: 74101132-74101132 on Build GRCh38
rs755536243 Chr.14: 74101253-74101253 on Build GRCh38
rs756482684 Chr.14: 74101127-74101127 on Build GRCh38
rs753078024 Chr.14: 74101274-74101274 on Build GRCh38
rs111771040 Chr.14: 74101267-74101267 on Build GRCh38
rs145058704 Chr.14: 74101216-74101216 on Build GRCh38
rs369694210 Chr.14: 74101197-74101197 on Build GRCh38
rs763025504 Chr.14: 74101196-74101196 on Build GRCh38
rs750822197 Chr.14: 74101105-74101105 on Build GRCh38
rs916462197 Chr.14: 74101250-74101250 on Build GRCh38
rs940809724 Chr.14: 74101299-74101299 on Build GRCh38
rs1034713415 Chr.14: 74101213-74101213 on Build GRCh38
rs939330695 Chr.14: 74101122-74101122 on Build GRCh38
rs745462464 Chr.14: 74101175-74101175 on Build GRCh38
rs762896596 Chr.14: 74101268-74101286 on Build GRCh38
rs775385431 Chr.14: 74101189-74101189 on Build GRCh38
rs751337129 Chr.14: 74101289-74101308 on Build GRCh38
rs374818493 Chr.14: 74101104-74101104 on Build GRCh38
rs9743467 Chr.14: 74101223-74101223 on Build GRCh38
rs200462903 Chr.14: 74101174-74101174 on Build GRCh38
rs74431323 Chr.14: 74101386-74101386 on Build GRCh38

View Full Product Details