Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 74199599-74200096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIN52
Entrez Gene ID
91750
Gene Name
lin-52 DREAM MuvB core complex component
Gene Aliases
C14orf46, c14_5549
UniGene
Hs.612866
Gene Chromosome Location
Chr.14: 74084831-74201255 on Build GRCh38
Transcripts
NM_001024674.2 XM_017021763.1

SNP Detail

SNP ID Chromosome Location
rs1019964612 Chr.14: 74199896-74199896 on Build GRCh38
rs140314407 Chr.14: 74199665-74199665 on Build GRCh38
rs573495817 Chr.14: 74199961-74199961 on Build GRCh38
rs1019373301 Chr.14: 74199775-74199775 on Build GRCh38
rs941465599 Chr.14: 74199699-74199699 on Build GRCh38
rs988134819 Chr.14: 74199872-74199872 on Build GRCh38
rs951307865 Chr.14: 74199723-74199723 on Build GRCh38
rs949579996 Chr.14: 74199924-74199924 on Build GRCh38
rs925341988 Chr.14: 74199857-74199857 on Build GRCh38
rs1055541605 Chr.14: 74199997-74199997 on Build GRCh38
rs542883616 Chr.14: 74199934-74199934 on Build GRCh38
rs1046700297 Chr.14: 74200070-74200070 on Build GRCh38
rs897171641 Chr.14: 74199752-74199752 on Build GRCh38
rs752422280 Chr.14: 74200059-74200059 on Build GRCh38
rs770141123 Chr.14: 74199645-74199645 on Build GRCh38
rs564072323 Chr.14: 74200055-74200055 on Build GRCh38
rs949995541 Chr.14: 74199794-74199794 on Build GRCh38
rs1025966522 Chr.14: 74199691-74199691 on Build GRCh38
rs930992868 Chr.14: 74200048-74200048 on Build GRCh38
rs1001244625 Chr.14: 74199635-74199635 on Build GRCh38
rs1037094118 Chr.14: 74199732-74199732 on Build GRCh38
rs964100224 Chr.14: 74199790-74199790 on Build GRCh38
rs145234008 Chr.14: 74199689-74199689 on Build GRCh38
rs887595 Chr.14: 74199938-74199938 on Build GRCh38
rs1001448813 Chr.14: 74199833-74199833 on Build GRCh38
rs5809655 Chr.14: 74199869-74199870 on Build GRCh38
rs182847294 Chr.14: 74199923-74199923 on Build GRCh38
rs965692726 Chr.14: 74199915-74199915 on Build GRCh38
rs773492477 Chr.14: 74199662-74199662 on Build GRCh38
rs562416329 Chr.14: 74199895-74199895 on Build GRCh38
rs1033290338 Chr.14: 74199858-74199858 on Build GRCh38
rs937105104 Chr.14: 74199996-74199996 on Build GRCh38
rs752068026 Chr.14: 74199660-74199660 on Build GRCh38
rs7154851 Chr.14: 74199862-74199862 on Build GRCh38
rs756512457 Chr.14: 74199891-74199891 on Build GRCh38
rs1055505645 Chr.14: 74199647-74199647 on Build GRCh38
rs975466657 Chr.14: 74199803-74199805 on Build GRCh38
rs922824343 Chr.14: 74199622-74199622 on Build GRCh38
rs191069891 Chr.14: 74199781-74199781 on Build GRCh38
rs766750931 Chr.14: 74199707-74199707 on Build GRCh38
rs898186446 Chr.14: 74200021-74200021 on Build GRCh38
rs910037834 Chr.14: 74199679-74199679 on Build GRCh38
rs1008031141 Chr.14: 74199744-74199744 on Build GRCh38
rs910059690 Chr.14: 74200032-74200032 on Build GRCh38
rs905816000 Chr.14: 74199817-74199817 on Build GRCh38
rs1045678070 Chr.14: 74199809-74199809 on Build GRCh38
rs764305131 Chr.14: 74199627-74199627 on Build GRCh38
rs750807792 Chr.14: 74199870-74199870 on Build GRCh38
rs545677575 Chr.14: 74200018-74200018 on Build GRCh38

View Full Product Details