Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 74261302-74261810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VSX2
Entrez Gene ID
338917
Gene Name
visual system homeobox 2
Gene Aliases
CHX10, HOX10, MCOP2, MCOPCB3, RET1
UniGene
Hs.449771
Gene Chromosome Location
Chr.14: 74239472-74262738 on Build GRCh38
Transcripts
NM_182894.2

SNP Detail

SNP ID Chromosome Location
rs557664123 Chr.14: 74261357-74261357 on Build GRCh38
rs996023682 Chr.14: 74261558-74261558 on Build GRCh38
rs924306526 Chr.14: 74261483-74261483 on Build GRCh38
rs150646795 Chr.14: 74261651-74261651 on Build GRCh38
rs954193363 Chr.14: 74261754-74261754 on Build GRCh38
rs1039826358 Chr.14: 74261514-74261514 on Build GRCh38
rs931746723 Chr.14: 74261475-74261475 on Build GRCh38
rs73309254 Chr.14: 74261374-74261374 on Build GRCh38
rs180856135 Chr.14: 74261394-74261394 on Build GRCh38
rs45556237 Chr.14: 74261589-74261589 on Build GRCh38
rs189778670 Chr.14: 74261335-74261335 on Build GRCh38
rs74694887 Chr.14: 74261738-74261738 on Build GRCh38
rs887144105 Chr.14: 74261588-74261588 on Build GRCh38
rs886050740 Chr.14: 74261694-74261694 on Build GRCh38
rs748532333 Chr.14: 74261756-74261756 on Build GRCh38
rs978522430 Chr.14: 74261429-74261429 on Build GRCh38
rs951398968 Chr.14: 74261364-74261364 on Build GRCh38
rs538170309 Chr.14: 74261553-74261553 on Build GRCh38
rs565478331 Chr.14: 74261404-74261404 on Build GRCh38
rs758472746 Chr.14: 74261383-74261383 on Build GRCh38
rs781492325 Chr.14: 74261693-74261693 on Build GRCh38
rs934341446 Chr.14: 74261494-74261494 on Build GRCh38
rs888475579 Chr.14: 74261629-74261629 on Build GRCh38
rs1056975167 Chr.14: 74261579-74261579 on Build GRCh38
rs45608043 Chr.14: 74261774-74261774 on Build GRCh38
rs968574379 Chr.14: 74261684-74261684 on Build GRCh38
rs968397429 Chr.14: 74261785-74261785 on Build GRCh38
rs1028510389 Chr.14: 74261749-74261749 on Build GRCh38
rs12588074 Chr.14: 74261498-74261498 on Build GRCh38
rs959914565 Chr.14: 74261626-74261626 on Build GRCh38
rs140678898 Chr.14: 74261701-74261701 on Build GRCh38
rs1027051676 Chr.14: 74261610-74261610 on Build GRCh38
rs938559899 Chr.14: 74261560-74261560 on Build GRCh38
rs551300880 Chr.14: 74261507-74261507 on Build GRCh38
rs955907677 Chr.14: 74261689-74261689 on Build GRCh38
rs1039917771 Chr.14: 74261755-74261755 on Build GRCh38
rs944225217 Chr.14: 74261505-74261505 on Build GRCh38
rs1004569826 Chr.14: 74261367-74261367 on Build GRCh38
rs563564973 Chr.14: 74261349-74261349 on Build GRCh38
rs557561819 Chr.14: 74261761-74261761 on Build GRCh38
rs1026999331 Chr.14: 74261361-74261361 on Build GRCh38
rs1048463906 Chr.14: 74261585-74261585 on Build GRCh38
rs1015834054 Chr.14: 74261375-74261375 on Build GRCh38
rs976943890 Chr.14: 74261403-74261403 on Build GRCh38
rs751673499 Chr.14: 74261502-74261502 on Build GRCh38
rs978957287 Chr.14: 74261686-74261686 on Build GRCh38
rs1012773672 Chr.14: 74261636-74261636 on Build GRCh38
rs944317830 Chr.14: 74261703-74261703 on Build GRCh38
rs985821553 Chr.14: 74261477-74261486 on Build GRCh38
rs899991726 Chr.14: 74261527-74261527 on Build GRCh38
rs977380848 Chr.14: 74261721-74261721 on Build GRCh38
rs559754393 Chr.14: 74261368-74261368 on Build GRCh38
rs185981255 Chr.14: 74261593-74261593 on Build GRCh38
rs915675131 Chr.14: 74261399-74261399 on Build GRCh38
rs924345564 Chr.14: 74261688-74261688 on Build GRCh38
rs574115764 Chr.14: 74261350-74261350 on Build GRCh38

View Full Product Details