Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 76865885-76866390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC74A
Entrez Gene ID
145497
Gene Name
leucine rich repeat containing 74A
Gene Aliases
C14orf166B, LRRC74
UniGene
Hs.147276
Gene Chromosome Location
Chr.14: 76826370-76870302 on Build GRCh38
Transcripts
NM_194287.2 XM_011536479.1 XM_011536483.1 XM_017021032.1 XM_017021033.1 XM_017021034.1 XM_017021035.1 XM_017021036.1 XM_017021037.1 XM_017021038.1 XM_017021039.1 XM_017021040.1 XM_017021041.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4524252 c.1261G>A p.D421N Chr.14: 76865977-76865977 on Build GRCh38
4148265 c.1349T>C p.V450A Chr.14: 76866065-76866065 on Build GRCh38
2139078 c.1341C>T p.F447F Chr.14: 76866057-76866057 on Build GRCh38
6435538 c.1334A>G p.D445G Chr.14: 76866050-76866050 on Build GRCh38
958092 c.1267C>T p.H423Y Chr.14: 76865983-76865983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747103607 Chr.14: 76866089-76866089 on Build GRCh38
rs948162044 Chr.14: 76866154-76866154 on Build GRCh38
rs1018456157 Chr.14: 76865913-76865913 on Build GRCh38
rs755318785 Chr.14: 76865972-76865972 on Build GRCh38
rs906755442 Chr.14: 76866212-76866212 on Build GRCh38
rs1053911800 Chr.14: 76866112-76866112 on Build GRCh38
rs574802083 Chr.14: 76865916-76865916 on Build GRCh38
rs140195914 Chr.14: 76865958-76865958 on Build GRCh38
rs756695494 Chr.14: 76865940-76865940 on Build GRCh38
rs1882844 Chr.14: 76866024-76866024 on Build GRCh38
rs763731895 Chr.14: 76865926-76865926 on Build GRCh38
rs756046626 Chr.14: 76866052-76866052 on Build GRCh38
rs777064100 Chr.14: 76866101-76866101 on Build GRCh38
rs981443858 Chr.14: 76866198-76866198 on Build GRCh38
rs868286524 Chr.14: 76866358-76866358 on Build GRCh38
rs995697051 Chr.14: 76866136-76866136 on Build GRCh38
rs947205106 Chr.14: 76866337-76866337 on Build GRCh38
rs13379451 Chr.14: 76866206-76866206 on Build GRCh38
rs937442972 Chr.14: 76865973-76865973 on Build GRCh38
rs761423172 Chr.14: 76866118-76866118 on Build GRCh38
rs202056854 Chr.14: 76865998-76865998 on Build GRCh38
rs749927887 Chr.14: 76866046-76866046 on Build GRCh38
rs1026660336 Chr.14: 76866336-76866336 on Build GRCh38
rs766336374 Chr.14: 76866053-76866053 on Build GRCh38
rs779265325 Chr.14: 76865974-76865974 on Build GRCh38
rs538205987 Chr.14: 76866245-76866245 on Build GRCh38
rs985270432 Chr.14: 76866041-76866041 on Build GRCh38
rs777750982 Chr.14: 76866077-76866077 on Build GRCh38
rs7160583 Chr.14: 76866065-76866065 on Build GRCh38
rs914414521 Chr.14: 76865993-76865993 on Build GRCh38
rs777387750 Chr.14: 76866152-76866152 on Build GRCh38
rs748253194 Chr.14: 76865994-76865994 on Build GRCh38
rs962919695 Chr.14: 76865925-76865925 on Build GRCh38
rs773789392 Chr.14: 76865997-76865997 on Build GRCh38
rs755962261 Chr.14: 76865956-76865956 on Build GRCh38
rs879507563 Chr.14: 76866244-76866244 on Build GRCh38
rs111669209 Chr.14: 76866226-76866226 on Build GRCh38
rs772299913 Chr.14: 76866034-76866039 on Build GRCh38
rs754765913 Chr.14: 76866058-76866058 on Build GRCh38
rs766891016 Chr.14: 76866045-76866045 on Build GRCh38
rs763746232 Chr.14: 76866023-76866023 on Build GRCh38
rs974251809 Chr.14: 76865981-76865981 on Build GRCh38
rs912441595 Chr.14: 76866060-76866060 on Build GRCh38
rs895239979 Chr.14: 76866142-76866142 on Build GRCh38
rs192440024 Chr.14: 76865944-76865944 on Build GRCh38
rs1055594811 Chr.14: 76865986-76865986 on Build GRCh38
rs560484499 Chr.14: 76865947-76865947 on Build GRCh38
rs770102189 Chr.14: 76866105-76866105 on Build GRCh38
rs72721328 Chr.14: 76866102-76866102 on Build GRCh38
rs748343191 Chr.14: 76866064-76866064 on Build GRCh38
rs753748442 Chr.14: 76866057-76866057 on Build GRCh38
rs747517137 Chr.14: 76866009-76866009 on Build GRCh38
rs771394586 Chr.14: 76866092-76866092 on Build GRCh38
rs375364160 Chr.14: 76865949-76865949 on Build GRCh38
rs775993218 Chr.14: 76866103-76866104 on Build GRCh38
rs781108697 Chr.14: 76866062-76866062 on Build GRCh38
rs953801247 Chr.14: 76866346-76866346 on Build GRCh38
rs770638863 Chr.14: 76866231-76866231 on Build GRCh38
rs896851133 Chr.14: 76866256-76866256 on Build GRCh38
rs201148819 Chr.14: 76866015-76866015 on Build GRCh38
rs985239151 Chr.14: 76866352-76866352 on Build GRCh38
rs564156258 Chr.14: 76866169-76866169 on Build GRCh38
rs771980956 Chr.14: 76865995-76865995 on Build GRCh38
rs369516926 Chr.14: 76865927-76865927 on Build GRCh38
rs779766401 Chr.14: 76865957-76865957 on Build GRCh38
rs759876913 Chr.14: 76866021-76866021 on Build GRCh38
rs754207024 Chr.14: 76865918-76865918 on Build GRCh38
rs936752277 Chr.14: 76866110-76866110 on Build GRCh38
rs535945793 Chr.14: 76866126-76866126 on Build GRCh38
rs762633758 Chr.14: 76865919-76865919 on Build GRCh38
rs962865950 Chr.14: 76866131-76866131 on Build GRCh38
rs774016479 Chr.14: 76866116-76866116 on Build GRCh38
rs771437650 Chr.14: 76866010-76866010 on Build GRCh38
rs201329545 Chr.14: 76866063-76866063 on Build GRCh38
rs914700168 Chr.14: 76866311-76866311 on Build GRCh38
rs761414654 Chr.14: 76866040-76866040 on Build GRCh38

View Full Product Details