Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77115193-77115690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIPC
Entrez Gene ID
85457
Gene Name
CLOCK interacting pacemaker
Gene Aliases
KIAA1737
UniGene
Hs.709066
Gene Chromosome Location
Chr.14: 77098235-77117287 on Build GRCh38
Transcripts
NM_033426.2

SNP Detail

SNP ID Chromosome Location
rs949772641 Chr.14: 77115609-77115609 on Build GRCh38
rs909548726 Chr.14: 77115562-77115562 on Build GRCh38
rs988644543 Chr.14: 77115296-77115296 on Build GRCh38
rs541836668 Chr.14: 77115275-77115275 on Build GRCh38
rs778890107 Chr.14: 77115256-77115257 on Build GRCh38
rs373374123 Chr.14: 77115628-77115628 on Build GRCh38
rs796778696 Chr.14: 77115242-77115243 on Build GRCh38
rs939676009 Chr.14: 77115569-77115569 on Build GRCh38
rs1002455357 Chr.14: 77115245-77115245 on Build GRCh38
rs531685115 Chr.14: 77115241-77115242 on Build GRCh38
rs557310155 Chr.14: 77115489-77115489 on Build GRCh38
rs1032567399 Chr.14: 77115655-77115655 on Build GRCh38
rs865855258 Chr.14: 77115348-77115348 on Build GRCh38
rs974397637 Chr.14: 77115438-77115438 on Build GRCh38
rs1046638369 Chr.14: 77115625-77115625 on Build GRCh38
rs189119330 Chr.14: 77115323-77115323 on Build GRCh38
rs1001085758 Chr.14: 77115608-77115608 on Build GRCh38
rs75764539 Chr.14: 77115242-77115242 on Build GRCh38
rs78012095 Chr.14: 77115255-77115255 on Build GRCh38
rs1042341706 Chr.14: 77115537-77115537 on Build GRCh38
rs878978012 Chr.14: 77115244-77115244 on Build GRCh38
rs432679 Chr.14: 77115452-77115452 on Build GRCh38
rs187182042 Chr.14: 77115474-77115474 on Build GRCh38
rs116726883 Chr.14: 77115485-77115485 on Build GRCh38
rs902530590 Chr.14: 77115595-77115595 on Build GRCh38
rs775676676 Chr.14: 77115553-77115553 on Build GRCh38
rs969660909 Chr.14: 77115243-77115243 on Build GRCh38
rs770783125 Chr.14: 77115522-77115523 on Build GRCh38
rs796893093 Chr.14: 77115255-77115255 on Build GRCh38
rs959810106 Chr.14: 77115244-77115245 on Build GRCh38
rs543309118 Chr.14: 77115652-77115656 on Build GRCh38
rs879227721 Chr.14: 77115242-77115243 on Build GRCh38
rs58321309 Chr.14: 77115255-77115256 on Build GRCh38
rs932610693 Chr.14: 77115493-77115493 on Build GRCh38
rs768014783 Chr.14: 77115661-77115661 on Build GRCh38
rs1002875250 Chr.14: 77115651-77115651 on Build GRCh38
rs79606373 Chr.14: 77115241-77115241 on Build GRCh38
rs551284168 Chr.14: 77115398-77115398 on Build GRCh38
rs961247710 Chr.14: 77115376-77115376 on Build GRCh38
rs895427728 Chr.14: 77115621-77115621 on Build GRCh38
rs182505212 Chr.14: 77115330-77115330 on Build GRCh38
rs950827035 Chr.14: 77115261-77115261 on Build GRCh38
rs908218004 Chr.14: 77115351-77115351 on Build GRCh38
rs528872443 Chr.14: 77115262-77115262 on Build GRCh38
rs11342878 Chr.14: 77115242-77115242 on Build GRCh38
rs912631418 Chr.14: 77115499-77115499 on Build GRCh38
rs965413665 Chr.14: 77115344-77115344 on Build GRCh38
rs895593786 Chr.14: 77115607-77115607 on Build GRCh38
rs140172245 Chr.14: 77115626-77115626 on Build GRCh38
rs976585243 Chr.14: 77115396-77115396 on Build GRCh38
rs1049612335 Chr.14: 77115490-77115490 on Build GRCh38
rs971158830 Chr.14: 77115334-77115334 on Build GRCh38
rs565423832 Chr.14: 77115333-77115333 on Build GRCh38
rs944146820 Chr.14: 77115516-77115516 on Build GRCh38
rs878855533 Chr.14: 77115242-77115244 on Build GRCh38
rs1025166564 Chr.14: 77115633-77115633 on Build GRCh38
rs1037208597 Chr.14: 77115573-77115573 on Build GRCh38
rs958030316 Chr.14: 77115263-77115263 on Build GRCh38
rs534759736 Chr.14: 77115397-77115397 on Build GRCh38

View Full Product Details