Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77730805-77731318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNW1
Entrez Gene ID
22938
Gene Name
SNW domain containing 1
Gene Aliases
Bx42, NCOA-62, PRPF45, Prp45, SKIIP, SKIP
UniGene
Hs.445498
Gene Chromosome Location
Chr.14: 77717599-77761220 on Build GRCh38
Transcripts
NM_001318844.1 NM_012245.2 XM_005267413.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7088038 c.1009G>A p.G337R Chr.14: 77731012-77731012 on Build GRCh38
6786367 c.1032_1033insA p.E345fs*7 Chr.14: 77730988-77730989 on Build GRCh38
4884270 c.1010G>T p.G337V Chr.14: 77731011-77731011 on Build GRCh38
3377471 c.949A>C p.K317Q Chr.14: 77731072-77731072 on Build GRCh38
5445656 c.999G>A p.E333E Chr.14: 77731022-77731022 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761167646 Chr.14: 77731046-77731046 on Build GRCh38
rs1002486026 Chr.14: 77731243-77731243 on Build GRCh38
rs754181399 Chr.14: 77731160-77731160 on Build GRCh38
rs368176828 Chr.14: 77731147-77731147 on Build GRCh38
rs759637791 Chr.14: 77731155-77731156 on Build GRCh38
rs568170099 Chr.14: 77730842-77730842 on Build GRCh38
rs2111829 Chr.14: 77730940-77730940 on Build GRCh38
rs773541873 Chr.14: 77730955-77730955 on Build GRCh38
rs1029164713 Chr.14: 77730957-77730957 on Build GRCh38
rs750300573 Chr.14: 77731125-77731125 on Build GRCh38
rs918310122 Chr.14: 77731182-77731182 on Build GRCh38
rs905737376 Chr.14: 77731229-77731229 on Build GRCh38
rs190433273 Chr.14: 77730877-77730877 on Build GRCh38
rs1033904122 Chr.14: 77731261-77731262 on Build GRCh38
rs778945389 Chr.14: 77731166-77731166 on Build GRCh38
rs534565504 Chr.14: 77730938-77730938 on Build GRCh38
rs572782998 Chr.14: 77731266-77731266 on Build GRCh38
rs201134818 Chr.14: 77731106-77731106 on Build GRCh38
rs929830797 Chr.14: 77730903-77730903 on Build GRCh38
rs919320617 Chr.14: 77730859-77730859 on Build GRCh38
rs762493739 Chr.14: 77731200-77731200 on Build GRCh38
rs987774669 Chr.14: 77731103-77731103 on Build GRCh38
rs756024662 Chr.14: 77731144-77731144 on Build GRCh38
rs114763 Chr.14: 77731022-77731022 on Build GRCh38
rs576598900 Chr.14: 77731167-77731167 on Build GRCh38
rs778383955 Chr.14: 77731170-77731170 on Build GRCh38
rs982990011 Chr.14: 77730917-77730917 on Build GRCh38
rs992122335 Chr.14: 77731287-77731287 on Build GRCh38
rs772647551 Chr.14: 77730952-77730952 on Build GRCh38
rs755257622 Chr.14: 77731161-77731161 on Build GRCh38
rs775170813 Chr.14: 77730998-77730998 on Build GRCh38
rs1022919592 Chr.14: 77731225-77731225 on Build GRCh38
rs758369802 Chr.14: 77731169-77731169 on Build GRCh38
rs577144121 Chr.14: 77730976-77730976 on Build GRCh38
rs61742193 Chr.14: 77731124-77731124 on Build GRCh38
rs1048665844 Chr.14: 77731214-77731214 on Build GRCh38
rs895804342 Chr.14: 77731275-77731275 on Build GRCh38
rs542404306 Chr.14: 77731197-77731197 on Build GRCh38
rs763658256 Chr.14: 77731028-77731028 on Build GRCh38
rs562214000 Chr.14: 77731236-77731236 on Build GRCh38
rs185390484 Chr.14: 77730863-77730863 on Build GRCh38
rs937249892 Chr.14: 77731265-77731265 on Build GRCh38
rs763981477 Chr.14: 77731176-77731179 on Build GRCh38
rs747490046 Chr.14: 77731172-77731172 on Build GRCh38
rs751081554 Chr.14: 77731032-77731032 on Build GRCh38
rs761132528 Chr.14: 77730967-77730967 on Build GRCh38
rs1009745044 Chr.14: 77731205-77731205 on Build GRCh38
rs968792962 Chr.14: 77730871-77730871 on Build GRCh38

View Full Product Details